Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150805772-150806370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSK
Entrez Gene ID
1513
Gene Name
cathepsin K
Gene Aliases
CTS02, CTSO, CTSO1, CTSO2, PKND, PYCD
UniGene
Hs.632466
Gene Chromosome Location
Chr.1: 150796208-150808441 on Build GRCh38
Transcripts
NM_000396.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5754187 c.151G>A p.E51K Chr.1: 150806194-150806194 on Build GRCh38
5769502 c.567C>T p.V189V Chr.1: 150805870-150805870 on Build GRCh38
2182539 c.152A>G p.E51G Chr.1: 150806193-150806193 on Build GRCh38
5795527 c.541C>T p.R181* Chr.1: 150805896-150805896 on Build GRCh38
895613 c.380T>C p.V127A Chr.1: 150805880-150805880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995485377 Chr.1: 150806107-150806107 on Build GRCh38
rs1057516725 Chr.1: 150805864-150805865 on Build GRCh38
rs587663163 Chr.1: 150806225-150806225 on Build GRCh38
rs201274159 Chr.1: 150805821-150805821 on Build GRCh38
rs765510109 Chr.1: 150806096-150806096 on Build GRCh38
rs145693431 Chr.1: 150806016-150806016 on Build GRCh38
rs200870056 Chr.1: 150806208-150806208 on Build GRCh38
rs776050755 Chr.1: 150805814-150805814 on Build GRCh38
rs747664835 Chr.1: 150806082-150806082 on Build GRCh38
rs74315305 Chr.1: 150806109-150806109 on Build GRCh38
rs912531584 Chr.1: 150805978-150805978 on Build GRCh38
rs184046317 Chr.1: 150806338-150806338 on Build GRCh38
rs781761916 Chr.1: 150806149-150806149 on Build GRCh38
rs193045250 Chr.1: 150806134-150806134 on Build GRCh38
rs772730842 Chr.1: 150805919-150805919 on Build GRCh38
rs1037452561 Chr.1: 150806071-150806071 on Build GRCh38
rs200621697 Chr.1: 150806084-150806084 on Build GRCh38
rs200337167 Chr.1: 150805855-150805855 on Build GRCh38
rs764990767 Chr.1: 150806263-150806263 on Build GRCh38
rs746005364 Chr.1: 150805955-150805955 on Build GRCh38
rs780202604 Chr.1: 150806132-150806132 on Build GRCh38
rs770173800 Chr.1: 150805837-150805842 on Build GRCh38
rs748594586 Chr.1: 150806161-150806161 on Build GRCh38
rs753642215 Chr.1: 150806080-150806080 on Build GRCh38
rs1057516587 Chr.1: 150806226-150806226 on Build GRCh38
rs201446793 Chr.1: 150806176-150806176 on Build GRCh38
rs142306783 Chr.1: 150805903-150805903 on Build GRCh38
rs753591279 Chr.1: 150805922-150805922 on Build GRCh38
rs1034096210 Chr.1: 150805825-150805825 on Build GRCh38
rs907283349 Chr.1: 150805969-150805969 on Build GRCh38
rs199655884 Chr.1: 150805826-150805826 on Build GRCh38
rs373300320 Chr.1: 150806036-150806036 on Build GRCh38
rs759107967 Chr.1: 150805896-150805896 on Build GRCh38
rs137874306 Chr.1: 150805938-150805938 on Build GRCh38
rs201667695 Chr.1: 150805909-150805909 on Build GRCh38
rs777647239 Chr.1: 150805914-150805914 on Build GRCh38
rs779047231 Chr.1: 150805948-150805948 on Build GRCh38
rs760699631 Chr.1: 150806053-150806053 on Build GRCh38
rs771607501 Chr.1: 150805992-150805994 on Build GRCh38
rs936779452 Chr.1: 150806187-150806187 on Build GRCh38
rs766065317 Chr.1: 150805885-150805885 on Build GRCh38
rs752649562 Chr.1: 150805902-150805902 on Build GRCh38
rs745520467 Chr.1: 150805977-150805978 on Build GRCh38
rs776743489 Chr.1: 150806187-150806187 on Build GRCh38
rs188468815 Chr.1: 150805852-150805852 on Build GRCh38
rs955729641 Chr.1: 150806303-150806303 on Build GRCh38
rs768992778 Chr.1: 150805986-150805986 on Build GRCh38
rs199909074 Chr.1: 150806205-150806205 on Build GRCh38
rs928459024 Chr.1: 150806343-150806343 on Build GRCh38
rs587742006 Chr.1: 150805965-150805965 on Build GRCh38
rs774922287 Chr.1: 150806024-150806026 on Build GRCh38
rs112299095 Chr.1: 150806018-150806018 on Build GRCh38
rs750165917 Chr.1: 150806271-150806271 on Build GRCh38
rs764148445 Chr.1: 150806213-150806213 on Build GRCh38
rs764168526 Chr.1: 150806186-150806187 on Build GRCh38
rs769029170 Chr.1: 150805853-150805853 on Build GRCh38
rs199802458 Chr.1: 150806040-150806040 on Build GRCh38
rs1057517263 Chr.1: 150806101-150806101 on Build GRCh38
rs980350501 Chr.1: 150806334-150806334 on Build GRCh38
rs780461752 Chr.1: 150805959-150805959 on Build GRCh38
rs748453667 Chr.1: 150806296-150806296 on Build GRCh38
rs760640027 Chr.1: 150806145-150806155 on Build GRCh38
rs892638977 Chr.1: 150806278-150806278 on Build GRCh38
rs762714840 Chr.1: 150805857-150805857 on Build GRCh38
rs989762442 Chr.1: 150806315-150806315 on Build GRCh38
rs774816040 Chr.1: 150806051-150806051 on Build GRCh38
rs750609110 Chr.1: 150806110-150806110 on Build GRCh38
rs773943327 Chr.1: 150805970-150805971 on Build GRCh38
rs966197225 Chr.1: 150806252-150806252 on Build GRCh38
rs200742781 Chr.1: 150805958-150805958 on Build GRCh38
rs371277428 Chr.1: 150806209-150806209 on Build GRCh38
rs201028711 Chr.1: 150806030-150806030 on Build GRCh38
rs199642127 Chr.1: 150805936-150805936 on Build GRCh38
rs201962431 Chr.1: 150805894-150805894 on Build GRCh38
rs1057516839 Chr.1: 150805859-150805859 on Build GRCh38
rs777061480 Chr.1: 150805992-150805992 on Build GRCh38
rs948599693 Chr.1: 150806326-150806326 on Build GRCh38
rs143688533 Chr.1: 150806129-150806129 on Build GRCh38
rs74315306 Chr.1: 150806191-150806191 on Build GRCh38
rs755526593 Chr.1: 150806140-150806140 on Build GRCh38
rs761654710 Chr.1: 150806215-150806220 on Build GRCh38
rs770769840 Chr.1: 150806005-150806005 on Build GRCh38
rs199976601 Chr.1: 150806265-150806265 on Build GRCh38
rs202078506 Chr.1: 150806328-150806328 on Build GRCh38
rs202035896 Chr.1: 150806251-150806251 on Build GRCh38
rs374906814 Chr.1: 150806319-150806319 on Build GRCh38
rs945614436 Chr.1: 150806309-150806309 on Build GRCh38
rs1054278735 Chr.1: 150805898-150805898 on Build GRCh38
rs1002925379 Chr.1: 150806070-150806070 on Build GRCh38
rs201008554 Chr.1: 150806264-150806264 on Build GRCh38
rs755849677 Chr.1: 150805854-150805854 on Build GRCh38
rs879232242 Chr.1: 150806207-150806207 on Build GRCh38
rs201308534 Chr.1: 150805866-150805866 on Build GRCh38
rs587775407 Chr.1: 150805926-150805926 on Build GRCh38
rs762212949 Chr.1: 150805997-150805997 on Build GRCh38
rs587708639 Chr.1: 150806162-150806162 on Build GRCh38
rs763870328 Chr.1: 150806061-150806061 on Build GRCh38
rs761635015 Chr.1: 150806239-150806239 on Build GRCh38
rs771586535 Chr.1: 150806206-150806206 on Build GRCh38
rs765141224 Chr.1: 150806230-150806230 on Build GRCh38

View Full Product Details