Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36233129-36233566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL2
Entrez Gene ID
23780
Gene Name
apolipoprotein L2
Gene Aliases
APOL-II, APOL3
UniGene
Hs.474740
Gene Chromosome Location
Chr.22: 36226209-36239954 on Build GRCh38
Transcripts
NM_030882.3 NM_145637.2 XM_011530074.1 XM_011530075.1 XM_011530076.2 XM_011530077.2 XM_011530078.1 XM_017028724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5859280 c.7C>T p.P3S Chr.22: 36233156-36233156 on Build GRCh38
5057794 c.2T>C p.M1T Chr.22: 36233161-36233161 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781262750 Chr.22: 36233189-36233189 on Build GRCh38
rs766844393 Chr.22: 36233171-36233171 on Build GRCh38
rs2010499 Chr.22: 36233420-36233420 on Build GRCh38
rs6000212 Chr.22: 36233262-36233262 on Build GRCh38
rs780192606 Chr.22: 36233367-36233367 on Build GRCh38
rs532980977 Chr.22: 36233405-36233405 on Build GRCh38
rs535199787 Chr.22: 36233459-36233459 on Build GRCh38
rs550561462 Chr.22: 36233373-36233373 on Build GRCh38
rs778152672 Chr.22: 36233441-36233442 on Build GRCh38
rs765544989 Chr.22: 36233251-36233251 on Build GRCh38
rs777062113 Chr.22: 36233247-36233247 on Build GRCh38
rs763424700 Chr.22: 36233265-36233265 on Build GRCh38
rs751867133 Chr.22: 36233272-36233272 on Build GRCh38
rs530946505 Chr.22: 36233284-36233284 on Build GRCh38
rs201950146 Chr.22: 36233177-36233177 on Build GRCh38
rs932499375 Chr.22: 36233506-36233506 on Build GRCh38
rs936153044 Chr.22: 36233401-36233401 on Build GRCh38
rs562037489 Chr.22: 36233261-36233261 on Build GRCh38
rs1024570646 Chr.22: 36233440-36233440 on Build GRCh38
rs755907908 Chr.22: 36233215-36233215 on Build GRCh38
rs570728336 Chr.22: 36233402-36233402 on Build GRCh38
rs924716308 Chr.22: 36233427-36233427 on Build GRCh38
rs878956888 Chr.22: 36233445-36233445 on Build GRCh38
rs747212160 Chr.22: 36233469-36233471 on Build GRCh38
rs985023202 Chr.22: 36233489-36233489 on Build GRCh38
rs762159429 Chr.22: 36233480-36233480 on Build GRCh38
rs1001553409 Chr.22: 36233454-36233454 on Build GRCh38
rs757355115 Chr.22: 36233282-36233282 on Build GRCh38
rs771624821 Chr.22: 36233245-36233245 on Build GRCh38
rs756146178 Chr.22: 36233355-36233355 on Build GRCh38
rs750348171 Chr.22: 36233286-36233286 on Build GRCh38
rs555063247 Chr.22: 36233467-36233467 on Build GRCh38
rs374380172 Chr.22: 36233196-36233196 on Build GRCh38
rs973561874 Chr.22: 36233279-36233279 on Build GRCh38
rs917500005 Chr.22: 36233356-36233356 on Build GRCh38
rs774097070 Chr.22: 36233241-36233241 on Build GRCh38
rs1050990892 Chr.22: 36233535-36233535 on Build GRCh38
rs149859426 Chr.22: 36233474-36233474 on Build GRCh38
rs775510430 Chr.22: 36233281-36233281 on Build GRCh38
rs765596236 Chr.22: 36233173-36233173 on Build GRCh38
rs772737650 Chr.22: 36233415-36233415 on Build GRCh38
rs764191642 Chr.22: 36233267-36233267 on Build GRCh38
rs1027851932 Chr.22: 36233297-36233297 on Build GRCh38
rs1014799067 Chr.22: 36233513-36233513 on Build GRCh38
rs754872731 Chr.22: 36233376-36233376 on Build GRCh38
rs369772901 Chr.22: 36233153-36233153 on Build GRCh38
rs764185135 Chr.22: 36233179-36233179 on Build GRCh38
rs958923909 Chr.22: 36233305-36233305 on Build GRCh38
rs1034416767 Chr.22: 36233464-36233464 on Build GRCh38
rs774371606 Chr.22: 36233475-36233475 on Build GRCh38
rs748924267 Chr.22: 36233222-36233222 on Build GRCh38
rs1054200551 Chr.22: 36233483-36233483 on Build GRCh38
rs566427736 Chr.22: 36233435-36233435 on Build GRCh38
rs546379454 Chr.22: 36233172-36233172 on Build GRCh38
rs528633842 Chr.22: 36233183-36233183 on Build GRCh38
rs1050107828 Chr.22: 36233351-36233351 on Build GRCh38
rs749009247 Chr.22: 36233369-36233369 on Build GRCh38
rs771501980 Chr.22: 36233411-36233411 on Build GRCh38
rs746322835 Chr.22: 36233418-36233418 on Build GRCh38
rs780029354 Chr.22: 36233219-36233219 on Build GRCh38
rs745815748 Chr.22: 36233191-36233191 on Build GRCh38
rs373430439 Chr.22: 36233184-36233184 on Build GRCh38
rs767658571 Chr.22: 36233494-36233494 on Build GRCh38
rs768175357 Chr.22: 36233234-36233234 on Build GRCh38
rs940654413 Chr.22: 36233242-36233242 on Build GRCh38
rs746369273 Chr.22: 36233354-36233354 on Build GRCh38
rs185233543 Chr.22: 36233250-36233250 on Build GRCh38
rs907173021 Chr.22: 36233447-36233447 on Build GRCh38
rs1004296906 Chr.22: 36233217-36233217 on Build GRCh38
rs537456625 Chr.22: 36233523-36233523 on Build GRCh38
rs761439163 Chr.22: 36233160-36233160 on Build GRCh38

View Full Product Details