Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48177669-48178226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC25A
Entrez Gene ID
993
Gene Name
cell division cycle 25A
Gene Aliases
CDC25A2
UniGene
Hs.437705
Gene Chromosome Location
Chr.3: 48157178-48188658 on Build GRCh38
Transcripts
NM_001789.2 NM_201567.1 XM_006713434.2 XM_006713435.2 XM_011534316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
25656 c.652T>A p.F218I Chr.3: 48177886-48177886 on Build GRCh38
1045735 c.606A>C p.T202T Chr.3: 48177932-48177932 on Build GRCh38
274456 c.654C>T p.F218F Chr.3: 48177884-48177884 on Build GRCh38
7360361 c.634T>G p.S212A Chr.3: 48177904-48177904 on Build GRCh38
5910349 c.550C>T p.L184F Chr.3: 48177988-48177988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200689858 Chr.3: 48177965-48177965 on Build GRCh38
rs750466195 Chr.3: 48177848-48177849 on Build GRCh38
rs771901906 Chr.3: 48177830-48177831 on Build GRCh38
rs770631650 Chr.3: 48177814-48177814 on Build GRCh38
rs750597538 Chr.3: 48177872-48177872 on Build GRCh38
rs959971751 Chr.3: 48177813-48177813 on Build GRCh38
rs3731513 Chr.3: 48178047-48178047 on Build GRCh38
rs371829889 Chr.3: 48177933-48177933 on Build GRCh38
rs534972297 Chr.3: 48177833-48177834 on Build GRCh38
rs374368943 Chr.3: 48177871-48177871 on Build GRCh38
rs975405163 Chr.3: 48177958-48177958 on Build GRCh38
rs765902652 Chr.3: 48177983-48177983 on Build GRCh38
rs182709683 Chr.3: 48178111-48178111 on Build GRCh38
rs756419756 Chr.3: 48178114-48178114 on Build GRCh38
rs775697668 Chr.3: 48177817-48177817 on Build GRCh38
rs766671573 Chr.3: 48177884-48177884 on Build GRCh38
rs764469793 Chr.3: 48177838-48177838 on Build GRCh38
rs763164594 Chr.3: 48177996-48177996 on Build GRCh38
rs896062881 Chr.3: 48177769-48177769 on Build GRCh38
rs780939908 Chr.3: 48177916-48177916 on Build GRCh38
rs1014445964 Chr.3: 48178174-48178174 on Build GRCh38
rs201435339 Chr.3: 48177893-48177893 on Build GRCh38
rs777212214 Chr.3: 48177959-48177959 on Build GRCh38
rs767983385 Chr.3: 48178069-48178069 on Build GRCh38
rs763044930 Chr.3: 48178011-48178013 on Build GRCh38
rs758679809 Chr.3: 48177883-48177883 on Build GRCh38
rs754829912 Chr.3: 48177902-48177902 on Build GRCh38
rs1037856080 Chr.3: 48178080-48178081 on Build GRCh38
rs770949612 Chr.3: 48177806-48177809 on Build GRCh38
rs753232432 Chr.3: 48178103-48178103 on Build GRCh38
rs10582115 Chr.3: 48177832-48177833 on Build GRCh38
rs560372365 Chr.3: 48177755-48177755 on Build GRCh38
rs142184745 Chr.3: 48177953-48177953 on Build GRCh38
rs398062310 Chr.3: 48177847-48177848 on Build GRCh38
rs1004119399 Chr.3: 48177771-48177771 on Build GRCh38
rs368466847 Chr.3: 48177891-48177891 on Build GRCh38
rs761144705 Chr.3: 48177826-48177826 on Build GRCh38
rs778011954 Chr.3: 48178191-48178191 on Build GRCh38
rs149708323 Chr.3: 48177848-48177848 on Build GRCh38
rs9816850 Chr.3: 48177756-48177756 on Build GRCh38
rs867455827 Chr.3: 48177860-48177861 on Build GRCh38
rs748848895 Chr.3: 48177924-48177924 on Build GRCh38
rs796490720 Chr.3: 48177842-48177843 on Build GRCh38
rs777256066 Chr.3: 48177923-48177923 on Build GRCh38
rs937040749 Chr.3: 48178106-48178106 on Build GRCh38
rs766468519 Chr.3: 48177998-48177998 on Build GRCh38
rs190444887 Chr.3: 48177731-48177731 on Build GRCh38
rs756027300 Chr.3: 48178024-48178024 on Build GRCh38
rs767346063 Chr.3: 48178012-48178012 on Build GRCh38
rs752549847 Chr.3: 48178023-48178023 on Build GRCh38
rs1036008556 Chr.3: 48178120-48178120 on Build GRCh38
rs3731512 Chr.3: 48178079-48178079 on Build GRCh38
rs751020472 Chr.3: 48178033-48178033 on Build GRCh38
rs577829791 Chr.3: 48177733-48177733 on Build GRCh38
rs201467989 Chr.3: 48177926-48177926 on Build GRCh38
rs1040191507 Chr.3: 48177699-48177699 on Build GRCh38
rs899889546 Chr.3: 48178014-48178014 on Build GRCh38
rs896094228 Chr.3: 48178157-48178157 on Build GRCh38
rs756114919 Chr.3: 48177922-48177922 on Build GRCh38
rs1035384075 Chr.3: 48177773-48177773 on Build GRCh38
rs545238807 Chr.3: 48177739-48177739 on Build GRCh38
rs761926570 Chr.3: 48177849-48177849 on Build GRCh38
rs536089329 Chr.3: 48177831-48177834 on Build GRCh38
rs1008338240 Chr.3: 48177695-48177695 on Build GRCh38
rs1048284998 Chr.3: 48178053-48178053 on Build GRCh38
rs142974626 Chr.3: 48177847-48177847 on Build GRCh38
rs3731514 Chr.3: 48177831-48177832 on Build GRCh38
rs142219036 Chr.3: 48177914-48177915 on Build GRCh38
rs199526294 Chr.3: 48178006-48178006 on Build GRCh38
rs796171631 Chr.3: 48177836-48177837 on Build GRCh38
rs367727718 Chr.3: 48178025-48178025 on Build GRCh38
rs773402032 Chr.3: 48177987-48177987 on Build GRCh38
rs1020851978 Chr.3: 48177859-48177859 on Build GRCh38
rs766123347 Chr.3: 48177999-48177999 on Build GRCh38
rs764966111 Chr.3: 48177712-48177712 on Build GRCh38
rs907715952 Chr.3: 48178060-48178060 on Build GRCh38

View Full Product Details