Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112775037-112775571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1A
Entrez Gene ID
3552
Gene Name
interleukin 1 alpha
Gene Aliases
IL-1A, IL1, IL1-ALPHA, IL1F1
UniGene
Hs.1722
Gene Chromosome Location
Chr.2: 112773915-112785398 on Build GRCh38
Transcripts
NM_000575.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5594704 c.686G>A p.G229D Chr.2: 112775197-112775197 on Build GRCh38
3406795 c.813G>A p.A271A Chr.2: 112775070-112775070 on Build GRCh38
6153665 c.715C>A p.H239N Chr.2: 112775168-112775168 on Build GRCh38
6087373 c.623C>G p.P208R Chr.2: 112775260-112775260 on Build GRCh38
4084280 c.804A>C p.E268D Chr.2: 112775079-112775079 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56291885 Chr.2: 112775361-112775361 on Build GRCh38
rs970755764 Chr.2: 112775514-112775514 on Build GRCh38
rs922321806 Chr.2: 112775327-112775327 on Build GRCh38
rs1028915432 Chr.2: 112775342-112775342 on Build GRCh38
rs770987672 Chr.2: 112775270-112775270 on Build GRCh38
rs774643628 Chr.2: 112775271-112775271 on Build GRCh38
rs780982683 Chr.2: 112775208-112775208 on Build GRCh38
rs970618356 Chr.2: 112775190-112775190 on Build GRCh38
rs3783588 Chr.2: 112775070-112775070 on Build GRCh38
rs867638751 Chr.2: 112775093-112775093 on Build GRCh38
rs750076678 Chr.2: 112775165-112775165 on Build GRCh38
rs957816968 Chr.2: 112775272-112775272 on Build GRCh38
rs769836380 Chr.2: 112775239-112775239 on Build GRCh38
rs761403801 Chr.2: 112775148-112775148 on Build GRCh38
rs759804652 Chr.2: 112775282-112775282 on Build GRCh38
rs563801411 Chr.2: 112775133-112775133 on Build GRCh38
rs777778675 Chr.2: 112775250-112775250 on Build GRCh38
rs571034268 Chr.2: 112775471-112775471 on Build GRCh38
rs767850241 Chr.2: 112775123-112775123 on Build GRCh38
rs1006315696 Chr.2: 112775383-112775383 on Build GRCh38
rs55910084 Chr.2: 112775086-112775086 on Build GRCh38
rs1044124612 Chr.2: 112775127-112775127 on Build GRCh38
rs890468599 Chr.2: 112775359-112775359 on Build GRCh38
rs202006254 Chr.2: 112775160-112775160 on Build GRCh38
rs775826557 Chr.2: 112775306-112775306 on Build GRCh38
rs938378887 Chr.2: 112775210-112775210 on Build GRCh38
rs3783550 Chr.2: 112775308-112775308 on Build GRCh38
rs528482563 Chr.2: 112775501-112775501 on Build GRCh38
rs889368950 Chr.2: 112775381-112775381 on Build GRCh38
rs962165835 Chr.2: 112775170-112775170 on Build GRCh38
rs779788855 Chr.2: 112775194-112775194 on Build GRCh38
rs147177010 Chr.2: 112775182-112775182 on Build GRCh38
rs1037892235 Chr.2: 112775385-112775385 on Build GRCh38
rs926948361 Chr.2: 112775149-112775149 on Build GRCh38
rs772553599 Chr.2: 112775288-112775288 on Build GRCh38
rs1006259966 Chr.2: 112775080-112775080 on Build GRCh38
rs945165951 Chr.2: 112775307-112775307 on Build GRCh38
rs774537851 Chr.2: 112775073-112775073 on Build GRCh38
rs758077128 Chr.2: 112775188-112775188 on Build GRCh38
rs559448121 Chr.2: 112775498-112775498 on Build GRCh38
rs996049760 Chr.2: 112775321-112775321 on Build GRCh38
rs759945149 Chr.2: 112775101-112775101 on Build GRCh38
rs144254850 Chr.2: 112775258-112775258 on Build GRCh38
rs748154335 Chr.2: 112775231-112775231 on Build GRCh38
rs747708905 Chr.2: 112775176-112775176 on Build GRCh38
rs547629579 Chr.2: 112775407-112775407 on Build GRCh38
rs913615593 Chr.2: 112775301-112775301 on Build GRCh38
rs1050674590 Chr.2: 112775352-112775352 on Build GRCh38
rs926438913 Chr.2: 112775264-112775264 on Build GRCh38
rs886250113 Chr.2: 112775268-112775268 on Build GRCh38
rs753034784 Chr.2: 112775278-112775280 on Build GRCh38
rs993624986 Chr.2: 112775436-112775436 on Build GRCh38
rs368867049 Chr.2: 112775195-112775195 on Build GRCh38
rs933567628 Chr.2: 112775332-112775332 on Build GRCh38
rs13420770 Chr.2: 112775339-112775339 on Build GRCh38
rs764722067 Chr.2: 112775311-112775311 on Build GRCh38
rs754817324 Chr.2: 112775198-112775198 on Build GRCh38

View Full Product Details