Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96290222-96290753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG2B
Entrez Gene ID
55102
Gene Name
autophagy related 2B
Gene Aliases
C14orf103
UniGene
Hs.168241
Gene Chromosome Location
Chr.14: 96281258-96363341 on Build GRCh38
Transcripts
XM_006720187.2 XM_017021397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6606675 c.5810C>T p.A1937V Chr.14: 96290482-96290482 on Build GRCh38
5805399 c.5779A>T p.R1927* Chr.14: 96290513-96290513 on Build GRCh38
4828298 c.5778G>C p.Q1926H Chr.14: 96290514-96290514 on Build GRCh38
1678244 c.5759G>A p.R1920H Chr.14: 96290533-96290533 on Build GRCh38
6505546 c.5824G>A p.E1942K Chr.14: 96290468-96290468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779999611 Chr.14: 96290671-96290671 on Build GRCh38
rs371151129 Chr.14: 96290650-96290650 on Build GRCh38
rs746908291 Chr.14: 96290613-96290613 on Build GRCh38
rs756177157 Chr.14: 96290643-96290643 on Build GRCh38
rs758987076 Chr.14: 96290381-96290381 on Build GRCh38
rs988116854 Chr.14: 96290316-96290316 on Build GRCh38
rs188383620 Chr.14: 96290491-96290491 on Build GRCh38
rs537904133 Chr.14: 96290676-96290676 on Build GRCh38
rs1026095158 Chr.14: 96290683-96290683 on Build GRCh38
rs753216718 Chr.14: 96290361-96290361 on Build GRCh38
rs766550939 Chr.14: 96290569-96290569 on Build GRCh38
rs753293154 Chr.14: 96290448-96290448 on Build GRCh38
rs773129778 Chr.14: 96290411-96290411 on Build GRCh38
rs532440666 Chr.14: 96290709-96290709 on Build GRCh38
rs757167490 Chr.14: 96290617-96290617 on Build GRCh38
rs559999403 Chr.14: 96290287-96290287 on Build GRCh38
rs753801459 Chr.14: 96290578-96290578 on Build GRCh38
rs115346468 Chr.14: 96290631-96290631 on Build GRCh38
rs143845208 Chr.14: 96290508-96290508 on Build GRCh38
rs777722930 Chr.14: 96290497-96290497 on Build GRCh38
rs946787159 Chr.14: 96290362-96290362 on Build GRCh38
rs751856575 Chr.14: 96290462-96290462 on Build GRCh38
rs551790263 Chr.14: 96290687-96290687 on Build GRCh38
rs776463804 Chr.14: 96290524-96290524 on Build GRCh38
rs201940277 Chr.14: 96290457-96290457 on Build GRCh38
rs764507068 Chr.14: 96290454-96290454 on Build GRCh38
rs758374879 Chr.14: 96290583-96290583 on Build GRCh38
rs746750812 Chr.14: 96290502-96290502 on Build GRCh38
rs767842219 Chr.14: 96290545-96290545 on Build GRCh38
rs750537439 Chr.14: 96290482-96290482 on Build GRCh38
rs370916276 Chr.14: 96290540-96290540 on Build GRCh38
rs374314757 Chr.14: 96290558-96290558 on Build GRCh38
rs879220039 Chr.14: 96290350-96290350 on Build GRCh38
rs529084704 Chr.14: 96290357-96290357 on Build GRCh38
rs554295409 Chr.14: 96290728-96290728 on Build GRCh38
rs974884841 Chr.14: 96290329-96290329 on Build GRCh38
rs148578456 Chr.14: 96290289-96290289 on Build GRCh38
rs763064757 Chr.14: 96290413-96290413 on Build GRCh38
rs756258909 Chr.14: 96290484-96290484 on Build GRCh38
rs763518763 Chr.14: 96290534-96290534 on Build GRCh38
rs546580016 Chr.14: 96290260-96290260 on Build GRCh38
rs987228532 Chr.14: 96290323-96290323 on Build GRCh38
rs947222328 Chr.14: 96290479-96290479 on Build GRCh38
rs950439457 Chr.14: 96290305-96290305 on Build GRCh38
rs777809183 Chr.14: 96290591-96290591 on Build GRCh38
rs767680652 Chr.14: 96290481-96290481 on Build GRCh38
rs771952867 Chr.14: 96290409-96290409 on Build GRCh38
rs912158588 Chr.14: 96290317-96290317 on Build GRCh38
rs748176146 Chr.14: 96290488-96290488 on Build GRCh38
rs556615112 Chr.14: 96290681-96290681 on Build GRCh38
rs939378522 Chr.14: 96290468-96290468 on Build GRCh38
rs759045775 Chr.14: 96290442-96290442 on Build GRCh38
rs751967764 Chr.14: 96290533-96290533 on Build GRCh38
rs776301761 Chr.14: 96290435-96290435 on Build GRCh38
rs763353608 Chr.14: 96290452-96290452 on Build GRCh38
rs879867905 Chr.14: 96290331-96290331 on Build GRCh38
rs757487463 Chr.14: 96290471-96290471 on Build GRCh38
rs760407167 Chr.14: 96290422-96290422 on Build GRCh38
rs1028471083 Chr.14: 96290724-96290724 on Build GRCh38
rs769262405 Chr.14: 96290641-96290641 on Build GRCh38
rs764529741 Chr.14: 96290535-96290535 on Build GRCh38
rs745483524 Chr.14: 96290529-96290529 on Build GRCh38
rs771117337 Chr.14: 96290431-96290431 on Build GRCh38
rs148160259 Chr.14: 96290546-96290546 on Build GRCh38
rs748005406 Chr.14: 96290396-96290396 on Build GRCh38
rs963132981 Chr.14: 96290256-96290256 on Build GRCh38
rs764753187 Chr.14: 96290428-96290428 on Build GRCh38
rs914450393 Chr.14: 96290470-96290470 on Build GRCh38
rs77294752 Chr.14: 96290636-96290636 on Build GRCh38
rs200959600 Chr.14: 96290487-96290487 on Build GRCh38

View Full Product Details