Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 12664449-12665043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPPED1
Entrez Gene ID
55313
Gene Name
calcineurin like phosphoesterase domain containing 1
Gene Aliases
CSTP1
UniGene
Hs.460002
Gene Chromosome Location
Chr.16: 12659799-12803887 on Build GRCh38
Transcripts
NM_001099455.1 NM_018340.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6555423 c.818G>A p.R273K Chr.16: 12665013-12665013 on Build GRCh38
7218442 c.852C>T p.T284T Chr.16: 12664979-12664979 on Build GRCh38
4674422 c.872G>A p.R291Q Chr.16: 12664959-12664959 on Build GRCh38
1684433 c.822delC p.H276fs*20 Chr.16: 12665009-12665009 on Build GRCh38
2143303 c.826delC p.H276fs*20 Chr.16: 12665005-12665005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778867274 Chr.16: 12665004-12665005 on Build GRCh38
rs781239364 Chr.16: 12664865-12664865 on Build GRCh38
rs927950834 Chr.16: 12664951-12664951 on Build GRCh38
rs755825407 Chr.16: 12664960-12664960 on Build GRCh38
rs904417382 Chr.16: 12664840-12664840 on Build GRCh38
rs369945677 Chr.16: 12664978-12664978 on Build GRCh38
rs201868301 Chr.16: 12664959-12664959 on Build GRCh38
rs755867525 Chr.16: 12665000-12665000 on Build GRCh38
rs75852522 Chr.16: 12664509-12664509 on Build GRCh38
rs765527165 Chr.16: 12664922-12664922 on Build GRCh38
rs751113154 Chr.16: 12664816-12664816 on Build GRCh38
rs184303669 Chr.16: 12664980-12664980 on Build GRCh38
rs559862740 Chr.16: 12664584-12664584 on Build GRCh38
rs746018723 Chr.16: 12665003-12665003 on Build GRCh38
rs998898637 Chr.16: 12664515-12664515 on Build GRCh38
rs925252809 Chr.16: 12664926-12664926 on Build GRCh38
rs574406755 Chr.16: 12664542-12664542 on Build GRCh38
rs757943303 Chr.16: 12664665-12664665 on Build GRCh38
rs750588214 Chr.16: 12664933-12664933 on Build GRCh38
rs531444451 Chr.16: 12664907-12664907 on Build GRCh38
rs1028441284 Chr.16: 12664575-12664575 on Build GRCh38
rs187369619 Chr.16: 12664970-12664970 on Build GRCh38
rs769289582 Chr.16: 12664981-12664981 on Build GRCh38
rs369886431 Chr.16: 12664874-12664879 on Build GRCh38
rs147152842 Chr.16: 12664988-12664988 on Build GRCh38
rs888719828 Chr.16: 12665008-12665008 on Build GRCh38
rs373744185 Chr.16: 12664874-12664874 on Build GRCh38
rs778672073 Chr.16: 12664969-12664969 on Build GRCh38
rs997955357 Chr.16: 12664561-12664561 on Build GRCh38
rs1046636697 Chr.16: 12664731-12664731 on Build GRCh38
rs755689329 Chr.16: 12664869-12664869 on Build GRCh38
rs537675910 Chr.16: 12664865-12664870 on Build GRCh38
rs374948695 Chr.16: 12664990-12664990 on Build GRCh38
rs562441710 Chr.16: 12664870-12664870 on Build GRCh38
rs143409742 Chr.16: 12664868-12664868 on Build GRCh38
rs183894919 Chr.16: 12664559-12664559 on Build GRCh38
rs554743567 Chr.16: 12664524-12664524 on Build GRCh38
rs754681484 Chr.16: 12664863-12664863 on Build GRCh38
rs773839952 Chr.16: 12664883-12664883 on Build GRCh38
rs537038258 Chr.16: 12664683-12664683 on Build GRCh38
rs1042923594 Chr.16: 12664827-12664827 on Build GRCh38
rs1012835566 Chr.16: 12664495-12664495 on Build GRCh38
rs894346207 Chr.16: 12664494-12664494 on Build GRCh38
rs750000145 Chr.16: 12664644-12664644 on Build GRCh38
rs746262783 Chr.16: 12664884-12664884 on Build GRCh38
rs994421943 Chr.16: 12664823-12664823 on Build GRCh38
rs796160360 Chr.16: 12664608-12664608 on Build GRCh38
rs887461503 Chr.16: 12664855-12664855 on Build GRCh38
rs781254681 Chr.16: 12664848-12664848 on Build GRCh38
rs764325680 Chr.16: 12664914-12664914 on Build GRCh38
rs749599146 Chr.16: 12664926-12664926 on Build GRCh38
rs1005265228 Chr.16: 12665015-12665015 on Build GRCh38
rs984562894 Chr.16: 12664563-12664563 on Build GRCh38
rs951844594 Chr.16: 12664556-12664556 on Build GRCh38
rs965462336 Chr.16: 12664557-12664557 on Build GRCh38
rs199618401 Chr.16: 12665002-12665002 on Build GRCh38
rs778279228 Chr.16: 12665019-12665019 on Build GRCh38
rs964593748 Chr.16: 12664606-12664606 on Build GRCh38
rs192282834 Chr.16: 12664979-12664979 on Build GRCh38
rs1055254360 Chr.16: 12664714-12664714 on Build GRCh38
rs1042873159 Chr.16: 12664972-12664972 on Build GRCh38
rs762140161 Chr.16: 12664585-12664585 on Build GRCh38
rs776101804 Chr.16: 12664895-12664895 on Build GRCh38
rs772298081 Chr.16: 12664885-12664885 on Build GRCh38
rs777238460 Chr.16: 12664986-12664986 on Build GRCh38
rs925295764 Chr.16: 12664673-12664673 on Build GRCh38
rs767213835 Chr.16: 12664939-12664939 on Build GRCh38
rs904177108 Chr.16: 12664510-12664510 on Build GRCh38
rs961893323 Chr.16: 12664908-12664908 on Build GRCh38
rs368474690 Chr.16: 12664953-12664953 on Build GRCh38
rs375153155 Chr.16: 12664952-12664952 on Build GRCh38
rs1031355535 Chr.16: 12664547-12664547 on Build GRCh38
rs765744731 Chr.16: 12664720-12664720 on Build GRCh38
rs993157797 Chr.16: 12664528-12664528 on Build GRCh38
rs934567384 Chr.16: 12664976-12664976 on Build GRCh38
rs961621378 Chr.16: 12664657-12664657 on Build GRCh38
rs575569665 Chr.16: 12664685-12664685 on Build GRCh38
rs910105964 Chr.16: 12664643-12664643 on Build GRCh38
rs367864945 Chr.16: 12664875-12664875 on Build GRCh38
rs755026639 Chr.16: 12664846-12664846 on Build GRCh38
rs973042127 Chr.16: 12664945-12664945 on Build GRCh38
rs200764803 Chr.16: 12665009-12665009 on Build GRCh38
rs755316542 Chr.16: 12664672-12664673 on Build GRCh38
rs745474668 Chr.16: 12664974-12664974 on Build GRCh38
rs915728897 Chr.16: 12664716-12664716 on Build GRCh38
rs11645068 Chr.16: 12664962-12664962 on Build GRCh38
rs897410892 Chr.16: 12664742-12664742 on Build GRCh38
rs1014230873 Chr.16: 12664484-12664484 on Build GRCh38
rs984321183 Chr.16: 12664624-12664624 on Build GRCh38
rs752496637 Chr.16: 12664996-12664996 on Build GRCh38
rs568900546 Chr.16: 12664943-12664943 on Build GRCh38
rs764979105 Chr.16: 12664626-12664626 on Build GRCh38
rs753746918 Chr.16: 12664840-12664841 on Build GRCh38
rs948563540 Chr.16: 12664721-12664721 on Build GRCh38
rs371646726 Chr.16: 12664915-12664915 on Build GRCh38
rs754842319 Chr.16: 12664934-12664934 on Build GRCh38
rs532120242 Chr.16: 12664684-12664684 on Build GRCh38
rs35337066 Chr.16: 12664953-12664954 on Build GRCh38
rs376818351 Chr.16: 12664975-12664975 on Build GRCh38

View Full Product Details