Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 124313694-124314293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM101A
Entrez Gene ID
144347
Gene Name
family with sequence similarity 101 member A
Gene Aliases
CFM2
UniGene
Hs.432901
Gene Chromosome Location
Chr.12: 124289164-124316024 on Build GRCh38
Transcripts
NM_181709.4 NM_001204299.1

Gene Symbol
ZNF664-FAM101A
Entrez Gene ID
100533183
Gene Name
filamin-interacting protein FAM101A
Gene Aliases
-
UniGene
Hs.432901
Gene Chromosome Location
Chr.12: 123973215-124316024 on Build GRCh38
Transcripts
NM_181709.4 NM_001204299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7132331 c.138C>T p.P46P Chr.12: 124314255-124314255 on Build GRCh38
3457775 c.344C>T p.S115L Chr.12: 124314218-124314218 on Build GRCh38
5681558 c.94T>C p.Y32H Chr.12: 124314211-124314211 on Build GRCh38
4848590 c.404G>T p.S135I Chr.12: 124314278-124314278 on Build GRCh38
6658017 c.171C>T p.I57I Chr.12: 124314288-124314288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757929264 Chr.12: 124314252-124314252 on Build GRCh38
rs776467052 Chr.12: 124314162-124314162 on Build GRCh38
rs746525811 Chr.12: 124313886-124313886 on Build GRCh38
rs954840309 Chr.12: 124313731-124313731 on Build GRCh38
rs937478296 Chr.12: 124313849-124313849 on Build GRCh38
rs181334168 Chr.12: 124313980-124313980 on Build GRCh38
rs113175048 Chr.12: 124314037-124314039 on Build GRCh38
rs116827239 Chr.12: 124313972-124313972 on Build GRCh38
rs1046220940 Chr.12: 124314083-124314083 on Build GRCh38
rs546328000 Chr.12: 124313957-124313957 on Build GRCh38
rs146502129 Chr.12: 124314057-124314057 on Build GRCh38
rs78753849 Chr.12: 124314265-124314265 on Build GRCh38
rs1049801921 Chr.12: 124314001-124314001 on Build GRCh38
rs767921361 Chr.12: 124314214-124314214 on Build GRCh38
rs530640495 Chr.12: 124313902-124313902 on Build GRCh38
rs561239086 Chr.12: 124313836-124313836 on Build GRCh38
rs565194359 Chr.12: 124314203-124314203 on Build GRCh38
rs935201273 Chr.12: 124313962-124313962 on Build GRCh38
rs770052960 Chr.12: 124314170-124314170 on Build GRCh38
rs775244486 Chr.12: 124314051-124314051 on Build GRCh38
rs12318072 Chr.12: 124314255-124314255 on Build GRCh38
rs747706977 Chr.12: 124314145-124314145 on Build GRCh38
rs902385616 Chr.12: 124313831-124313831 on Build GRCh38
rs898823633 Chr.12: 124313897-124313897 on Build GRCh38
rs978788929 Chr.12: 124313826-124313826 on Build GRCh38
rs867806328 Chr.12: 124314077-124314077 on Build GRCh38
rs115451914 Chr.12: 124313919-124313919 on Build GRCh38
rs368418511 Chr.12: 124314254-124314254 on Build GRCh38
rs750861270 Chr.12: 124314219-124314219 on Build GRCh38
rs571215734 Chr.12: 124313971-124313971 on Build GRCh38
rs80187036 Chr.12: 124313899-124313899 on Build GRCh38
rs139818421 Chr.12: 124314256-124314256 on Build GRCh38
rs189697118 Chr.12: 124313896-124313896 on Build GRCh38
rs77026273 Chr.12: 124314142-124314142 on Build GRCh38
rs150677447 Chr.12: 124314209-124314209 on Build GRCh38
rs148465703 Chr.12: 124313819-124313819 on Build GRCh38
rs979408686 Chr.12: 124313947-124313947 on Build GRCh38
rs779438484 Chr.12: 124313873-124313873 on Build GRCh38
rs372026151 Chr.12: 124314264-124314264 on Build GRCh38
rs1028341039 Chr.12: 124313878-124313878 on Build GRCh38
rs754653351 Chr.12: 124314238-124314238 on Build GRCh38
rs142620622 Chr.12: 124313892-124313892 on Build GRCh38
rs975991284 Chr.12: 124314248-124314248 on Build GRCh38
rs763124898 Chr.12: 124314185-124314185 on Build GRCh38
rs946267454 Chr.12: 124313824-124313824 on Build GRCh38
rs990498278 Chr.12: 124313926-124313926 on Build GRCh38
rs199963697 Chr.12: 124314213-124314213 on Build GRCh38
rs116090335 Chr.12: 124314251-124314251 on Build GRCh38
rs61640933 Chr.12: 124313891-124313891 on Build GRCh38
rs758216685 Chr.12: 124314148-124314148 on Build GRCh38
rs775500450 Chr.12: 124314184-124314184 on Build GRCh38
rs201083008 Chr.12: 124314208-124314208 on Build GRCh38
rs946385063 Chr.12: 124313933-124313933 on Build GRCh38
rs925914319 Chr.12: 124313838-124313838 on Build GRCh38
rs746715548 Chr.12: 124314153-124314153 on Build GRCh38
rs114386636 Chr.12: 124314117-124314117 on Build GRCh38
rs765000215 Chr.12: 124314244-124314244 on Build GRCh38
rs746084849 Chr.12: 124314169-124314169 on Build GRCh38
rs916252834 Chr.12: 124313930-124313930 on Build GRCh38
rs141134804 Chr.12: 124314070-124314070 on Build GRCh38
rs757117436 Chr.12: 124314148-124314148 on Build GRCh38
rs987751586 Chr.12: 124313818-124313818 on Build GRCh38
rs548734808 Chr.12: 124313915-124313915 on Build GRCh38
rs749272800 Chr.12: 124314271-124314271 on Build GRCh38
rs575971708 Chr.12: 124314040-124314040 on Build GRCh38
rs958960693 Chr.12: 124314273-124314273 on Build GRCh38
rs765893793 Chr.12: 124314141-124314141 on Build GRCh38
rs949232753 Chr.12: 124313872-124313872 on Build GRCh38
rs907692914 Chr.12: 124314085-124314085 on Build GRCh38
rs77570261 Chr.12: 124314270-124314270 on Build GRCh38
rs770773312 Chr.12: 124314154-124314154 on Build GRCh38
rs573269008 Chr.12: 124313866-124313866 on Build GRCh38
rs1035161165 Chr.12: 124314129-124314129 on Build GRCh38
rs11610586 Chr.12: 124313867-124313867 on Build GRCh38
rs954416722 Chr.12: 124313920-124313920 on Build GRCh38
rs926016643 Chr.12: 124314021-124314021 on Build GRCh38
rs917348614 Chr.12: 124314063-124314063 on Build GRCh38

View Full Product Details