Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123612915-123613488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX55
Entrez Gene ID
57696
Gene Name
DEAD-box helicase 55
Gene Aliases
-
UniGene
Hs.286173
Gene Chromosome Location
Chr.12: 123602077-123620943 on Build GRCh38
Transcripts
NM_020936.2 NR_135104.1 NR_135105.1 XM_011538608.2 XM_017019710.1 XM_017019711.1 XM_017019712.1 XM_017019713.1 XM_017019714.1 XM_017019715.1 XM_017019716.1 XM_017019717.1 XM_017019718.1 XM_017019719.1 XM_017019720.1 XM_017019721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7153074 c.784C>T p.L262F Chr.12: 123613212-123613212 on Build GRCh38
936462 c.787C>T p.R263C Chr.12: 123613215-123613215 on Build GRCh38
5374499 c.777C>T p.V259V Chr.12: 123613205-123613205 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs571473029 Chr.12: 123613338-123613338 on Build GRCh38
rs866567581 Chr.12: 123613129-123613129 on Build GRCh38
rs884892 Chr.12: 123612981-123612981 on Build GRCh38
rs779573777 Chr.12: 123613194-123613194 on Build GRCh38
rs373062073 Chr.12: 123613281-123613281 on Build GRCh38
rs11057306 Chr.12: 123613219-123613219 on Build GRCh38
rs3748268 Chr.12: 123613277-123613277 on Build GRCh38
rs750469758 Chr.12: 123613278-123613278 on Build GRCh38
rs903392043 Chr.12: 123613047-123613047 on Build GRCh38
rs952764501 Chr.12: 123613432-123613432 on Build GRCh38
rs762990636 Chr.12: 123613143-123613143 on Build GRCh38
rs914281701 Chr.12: 123613354-123613354 on Build GRCh38
rs565144643 Chr.12: 123613173-123613173 on Build GRCh38
rs34407996 Chr.12: 123613431-123613431 on Build GRCh38
rs940538158 Chr.12: 123612985-123612985 on Build GRCh38
rs575175523 Chr.12: 123613072-123613072 on Build GRCh38
rs181740577 Chr.12: 123613112-123613112 on Build GRCh38
rs937051032 Chr.12: 123613005-123613007 on Build GRCh38
rs575280431 Chr.12: 123613428-123613428 on Build GRCh38
rs928649858 Chr.12: 123612973-123612973 on Build GRCh38
rs906966343 Chr.12: 123613002-123613003 on Build GRCh38
rs554155680 Chr.12: 123613438-123613438 on Build GRCh38
rs757969750 Chr.12: 123613162-123613162 on Build GRCh38
rs760704695 Chr.12: 123613280-123613280 on Build GRCh38
rs903447098 Chr.12: 123613393-123613393 on Build GRCh38
rs1037709054 Chr.12: 123613036-123613036 on Build GRCh38
rs915773122 Chr.12: 123613254-123613254 on Build GRCh38
rs769085165 Chr.12: 123613268-123613269 on Build GRCh38
rs532452790 Chr.12: 123613200-123613200 on Build GRCh38
rs141932661 Chr.12: 123613216-123613216 on Build GRCh38
rs775405809 Chr.12: 123613139-123613139 on Build GRCh38
rs752493803 Chr.12: 123613000-123613001 on Build GRCh38
rs928435297 Chr.12: 123613315-123613315 on Build GRCh38
rs1011531697 Chr.12: 123613406-123613406 on Build GRCh38
rs1015987722 Chr.12: 123613462-123613462 on Build GRCh38
rs369679658 Chr.12: 123613181-123613181 on Build GRCh38
rs920164251 Chr.12: 123613312-123613312 on Build GRCh38
rs755317393 Chr.12: 123613214-123613214 on Build GRCh38
rs1040116737 Chr.12: 123613339-123613339 on Build GRCh38
rs901469476 Chr.12: 123613355-123613355 on Build GRCh38
rs57153600 Chr.12: 123613001-123613002 on Build GRCh38
rs998932941 Chr.12: 123613367-123613367 on Build GRCh38
rs148148585 Chr.12: 123613215-123613215 on Build GRCh38
rs888861485 Chr.12: 123613178-123613178 on Build GRCh38
rs79514038 Chr.12: 123613336-123613336 on Build GRCh38
rs761806902 Chr.12: 123613273-123613273 on Build GRCh38
rs140634646 Chr.12: 123613185-123613185 on Build GRCh38
rs59168328 Chr.12: 123613387-123613387 on Build GRCh38
rs1045746114 Chr.12: 123612956-123612956 on Build GRCh38
rs149103406 Chr.12: 123613253-123613253 on Build GRCh38
rs767943918 Chr.12: 123613137-123613137 on Build GRCh38
rs948824382 Chr.12: 123612949-123612949 on Build GRCh38
rs962011776 Chr.12: 123613463-123613463 on Build GRCh38
rs753949936 Chr.12: 123613454-123613454 on Build GRCh38
rs757297594 Chr.12: 123613159-123613159 on Build GRCh38
rs892794816 Chr.12: 123613026-123613026 on Build GRCh38
rs931622949 Chr.12: 123613314-123613314 on Build GRCh38
rs76443868 Chr.12: 123613001-123613001 on Build GRCh38
rs766631135 Chr.12: 123613449-123613449 on Build GRCh38
rs984711750 Chr.12: 123612976-123612976 on Build GRCh38
rs762211199 Chr.12: 123613135-123613135 on Build GRCh38
rs186434033 Chr.12: 123613204-123613204 on Build GRCh38
rs778351991 Chr.12: 123613138-123613139 on Build GRCh38
rs545098868 Chr.12: 123613043-123613043 on Build GRCh38
rs751467847 Chr.12: 123613156-123613156 on Build GRCh38
rs767519190 Chr.12: 123613172-123613172 on Build GRCh38
rs534934005 Chr.12: 123613332-123613332 on Build GRCh38
rs76608317 Chr.12: 123613402-123613402 on Build GRCh38
rs1055494053 Chr.12: 123613021-123613021 on Build GRCh38
rs777140516 Chr.12: 123613275-123613276 on Build GRCh38
rs375579705 Chr.12: 123613064-123613064 on Build GRCh38
rs745351231 Chr.12: 123613145-123613145 on Build GRCh38
rs769543037 Chr.12: 123613241-123613241 on Build GRCh38
rs981485962 Chr.12: 123613439-123613439 on Build GRCh38
rs528778850 Chr.12: 123613114-123613114 on Build GRCh38
rs970119553 Chr.12: 123613256-123613256 on Build GRCh38
rs952707396 Chr.12: 123613240-123613240 on Build GRCh38
rs746737191 Chr.12: 123613250-123613250 on Build GRCh38
rs540390630 Chr.12: 123613145-123613145 on Build GRCh38
rs117200049 Chr.12: 123613230-123613230 on Build GRCh38
rs1037253735 Chr.12: 123613308-123613308 on Build GRCh38
rs750700562 Chr.12: 123613175-123613175 on Build GRCh38
rs377149419 Chr.12: 123613260-123613260 on Build GRCh38
rs542302586 Chr.12: 123613077-123613077 on Build GRCh38
rs751318491 Chr.12: 123613264-123613264 on Build GRCh38
rs143703504 Chr.12: 123613210-123613210 on Build GRCh38
rs567539448 Chr.12: 123613288-123613288 on Build GRCh38
rs768442362 Chr.12: 123613262-123613262 on Build GRCh38
rs191155414 Chr.12: 123613429-123613429 on Build GRCh38
rs188921944 Chr.12: 123613044-123613044 on Build GRCh38

View Full Product Details