Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69462452-69462902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5B
Entrez Gene ID
80777
Gene Name
cytochrome b5 type B
Gene Aliases
CYB5-M, CYPB5M, OMB5
UniGene
Hs.461131
Gene Chromosome Location
Chr.16: 69424595-69466264 on Build GRCh38
Transcripts
NM_030579.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
972893 c.388A>G p.I130V Chr.16: 69462455-69462455 on Build GRCh38
6555827 c.434C>T p.S145L Chr.16: 69462501-69462501 on Build GRCh38
1202768 c.415C>A p.L139M Chr.16: 69462482-69462482 on Build GRCh38
352615 c.425A>G p.Y142C Chr.16: 69462492-69462492 on Build GRCh38
4675778 c.422G>A p.R141H Chr.16: 69462489-69462489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546818684 Chr.16: 69462774-69462774 on Build GRCh38
rs761436691 Chr.16: 69462529-69462529 on Build GRCh38
rs534618389 Chr.16: 69462534-69462534 on Build GRCh38
rs190207594 Chr.16: 69462746-69462746 on Build GRCh38
rs899639094 Chr.16: 69462682-69462682 on Build GRCh38
rs200320532 Chr.16: 69462524-69462524 on Build GRCh38
rs771877684 Chr.16: 69462646-69462646 on Build GRCh38
rs767084029 Chr.16: 69462531-69462531 on Build GRCh38
rs762722460 Chr.16: 69462514-69462514 on Build GRCh38
rs1053982707 Chr.16: 69462516-69462516 on Build GRCh38
rs548935986 Chr.16: 69462731-69462731 on Build GRCh38
rs1003124250 Chr.16: 69462874-69462874 on Build GRCh38
rs77720661 Chr.16: 69462843-69462843 on Build GRCh38
rs563601110 Chr.16: 69462663-69462663 on Build GRCh38
rs779109350 Chr.16: 69462481-69462481 on Build GRCh38
rs754990243 Chr.16: 69462617-69462617 on Build GRCh38
rs369459264 Chr.16: 69462723-69462723 on Build GRCh38
rs574371101 Chr.16: 69462738-69462738 on Build GRCh38
rs745781758 Chr.16: 69462505-69462505 on Build GRCh38
rs369428215 Chr.16: 69462506-69462506 on Build GRCh38
rs760239341 Chr.16: 69462655-69462655 on Build GRCh38
rs939605566 Chr.16: 69462870-69462870 on Build GRCh38
rs184570827 Chr.16: 69462526-69462526 on Build GRCh38
rs772685298 Chr.16: 69462647-69462647 on Build GRCh38
rs374964865 Chr.16: 69462572-69462572 on Build GRCh38
rs1029530171 Chr.16: 69462704-69462704 on Build GRCh38
rs71710859 Chr.16: 69462860-69462863 on Build GRCh38
rs973821206 Chr.16: 69462763-69462763 on Build GRCh38
rs539209842 Chr.16: 69462860-69462860 on Build GRCh38
rs1186093 Chr.16: 69462634-69462634 on Build GRCh38
rs140114091 Chr.16: 69462499-69462499 on Build GRCh38
rs997064784 Chr.16: 69462692-69462692 on Build GRCh38
rs755154659 Chr.16: 69462476-69462476 on Build GRCh38
rs144106394 Chr.16: 69462857-69462860 on Build GRCh38
rs554220407 Chr.16: 69462652-69462652 on Build GRCh38
rs765362758 Chr.16: 69462553-69462553 on Build GRCh38
rs373628023 Chr.16: 69462651-69462651 on Build GRCh38
rs910214669 Chr.16: 69462727-69462727 on Build GRCh38
rs946785259 Chr.16: 69462630-69462630 on Build GRCh38
rs915012822 Chr.16: 69462741-69462741 on Build GRCh38
rs772074303 Chr.16: 69462492-69462492 on Build GRCh38
rs950610208 Chr.16: 69462641-69462641 on Build GRCh38
rs751019701 Chr.16: 69462543-69462543 on Build GRCh38
rs761237649 Chr.16: 69462561-69462561 on Build GRCh38
rs761664954 Chr.16: 69462631-69462631 on Build GRCh38
rs1011715905 Chr.16: 69462532-69462532 on Build GRCh38
rs533410178 Chr.16: 69462623-69462623 on Build GRCh38
rs376387682 Chr.16: 69462489-69462489 on Build GRCh38
rs958998385 Chr.16: 69462638-69462638 on Build GRCh38
rs765804282 Chr.16: 69462658-69462658 on Build GRCh38
rs775141212 Chr.16: 69462507-69462507 on Build GRCh38
rs903429736 Chr.16: 69462586-69462586 on Build GRCh38
rs903397023 Chr.16: 69462844-69462844 on Build GRCh38
rs781139329 Chr.16: 69462502-69462502 on Build GRCh38
rs376679722 Chr.16: 69462748-69462748 on Build GRCh38
rs943029771 Chr.16: 69462729-69462729 on Build GRCh38
rs551563786 Chr.16: 69462640-69462640 on Build GRCh38
rs540803071 Chr.16: 69462601-69462601 on Build GRCh38
rs565853387 Chr.16: 69462606-69462606 on Build GRCh38
rs969859115 Chr.16: 69462806-69462806 on Build GRCh38
rs567347570 Chr.16: 69462740-69462740 on Build GRCh38
rs766837387 Chr.16: 69462562-69462562 on Build GRCh38
rs954635602 Chr.16: 69462709-69462709 on Build GRCh38

View Full Product Details