Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4170604-4170965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKFY1
Entrez Gene ID
51479
Gene Name
ankyrin repeat and FYVE domain containing 1
Gene Aliases
ANKHZN, BTBD23, ZFYVE14
UniGene
Hs.696087
Gene Chromosome Location
Chr.17: 4163371-4264026 on Build GRCh38
Transcripts
NM_001257999.1 NM_016376.3 NR_047571.1 XM_005256679.3 XM_011523926.1 XM_011523928.2 XM_011523933.1 XM_017024733.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6147043 c.3331C>T p.R1111C Chr.17: 4170796-4170796 on Build GRCh38
6342439 c.3159C>A p.Y1053* Chr.17: 4170845-4170845 on Build GRCh38
7355516 c.3332G>A p.R1111H Chr.17: 4170795-4170795 on Build GRCh38
1521509 c.3208C>T p.R1070C Chr.17: 4170796-4170796 on Build GRCh38
3717380 c.3186C>T p.R1062R Chr.17: 4170818-4170818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780510222 Chr.17: 4170879-4170879 on Build GRCh38
rs772589429 Chr.17: 4170878-4170878 on Build GRCh38
rs191200704 Chr.17: 4170922-4170922 on Build GRCh38
rs759455991 Chr.17: 4170811-4170811 on Build GRCh38
rs777564479 Chr.17: 4170837-4170837 on Build GRCh38
rs755008788 Chr.17: 4170671-4170671 on Build GRCh38
rs764144399 Chr.17: 4170909-4170909 on Build GRCh38
rs748559930 Chr.17: 4170647-4170647 on Build GRCh38
rs369791240 Chr.17: 4170700-4170700 on Build GRCh38
rs200602880 Chr.17: 4170881-4170881 on Build GRCh38
rs765916649 Chr.17: 4170704-4170704 on Build GRCh38
rs776707196 Chr.17: 4170812-4170812 on Build GRCh38
rs376973799 Chr.17: 4170844-4170844 on Build GRCh38
rs200896506 Chr.17: 4170681-4170681 on Build GRCh38
rs372207094 Chr.17: 4170820-4170820 on Build GRCh38
rs760447087 Chr.17: 4170903-4170903 on Build GRCh38
rs947729298 Chr.17: 4170796-4170796 on Build GRCh38
rs375331862 Chr.17: 4170674-4170674 on Build GRCh38
rs865832436 Chr.17: 4170850-4170850 on Build GRCh38
rs1018176556 Chr.17: 4170651-4170651 on Build GRCh38
rs200685853 Chr.17: 4170795-4170795 on Build GRCh38
rs368037383 Chr.17: 4170817-4170817 on Build GRCh38
rs759818684 Chr.17: 4170703-4170703 on Build GRCh38
rs773916502 Chr.17: 4170799-4170799 on Build GRCh38
rs1016369634 Chr.17: 4170918-4170918 on Build GRCh38
rs773505106 Chr.17: 4170896-4170896 on Build GRCh38
rs368663243 Chr.17: 4170832-4170832 on Build GRCh38
rs199727847 Chr.17: 4170791-4170791 on Build GRCh38
rs767490580 Chr.17: 4170838-4170838 on Build GRCh38
rs372382822 Chr.17: 4170833-4170833 on Build GRCh38
rs776980697 Chr.17: 4170887-4170887 on Build GRCh38
rs778670003 Chr.17: 4170868-4170868 on Build GRCh38
rs761217096 Chr.17: 4170807-4170807 on Build GRCh38
rs888450476 Chr.17: 4170646-4170646 on Build GRCh38
rs766092499 Chr.17: 4170746-4170746 on Build GRCh38
rs769719497 Chr.17: 4170895-4170895 on Build GRCh38
rs771208566 Chr.17: 4170696-4170696 on Build GRCh38
rs529196360 Chr.17: 4170939-4170939 on Build GRCh38
rs748037483 Chr.17: 4170777-4170777 on Build GRCh38
rs956538445 Chr.17: 4170904-4170904 on Build GRCh38
rs537026576 Chr.17: 4170689-4170689 on Build GRCh38
rs114686778 Chr.17: 4170818-4170818 on Build GRCh38
rs751428903 Chr.17: 4170748-4170748 on Build GRCh38
rs753448811 Chr.17: 4170840-4170840 on Build GRCh38
rs776174515 Chr.17: 4170804-4170804 on Build GRCh38
rs763099085 Chr.17: 4170897-4170897 on Build GRCh38
rs779851898 Chr.17: 4170667-4170667 on Build GRCh38
rs186434827 Chr.17: 4170803-4170803 on Build GRCh38
rs367586240 Chr.17: 4170819-4170819 on Build GRCh38
rs144059556 Chr.17: 4170652-4170652 on Build GRCh38
rs144397517 Chr.17: 4170830-4170830 on Build GRCh38
rs776121056 Chr.17: 4170707-4170707 on Build GRCh38
rs746934373 Chr.17: 4170670-4170670 on Build GRCh38
rs753579331 Chr.17: 4170660-4170660 on Build GRCh38
rs375906756 Chr.17: 4170889-4170889 on Build GRCh38
rs756826513 Chr.17: 4170911-4170911 on Build GRCh38
rs757442296 Chr.17: 4170745-4170745 on Build GRCh38
rs747703887 Chr.17: 4170673-4170673 on Build GRCh38
rs781145478 Chr.17: 4170754-4170754 on Build GRCh38
rs753690594 Chr.17: 4170910-4170910 on Build GRCh38
rs752449791 Chr.17: 4170859-4170859 on Build GRCh38
rs202011309 Chr.17: 4170870-4170870 on Build GRCh38
rs201907808 Chr.17: 4170880-4170880 on Build GRCh38
rs200913206 Chr.17: 4170899-4170899 on Build GRCh38
rs754744722 Chr.17: 4170751-4170751 on Build GRCh38
rs748894341 Chr.17: 4170790-4170790 on Build GRCh38
rs755640421 Chr.17: 4170784-4170784 on Build GRCh38
rs764229912 Chr.17: 4170726-4170726 on Build GRCh38
rs777204989 Chr.17: 4170787-4170787 on Build GRCh38
rs754031562 Chr.17: 4170728-4170728 on Build GRCh38
rs927580880 Chr.17: 4170930-4170930 on Build GRCh38
rs866427741 Chr.17: 4170777-4170778 on Build GRCh38
rs980481200 Chr.17: 4170923-4170923 on Build GRCh38

View Full Product Details