Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 74947478-74948073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf40
Entrez Gene ID
55071
Gene Name
chromosome 9 open reading frame 40
Gene Aliases
-
UniGene
Hs.532296
Gene Chromosome Location
Chr.9: 74946583-74952886 on Build GRCh38
Transcripts
NM_017998.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4984767 c.569T>C p.V190A Chr.9: 74948064-74948064 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs902103464 Chr.9: 74947694-74947694 on Build GRCh38
rs747294638 Chr.9: 74948041-74948041 on Build GRCh38
rs1007691177 Chr.9: 74947682-74947682 on Build GRCh38
rs934672913 Chr.9: 74947913-74947913 on Build GRCh38
rs867724046 Chr.9: 74947941-74947941 on Build GRCh38
rs893276888 Chr.9: 74947673-74947673 on Build GRCh38
rs540415846 Chr.9: 74947960-74947960 on Build GRCh38
rs11144168 Chr.9: 74947779-74947779 on Build GRCh38
rs755265761 Chr.9: 74948045-74948045 on Build GRCh38
rs774256783 Chr.9: 74947934-74947934 on Build GRCh38
rs983509748 Chr.9: 74947882-74947882 on Build GRCh38
rs144601043 Chr.9: 74947565-74947565 on Build GRCh38
rs147106981 Chr.9: 74948052-74948052 on Build GRCh38
rs999902162 Chr.9: 74947750-74947750 on Build GRCh38
rs900771868 Chr.9: 74947622-74947622 on Build GRCh38
rs950486994 Chr.9: 74947501-74947501 on Build GRCh38
rs978027112 Chr.9: 74947897-74947897 on Build GRCh38
rs893215464 Chr.9: 74947954-74947954 on Build GRCh38
rs958902889 Chr.9: 74947769-74947769 on Build GRCh38
rs35747296 Chr.9: 74947720-74947721 on Build GRCh38
rs756684380 Chr.9: 74948028-74948028 on Build GRCh38
rs908003591 Chr.9: 74947924-74947924 on Build GRCh38
rs754599185 Chr.9: 74947614-74947615 on Build GRCh38
rs993094657 Chr.9: 74947563-74947563 on Build GRCh38
rs968334751 Chr.9: 74947537-74947537 on Build GRCh38
rs544572606 Chr.9: 74947631-74947631 on Build GRCh38
rs917846804 Chr.9: 74947583-74947583 on Build GRCh38
rs1045759379 Chr.9: 74947606-74947606 on Build GRCh38
rs917794527 Chr.9: 74947874-74947874 on Build GRCh38
rs1020010943 Chr.9: 74947696-74947697 on Build GRCh38
rs890604209 Chr.9: 74947641-74947641 on Build GRCh38
rs753353619 Chr.9: 74948022-74948022 on Build GRCh38
rs558952419 Chr.9: 74947526-74947526 on Build GRCh38
rs958647991 Chr.9: 74947548-74947548 on Build GRCh38
rs765247019 Chr.9: 74948029-74948029 on Build GRCh38
rs924613092 Chr.9: 74947899-74947899 on Build GRCh38
rs181476049 Chr.9: 74947624-74947624 on Build GRCh38
rs775160241 Chr.9: 74948009-74948009 on Build GRCh38
rs763648355 Chr.9: 74948019-74948019 on Build GRCh38
rs927230245 Chr.9: 74947539-74947539 on Build GRCh38
rs990654038 Chr.9: 74947814-74947814 on Build GRCh38
rs902046314 Chr.9: 74947988-74947988 on Build GRCh38
rs934744193 Chr.9: 74947623-74947623 on Build GRCh38
rs943498371 Chr.9: 74947978-74947978 on Build GRCh38
rs748288518 Chr.9: 74948047-74948047 on Build GRCh38
rs556052232 Chr.9: 74947564-74947564 on Build GRCh38
rs578041416 Chr.9: 74947815-74947815 on Build GRCh38
rs1017774480 Chr.9: 74947686-74947686 on Build GRCh38
rs950457012 Chr.9: 74947754-74947754 on Build GRCh38
rs111660354 Chr.9: 74947996-74947996 on Build GRCh38
rs909026014 Chr.9: 74947543-74947543 on Build GRCh38
rs760314227 Chr.9: 74948017-74948017 on Build GRCh38
rs984973217 Chr.9: 74947531-74947531 on Build GRCh38
rs890627417 Chr.9: 74948003-74948003 on Build GRCh38
rs978682995 Chr.9: 74947539-74947539 on Build GRCh38
rs369913991 Chr.9: 74948036-74948036 on Build GRCh38
rs997759661 Chr.9: 74947710-74947710 on Build GRCh38
rs949275134 Chr.9: 74947888-74947888 on Build GRCh38
rs1009991196 Chr.9: 74947685-74947685 on Build GRCh38
rs767956703 Chr.9: 74948031-74948031 on Build GRCh38
rs886075914 Chr.9: 74948033-74948033 on Build GRCh38
rs907885749 Chr.9: 74947602-74947602 on Build GRCh38
rs140264373 Chr.9: 74947987-74947987 on Build GRCh38
rs932243267 Chr.9: 74947986-74947986 on Build GRCh38
rs374853709 Chr.9: 74948037-74948037 on Build GRCh38
rs1025964302 Chr.9: 74947741-74947741 on Build GRCh38
rs949992112 Chr.9: 74947601-74947601 on Build GRCh38
rs766626425 Chr.9: 74948008-74948008 on Build GRCh38
rs903212003 Chr.9: 74947615-74947615 on Build GRCh38
rs781497162 Chr.9: 74948046-74948046 on Build GRCh38
rs138629715 Chr.9: 74947775-74947775 on Build GRCh38
rs984531740 Chr.9: 74947761-74947761 on Build GRCh38
rs959201030 Chr.9: 74947795-74947795 on Build GRCh38

View Full Product Details