Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 190387979-190388523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN16
Entrez Gene ID
10686
Gene Name
claudin 16
Gene Aliases
HOMG3, PCLN1
UniGene
Hs.251391
Gene Chromosome Location
Chr.3: 190387872-190412143 on Build GRCh38
Transcripts
NM_006580.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1042520 c.211A>C p.M71L Chr.3: 190388330-190388330 on Build GRCh38
3721265 c.226C>A p.Q76K Chr.3: 190388345-190388345 on Build GRCh38
4952471 c.310G>T p.D104Y Chr.3: 190388429-190388429 on Build GRCh38
1042519 c.86A>C p.K29T Chr.3: 190388205-190388205 on Build GRCh38
7164946 c.297G>C p.W99C Chr.3: 190388416-190388416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751220222 Chr.3: 190388233-190388233 on Build GRCh38
rs769827846 Chr.3: 190388357-190388357 on Build GRCh38
rs563081774 Chr.3: 190388488-190388488 on Build GRCh38
rs749994123 Chr.3: 190388128-190388128 on Build GRCh38
rs777659077 Chr.3: 190388175-190388175 on Build GRCh38
rs772401894 Chr.3: 190388183-190388183 on Build GRCh38
rs376639837 Chr.3: 190388085-190388085 on Build GRCh38
rs767771669 Chr.3: 190388457-190388457 on Build GRCh38
rs767080609 Chr.3: 190388238-190388238 on Build GRCh38
rs768569589 Chr.3: 190388275-190388275 on Build GRCh38
rs866440005 Chr.3: 190388359-190388361 on Build GRCh38
rs929445776 Chr.3: 190388482-190388482 on Build GRCh38
rs960613047 Chr.3: 190388159-190388159 on Build GRCh38
rs775073665 Chr.3: 190388410-190388410 on Build GRCh38
rs967573821 Chr.3: 190388380-190388380 on Build GRCh38
rs774637593 Chr.3: 190388059-190388059 on Build GRCh38
rs916967607 Chr.3: 190388484-190388484 on Build GRCh38
rs752540574 Chr.3: 190388246-190388246 on Build GRCh38
rs372525072 Chr.3: 190388333-190388333 on Build GRCh38
rs149955797 Chr.3: 190388289-190388289 on Build GRCh38
rs952746630 Chr.3: 190388003-190388003 on Build GRCh38
rs756139771 Chr.3: 190388478-190388478 on Build GRCh38
rs149116671 Chr.3: 190388374-190388374 on Build GRCh38
rs777259905 Chr.3: 190388115-190388115 on Build GRCh38
rs886058244 Chr.3: 190388157-190388157 on Build GRCh38
rs773763573 Chr.3: 190388288-190388288 on Build GRCh38
rs867514971 Chr.3: 190388346-190388346 on Build GRCh38
rs368215264 Chr.3: 190388349-190388349 on Build GRCh38
rs375640819 Chr.3: 190388348-190388348 on Build GRCh38
rs774670118 Chr.3: 190388383-190388383 on Build GRCh38
rs767495204 Chr.3: 190388122-190388122 on Build GRCh38
rs768702333 Chr.3: 190388055-190388055 on Build GRCh38
rs773530495 Chr.3: 190388210-190388210 on Build GRCh38
rs548797774 Chr.3: 190388060-190388060 on Build GRCh38
rs1020861027 Chr.3: 190388367-190388367 on Build GRCh38
rs761873372 Chr.3: 190388435-190388435 on Build GRCh38
rs752863742 Chr.3: 190388473-190388473 on Build GRCh38
rs763695473 Chr.3: 190388285-190388286 on Build GRCh38
rs767119879 Chr.3: 190388305-190388305 on Build GRCh38
rs929540511 Chr.3: 190388217-190388217 on Build GRCh38
rs886058243 Chr.3: 190388038-190388038 on Build GRCh38
rs778198632 Chr.3: 190388358-190388358 on Build GRCh38
rs201545856 Chr.3: 190388428-190388428 on Build GRCh38
rs759868163 Chr.3: 190388389-190388389 on Build GRCh38
rs765474628 Chr.3: 190388202-190388202 on Build GRCh38
rs199839143 Chr.3: 190388342-190388342 on Build GRCh38
rs1047717394 Chr.3: 190388075-190388075 on Build GRCh38
rs758074559 Chr.3: 190388132-190388132 on Build GRCh38
rs866648881 Chr.3: 190388297-190388297 on Build GRCh38
rs104893724 Chr.3: 190388331-190388331 on Build GRCh38
rs766056344 Chr.3: 190388141-190388141 on Build GRCh38
rs765304853 Chr.3: 190388437-190388437 on Build GRCh38
rs753777323 Chr.3: 190388429-190388429 on Build GRCh38
rs775681526 Chr.3: 190388110-190388110 on Build GRCh38
rs192579160 Chr.3: 190388079-190388079 on Build GRCh38
rs748133024 Chr.3: 190388155-190388155 on Build GRCh38
rs767846686 Chr.3: 190388304-190388304 on Build GRCh38
rs748213791 Chr.3: 190388284-190388284 on Build GRCh38
rs553547888 Chr.3: 190388281-190388282 on Build GRCh38
rs776943777 Chr.3: 190388197-190388197 on Build GRCh38
rs781362926 Chr.3: 190388277-190388277 on Build GRCh38
rs773947198 Chr.3: 190388295-190388295 on Build GRCh38
rs760754693 Chr.3: 190388283-190388285 on Build GRCh38
rs113105686 Chr.3: 190388094-190388094 on Build GRCh38
rs757186184 Chr.3: 190388263-190388263 on Build GRCh38
rs1491994 Chr.3: 190388453-190388453 on Build GRCh38
rs145475632 Chr.3: 190388196-190388196 on Build GRCh38
rs757743541 Chr.3: 190388493-190388493 on Build GRCh38
rs759268862 Chr.3: 190388301-190388301 on Build GRCh38
rs747082222 Chr.3: 190388186-190388186 on Build GRCh38
rs760160835 Chr.3: 190388309-190388309 on Build GRCh38
rs923427514 Chr.3: 190388472-190388472 on Build GRCh38
rs772239566 Chr.3: 190388108-190388108 on Build GRCh38
rs368234054 Chr.3: 190388285-190388285 on Build GRCh38
rs558573665 Chr.3: 190388265-190388265 on Build GRCh38
rs200322099 Chr.3: 190388117-190388117 on Build GRCh38
rs750369977 Chr.3: 190388235-190388235 on Build GRCh38
rs977137021 Chr.3: 190388366-190388366 on Build GRCh38
rs139923283 Chr.3: 190388229-190388229 on Build GRCh38
rs967711721 Chr.3: 190388192-190388192 on Build GRCh38
rs386669518 Chr.3: 190388284-190388285 on Build GRCh38
rs910725523 Chr.3: 190388031-190388031 on Build GRCh38
rs747245603 Chr.3: 190388272-190388272 on Build GRCh38
rs771301899 Chr.3: 190388292-190388292 on Build GRCh38
rs749819466 Chr.3: 190388360-190388360 on Build GRCh38
rs762080532 Chr.3: 190388232-190388232 on Build GRCh38
rs976244851 Chr.3: 190388466-190388466 on Build GRCh38
rs754537883 Chr.3: 190388147-190388147 on Build GRCh38
rs763354782 Chr.3: 190388214-190388214 on Build GRCh38
rs73192454 Chr.3: 190388492-190388492 on Build GRCh38
rs372129081 Chr.3: 190388353-190388353 on Build GRCh38
rs764125350 Chr.3: 190388313-190388313 on Build GRCh38
rs545693736 Chr.3: 190388417-190388417 on Build GRCh38
rs987831446 Chr.3: 190388166-190388166 on Build GRCh38
rs369250510 Chr.3: 190388456-190388456 on Build GRCh38
rs965435011 Chr.3: 190388354-190388354 on Build GRCh38
rs768547943 Chr.3: 190388078-190388078 on Build GRCh38
rs144731880 Chr.3: 190388181-190388181 on Build GRCh38
rs756048104 Chr.3: 190388248-190388248 on Build GRCh38
rs749083950 Chr.3: 190388262-190388262 on Build GRCh38
rs557493885 Chr.3: 190388114-190388114 on Build GRCh38
rs140781331 Chr.3: 190388148-190388148 on Build GRCh38
rs752230428 Chr.3: 190388307-190388307 on Build GRCh38
rs943528079 Chr.3: 190388038-190388038 on Build GRCh38
rs978876319 Chr.3: 190388131-190388131 on Build GRCh38
rs200400125 Chr.3: 190388264-190388264 on Build GRCh38
rs145118503 Chr.3: 190388351-190388351 on Build GRCh38
rs576643238 Chr.3: 190388040-190388040 on Build GRCh38
rs755820991 Chr.3: 190388156-190388156 on Build GRCh38
rs771742472 Chr.3: 190388391-190388391 on Build GRCh38
rs751044369 Chr.3: 190388143-190388143 on Build GRCh38
rs762405207 Chr.3: 190388198-190388198 on Build GRCh38
rs1041030100 Chr.3: 190388043-190388043 on Build GRCh38
rs3214506 Chr.3: 190388285-190388285 on Build GRCh38
rs777620076 Chr.3: 190388261-190388261 on Build GRCh38
rs1006196806 Chr.3: 190388280-190388280 on Build GRCh38
rs775914662 Chr.3: 190388185-190388185 on Build GRCh38
rs865841365 Chr.3: 190388311-190388312 on Build GRCh38
rs753832878 Chr.3: 190388318-190388318 on Build GRCh38
rs201380153 Chr.3: 190388283-190388283 on Build GRCh38
rs1040512329 Chr.3: 190388200-190388200 on Build GRCh38
rs909425040 Chr.3: 190388495-190388495 on Build GRCh38

View Full Product Details