Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 88140352-88140939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNR1
Entrez Gene ID
1268
Gene Name
cannabinoid receptor 1
Gene Aliases
CANN6, CB-R, CB1, CB1A, CB1K5, CB1R, CNR
UniGene
Hs.75110
Gene Chromosome Location
Chr.6: 88139864-88167429 on Build GRCh38
Transcripts
NM_001160226.1 NM_001160258.1 NM_001160259.1 NM_016083.4 NM_033181.3 XM_006715330.3 XM_011535424.1 XM_011535425.1 XM_011535426.1 XM_011535428.1 XM_017010238.1 XM_017010239.1 XM_017010240.1

SNP Detail

SNP ID Chromosome Location
rs897414456 Chr.6: 88140803-88140803 on Build GRCh38
rs1022419199 Chr.6: 88140884-88140884 on Build GRCh38
rs1035994630 Chr.6: 88140795-88140799 on Build GRCh38
rs185438180 Chr.6: 88140827-88140827 on Build GRCh38
rs538551548 Chr.6: 88140407-88140407 on Build GRCh38
rs548191602 Chr.6: 88140424-88140424 on Build GRCh38
rs756859003 Chr.6: 88140526-88140526 on Build GRCh38
rs1051278830 Chr.6: 88140839-88140839 on Build GRCh38
rs995006215 Chr.6: 88140816-88140816 on Build GRCh38
rs887479403 Chr.6: 88140399-88140399 on Build GRCh38
rs998363438 Chr.6: 88140433-88140433 on Build GRCh38
rs879188333 Chr.6: 88140473-88140473 on Build GRCh38
rs193267974 Chr.6: 88140691-88140691 on Build GRCh38
rs117523241 Chr.6: 88140905-88140905 on Build GRCh38
rs1054240893 Chr.6: 88140701-88140701 on Build GRCh38
rs778274865 Chr.6: 88140553-88140553 on Build GRCh38
rs74527916 Chr.6: 88140416-88140416 on Build GRCh38
rs935830455 Chr.6: 88140673-88140673 on Build GRCh38
rs758961475 Chr.6: 88140465-88140465 on Build GRCh38
rs978497607 Chr.6: 88140648-88140648 on Build GRCh38
rs908953335 Chr.6: 88140772-88140772 on Build GRCh38
rs954129430 Chr.6: 88140472-88140472 on Build GRCh38
rs900694958 Chr.6: 88140795-88140795 on Build GRCh38
rs984580726 Chr.6: 88140746-88140746 on Build GRCh38
rs950015800 Chr.6: 88140545-88140545 on Build GRCh38
rs1030911938 Chr.6: 88140671-88140671 on Build GRCh38
rs374574280 Chr.6: 88140785-88140785 on Build GRCh38
rs960571329 Chr.6: 88140688-88140688 on Build GRCh38
rs920665435 Chr.6: 88140431-88140431 on Build GRCh38
rs1050758673 Chr.6: 88140769-88140769 on Build GRCh38
rs113578046 Chr.6: 88140529-88140529 on Build GRCh38
rs890768399 Chr.6: 88140773-88140773 on Build GRCh38
rs889989523 Chr.6: 88140850-88140850 on Build GRCh38
rs868685038 Chr.6: 88140738-88140738 on Build GRCh38
rs917207341 Chr.6: 88140535-88140535 on Build GRCh38
rs76731415 Chr.6: 88140394-88140394 on Build GRCh38
rs34532735 Chr.6: 88140675-88140676 on Build GRCh38
rs968101772 Chr.6: 88140635-88140635 on Build GRCh38
rs940402103 Chr.6: 88140782-88140782 on Build GRCh38
rs751495962 Chr.6: 88140532-88140535 on Build GRCh38
rs806368 Chr.6: 88140381-88140381 on Build GRCh38
rs751993889 Chr.6: 88140881-88140881 on Build GRCh38
rs544173335 Chr.6: 88140612-88140612 on Build GRCh38
rs945740235 Chr.6: 88140706-88140706 on Build GRCh38
rs973921719 Chr.6: 88140418-88140418 on Build GRCh38
rs950803251 Chr.6: 88140882-88140882 on Build GRCh38
rs891548257 Chr.6: 88140705-88140705 on Build GRCh38
rs927137819 Chr.6: 88140599-88140599 on Build GRCh38
rs1021256944 Chr.6: 88140378-88140378 on Build GRCh38
rs1029797143 Chr.6: 88140441-88140441 on Build GRCh38
rs1004936403 Chr.6: 88140400-88140400 on Build GRCh38
rs75853128 Chr.6: 88140384-88140384 on Build GRCh38
rs950750969 Chr.6: 88140457-88140457 on Build GRCh38
rs1037325117 Chr.6: 88140417-88140417 on Build GRCh38
rs1017839485 Chr.6: 88140790-88140790 on Build GRCh38
rs533578074 Chr.6: 88140383-88140383 on Build GRCh38
rs947865244 Chr.6: 88140726-88140726 on Build GRCh38
rs1006947201 Chr.6: 88140519-88140519 on Build GRCh38

View Full Product Details