Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 131219615-131220155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP7
Entrez Gene ID
9465
Gene Name
A-kinase anchoring protein 7
Gene Aliases
AKAP15, AKAP18
UniGene
Hs.486483
Gene Chromosome Location
Chr.6: 131125698-131283535 on Build GRCh38
Transcripts
NM_016377.3 XM_005267228.1 XM_005267229.4 XM_006715614.2 XM_011536262.1 XM_017011506.1 XM_017011508.1 XM_017011509.1 XM_017011510.1 XM_017011511.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7234757 c.764G>T p.C255F Chr.6: 131219788-131219788 on Build GRCh38
5112075 c.642_643insA p.I217fs*4 Chr.6: 131219666-131219667 on Build GRCh38
3173054 c.643delA p.I217fs*1 Chr.6: 131219667-131219667 on Build GRCh38
5177008 c.784+9C>T p.? Chr.6: 131219817-131219817 on Build GRCh38
3858259 c.730C>T p.L244L Chr.6: 131219754-131219754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751894374 Chr.6: 131219816-131219816 on Build GRCh38
rs886915969 Chr.6: 131220008-131220008 on Build GRCh38
rs754751702 Chr.6: 131219721-131219721 on Build GRCh38
rs777206779 Chr.6: 131219759-131219760 on Build GRCh38
rs570019576 Chr.6: 131219874-131219874 on Build GRCh38
rs748153334 Chr.6: 131219729-131219729 on Build GRCh38
rs973755797 Chr.6: 131219849-131219850 on Build GRCh38
rs770861526 Chr.6: 131219827-131219830 on Build GRCh38
rs750480907 Chr.6: 131219694-131219694 on Build GRCh38
rs201234046 Chr.6: 131219808-131219808 on Build GRCh38
rs939206550 Chr.6: 131219998-131219998 on Build GRCh38
rs530547185 Chr.6: 131219651-131219651 on Build GRCh38
rs752273563 Chr.6: 131219645-131219646 on Build GRCh38
rs368804335 Chr.6: 131219836-131219836 on Build GRCh38
rs751409940 Chr.6: 131219717-131219717 on Build GRCh38
rs765730212 Chr.6: 131219807-131219807 on Build GRCh38
rs1038375928 Chr.6: 131220048-131220048 on Build GRCh38
rs949517599 Chr.6: 131219951-131219951 on Build GRCh38
rs74565557 Chr.6: 131220118-131220118 on Build GRCh38
rs574004835 Chr.6: 131220025-131220025 on Build GRCh38
rs200010554 Chr.6: 131219687-131219687 on Build GRCh38
rs147577265 Chr.6: 131219733-131219733 on Build GRCh38
rs781181072 Chr.6: 131219825-131219825 on Build GRCh38
rs996274838 Chr.6: 131220063-131220063 on Build GRCh38
rs140783900 Chr.6: 131219734-131219734 on Build GRCh38
rs762271442 Chr.6: 131219780-131219780 on Build GRCh38
rs537192546 Chr.6: 131219895-131219895 on Build GRCh38
rs1046912924 Chr.6: 131220005-131220005 on Build GRCh38
rs769069491 Chr.6: 131219747-131219747 on Build GRCh38
rs914914179 Chr.6: 131219914-131219914 on Build GRCh38
rs200644883 Chr.6: 131219730-131219730 on Build GRCh38
rs751431804 Chr.6: 131219656-131219656 on Build GRCh38
rs959132788 Chr.6: 131219901-131219901 on Build GRCh38
rs757909211 Chr.6: 131219725-131219727 on Build GRCh38
rs577536424 Chr.6: 131220098-131220098 on Build GRCh38
rs769252465 Chr.6: 131219645-131219645 on Build GRCh38
rs1005571118 Chr.6: 131219716-131219716 on Build GRCh38
rs1029910345 Chr.6: 131219791-131219791 on Build GRCh38
rs755236901 Chr.6: 131219818-131219818 on Build GRCh38
rs377486720 Chr.6: 131219646-131219646 on Build GRCh38
rs746940529 Chr.6: 131219809-131219812 on Build GRCh38
rs747664668 Chr.6: 131219644-131219644 on Build GRCh38
rs776649944 Chr.6: 131219643-131219643 on Build GRCh38
rs765285496 Chr.6: 131219692-131219692 on Build GRCh38
rs762553677 Chr.6: 131219647-131219647 on Build GRCh38
rs754411055 Chr.6: 131219673-131219673 on Build GRCh38
rs891359071 Chr.6: 131220115-131220115 on Build GRCh38
rs996792957 Chr.6: 131220096-131220096 on Build GRCh38
rs1031191294 Chr.6: 131219860-131219860 on Build GRCh38
rs1053774861 Chr.6: 131220035-131220035 on Build GRCh38
rs763276020 Chr.6: 131219679-131219679 on Build GRCh38
rs990503209 Chr.6: 131219911-131219911 on Build GRCh38
rs899880807 Chr.6: 131220067-131220067 on Build GRCh38
rs776961748 Chr.6: 131219772-131219772 on Build GRCh38
rs9321278 Chr.6: 131219826-131219826 on Build GRCh38
rs757773619 Chr.6: 131219677-131219677 on Build GRCh38
rs947699026 Chr.6: 131219931-131219931 on Build GRCh38
rs549046787 Chr.6: 131219706-131219706 on Build GRCh38
rs766710915 Chr.6: 131219813-131219813 on Build GRCh38
rs1042629217 Chr.6: 131219977-131219977 on Build GRCh38
rs552714488 Chr.6: 131220069-131220069 on Build GRCh38
rs777180727 Chr.6: 131219731-131219731 on Build GRCh38
rs566383205 Chr.6: 131219739-131219739 on Build GRCh38
rs780371314 Chr.6: 131219712-131219712 on Build GRCh38
rs534519645 Chr.6: 131220060-131220060 on Build GRCh38
rs879452709 Chr.6: 131220038-131220038 on Build GRCh38
rs919707608 Chr.6: 131219898-131219898 on Build GRCh38
rs1030354659 Chr.6: 131220105-131220105 on Build GRCh38
rs767626434 Chr.6: 131219952-131219952 on Build GRCh38
rs113424612 Chr.6: 131220126-131220126 on Build GRCh38
rs145033953 Chr.6: 131219709-131219709 on Build GRCh38
rs562183592 Chr.6: 131219726-131219726 on Build GRCh38
rs551934342 Chr.6: 131219988-131219988 on Build GRCh38
rs949653475 Chr.6: 131219653-131219653 on Build GRCh38
rs546260268 Chr.6: 131219655-131219655 on Build GRCh38
rs756028361 Chr.6: 131219847-131219847 on Build GRCh38
rs1015551906 Chr.6: 131220133-131220133 on Build GRCh38
rs956461596 Chr.6: 131219863-131219863 on Build GRCh38

View Full Product Details