Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134477921-134478464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC13
Entrez Gene ID
25847
Gene Name
anaphase promoting complex subunit 13
Gene Aliases
APC13, SWM1
UniGene
Hs.106909
Gene Chromosome Location
Chr.3: 134477704-134486023 on Build GRCh38
Transcripts
NM_001242374.1 NM_001242375.1 NM_015391.3

SNP Detail

SNP ID Chromosome Location
rs751005904 Chr.3: 134478440-134478440 on Build GRCh38
rs940418767 Chr.3: 134478118-134478118 on Build GRCh38
rs763861003 Chr.3: 134478345-134478345 on Build GRCh38
rs563874669 Chr.3: 134478071-134478071 on Build GRCh38
rs894377250 Chr.3: 134478251-134478251 on Build GRCh38
rs766861108 Chr.3: 134478202-134478202 on Build GRCh38
rs1033156824 Chr.3: 134478397-134478397 on Build GRCh38
rs139367774 Chr.3: 134478398-134478398 on Build GRCh38
rs776984222 Chr.3: 134478006-134478006 on Build GRCh38
rs902481275 Chr.3: 134478443-134478443 on Build GRCh38
rs755553613 Chr.3: 134478026-134478026 on Build GRCh38
rs775212879 Chr.3: 134478206-134478206 on Build GRCh38
rs546237017 Chr.3: 134478171-134478171 on Build GRCh38
rs987153773 Chr.3: 134478096-134478096 on Build GRCh38
rs978025022 Chr.3: 134477969-134477969 on Build GRCh38
rs1014709339 Chr.3: 134478257-134478257 on Build GRCh38
rs564717554 Chr.3: 134478320-134478320 on Build GRCh38
rs150525719 Chr.3: 134478286-134478286 on Build GRCh38
rs887563981 Chr.3: 134478283-134478283 on Build GRCh38
rs1021402510 Chr.3: 134478020-134478022 on Build GRCh38
rs998147891 Chr.3: 134478386-134478386 on Build GRCh38
rs141870909 Chr.3: 134478014-134478014 on Build GRCh38
rs371499164 Chr.3: 134478106-134478106 on Build GRCh38
rs966713805 Chr.3: 134478043-134478043 on Build GRCh38
rs1039635723 Chr.3: 134478346-134478346 on Build GRCh38
rs1024299581 Chr.3: 134478285-134478285 on Build GRCh38
rs921505940 Chr.3: 134478018-134478018 on Build GRCh38
rs981393101 Chr.3: 134478151-134478151 on Build GRCh38
rs967484207 Chr.3: 134477960-134477960 on Build GRCh38
rs755144972 Chr.3: 134478425-134478426 on Build GRCh38
rs1032434852 Chr.3: 134478085-134478085 on Build GRCh38
rs975536555 Chr.3: 134478406-134478406 on Build GRCh38
rs1055748429 Chr.3: 134478250-134478250 on Build GRCh38
rs957183893 Chr.3: 134478093-134478093 on Build GRCh38
rs991359118 Chr.3: 134478105-134478105 on Build GRCh38
rs975610163 Chr.3: 134478012-134478012 on Build GRCh38
rs902081389 Chr.3: 134478381-134478381 on Build GRCh38
rs1036607415 Chr.3: 134478164-134478164 on Build GRCh38
rs761069990 Chr.3: 134478028-134478028 on Build GRCh38
rs950035961 Chr.3: 134478149-134478149 on Build GRCh38
rs144044996 Chr.3: 134478210-134478210 on Build GRCh38
rs1033144679 Chr.3: 134477979-134477979 on Build GRCh38

View Full Product Details