Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93993417-93994006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BET1
Entrez Gene ID
10282
Gene Name
Bet1 golgi vesicular membrane trafficking protein
Gene Aliases
HBET1
UniGene
Hs.489132
Gene Chromosome Location
Chr.7: 93962770-94004382 on Build GRCh38
Transcripts
NM_001317739.1 NM_005868.5 NR_133908.1 NR_133909.1

SNP Detail

SNP ID Chromosome Location
rs768693592 Chr.7: 93993924-93993924 on Build GRCh38
rs543342019 Chr.7: 93993645-93993645 on Build GRCh38
rs573509116 Chr.7: 93993652-93993652 on Build GRCh38
rs897084171 Chr.7: 93993929-93993929 on Build GRCh38
rs939117973 Chr.7: 93993708-93993708 on Build GRCh38
rs563231481 Chr.7: 93993651-93993651 on Build GRCh38
rs966401114 Chr.7: 93993702-93993702 on Build GRCh38
rs575220648 Chr.7: 93993628-93993628 on Build GRCh38
rs867228617 Chr.7: 93993945-93993945 on Build GRCh38
rs1009042044 Chr.7: 93993622-93993622 on Build GRCh38
rs558877959 Chr.7: 93993467-93993467 on Build GRCh38
rs964977021 Chr.7: 93993657-93993657 on Build GRCh38
rs929267429 Chr.7: 93993960-93993960 on Build GRCh38
rs771038817 Chr.7: 93993946-93993946 on Build GRCh38
rs927660069 Chr.7: 93993707-93993707 on Build GRCh38
rs745385088 Chr.7: 93993552-93993552 on Build GRCh38
rs916430881 Chr.7: 93993868-93993868 on Build GRCh38
rs779888237 Chr.7: 93993866-93993866 on Build GRCh38
rs990642108 Chr.7: 93993713-93993713 on Build GRCh38
rs977760898 Chr.7: 93993703-93993703 on Build GRCh38
rs777714003 Chr.7: 93993936-93993936 on Build GRCh38
rs35788669 Chr.7: 93993610-93993610 on Build GRCh38
rs528151776 Chr.7: 93993848-93993848 on Build GRCh38
rs147576087 Chr.7: 93993968-93993968 on Build GRCh38
rs890541143 Chr.7: 93993602-93993602 on Build GRCh38
rs993532698 Chr.7: 93993534-93993534 on Build GRCh38
rs1037696908 Chr.7: 93993455-93993455 on Build GRCh38
rs1021966092 Chr.7: 93993685-93993685 on Build GRCh38
rs751646614 Chr.7: 93993771-93993771 on Build GRCh38
rs200336741 Chr.7: 93993888-93993888 on Build GRCh38
rs771642519 Chr.7: 93993594-93993594 on Build GRCh38
rs113320403 Chr.7: 93993662-93993662 on Build GRCh38
rs764209523 Chr.7: 93993523-93993523 on Build GRCh38
rs183070347 Chr.7: 93993911-93993911 on Build GRCh38
rs559204290 Chr.7: 93993701-93993701 on Build GRCh38
rs565189520 Chr.7: 93993488-93993488 on Build GRCh38
rs941271585 Chr.7: 93993876-93993876 on Build GRCh38
rs770108629 Chr.7: 93993940-93993940 on Build GRCh38
rs530513629 Chr.7: 93993943-93993943 on Build GRCh38

View Full Product Details