Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107566972-107567431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS4L
Entrez Gene ID
11062
Gene Name
dihydrouridine synthase 4 like
Gene Aliases
DUS4, PP35
UniGene
Hs.97627
Gene Chromosome Location
Chr.7: 107563957-107578523 on Build GRCh38
Transcripts
NM_001270419.1 NM_181581.2 NR_073002.1 NR_073003.1 NR_073004.1 NR_073005.1 XM_005250125.4 XM_006715844.2 XM_006715845.3 XM_011515766.2 XM_011515767.2 XM_017011708.1 XM_017011709.1 XM_017011710.1 XM_017011711.1 XM_017011712.1 XM_017011713.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4791020 c.28A>G p.I10V Chr.7: 107567098-107567098 on Build GRCh38
5557723 c.48A>G p.K16K Chr.7: 107567118-107567118 on Build GRCh38
3254769 c.41_42insA p.D17fs*27 Chr.7: 107567111-107567112 on Build GRCh38
1699159 c.106C>T p.R36* Chr.7: 107567176-107567176 on Build GRCh38
1699158 c.52C>T p.P18S Chr.7: 107567122-107567122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184421763 Chr.7: 107567340-107567340 on Build GRCh38
rs775695021 Chr.7: 107567205-107567205 on Build GRCh38
rs756085414 Chr.7: 107567113-107567113 on Build GRCh38
rs539607032 Chr.7: 107567343-107567343 on Build GRCh38
rs199908673 Chr.7: 107567197-107567197 on Build GRCh38
rs563328208 Chr.7: 107567027-107567027 on Build GRCh38
rs962024903 Chr.7: 107567133-107567133 on Build GRCh38
rs531104020 Chr.7: 107567275-107567275 on Build GRCh38
rs576042437 Chr.7: 107567067-107567067 on Build GRCh38
rs560362034 Chr.7: 107567273-107567273 on Build GRCh38
rs769741882 Chr.7: 107567153-107567154 on Build GRCh38
rs117550357 Chr.7: 107567249-107567249 on Build GRCh38
rs776857473 Chr.7: 107567068-107567068 on Build GRCh38
rs752666229 Chr.7: 107567110-107567110 on Build GRCh38
rs763383830 Chr.7: 107567182-107567182 on Build GRCh38
rs773862728 Chr.7: 107567032-107567032 on Build GRCh38
rs200681046 Chr.7: 107567194-107567194 on Build GRCh38
rs746130506 Chr.7: 107567064-107567065 on Build GRCh38
rs781424804 Chr.7: 107567059-107567060 on Build GRCh38
rs551304855 Chr.7: 107567277-107567277 on Build GRCh38
rs745462122 Chr.7: 107567168-107567168 on Build GRCh38
rs959428133 Chr.7: 107567333-107567333 on Build GRCh38
rs771648744 Chr.7: 107567052-107567052 on Build GRCh38
rs528209740 Chr.7: 107567242-107567242 on Build GRCh38
rs752823751 Chr.7: 107567236-107567236 on Build GRCh38
rs771738749 Chr.7: 107567170-107567170 on Build GRCh38
rs773584666 Chr.7: 107567181-107567181 on Build GRCh38
rs900253586 Chr.7: 107567015-107567015 on Build GRCh38
rs1038715891 Chr.7: 107567014-107567014 on Build GRCh38
rs201466996 Chr.7: 107567094-107567094 on Build GRCh38
rs777504979 Chr.7: 107567020-107567020 on Build GRCh38
rs199510674 Chr.7: 107567152-107567152 on Build GRCh38
rs4730250 Chr.7: 107567250-107567250 on Build GRCh38
rs761650189 Chr.7: 107567224-107567224 on Build GRCh38
rs371705355 Chr.7: 107567024-107567024 on Build GRCh38
rs376721875 Chr.7: 107567093-107567093 on Build GRCh38
rs767541229 Chr.7: 107567105-107567105 on Build GRCh38
rs779747286 Chr.7: 107567175-107567175 on Build GRCh38
rs778639375 Chr.7: 107567154-107567154 on Build GRCh38
rs777666666 Chr.7: 107567115-107567115 on Build GRCh38
rs918559827 Chr.7: 107567202-107567202 on Build GRCh38
rs770479604 Chr.7: 107567177-107567177 on Build GRCh38
rs777299916 Chr.7: 107567061-107567061 on Build GRCh38
rs767696689 Chr.7: 107567229-107567229 on Build GRCh38
rs759627784 Chr.7: 107567096-107567096 on Build GRCh38
rs375742817 Chr.7: 107567203-107567203 on Build GRCh38
rs753726431 Chr.7: 107567119-107567119 on Build GRCh38
rs970969941 Chr.7: 107567209-107567209 on Build GRCh38
rs577153490 Chr.7: 107567176-107567176 on Build GRCh38
rs756933796 Chr.7: 107567144-107567144 on Build GRCh38
rs765804979 Chr.7: 107567062-107567062 on Build GRCh38

View Full Product Details