Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 112775521-112775693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APC
Entrez Gene ID
324
Gene Name
APC, WNT signaling pathway regulator
Gene Aliases
BTPS2, DP2, DP2.5, DP3, GS, PPP1R46
UniGene
Hs.158932
Gene Chromosome Location
Chr.5: 112707505-112846239 on Build GRCh38
Transcripts
NM_000038.5 NM_001127510.2 NM_001127511.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
588989 c.423-1G>T p.? Chr.5: 112775628-112775628 on Build GRCh38
4934835 c.477C>G p.Y159* Chr.5: 112775683-112775683 on Build GRCh38
1432142 c.423-2A>G p.? Chr.5: 112775627-112775627 on Build GRCh38
19338 c.423-4delA p.? Chr.5: 112775625-112775625 on Build GRCh38
6602019 c.477C>A p.Y159* Chr.5: 112775683-112775683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763181290 Chr.5: 112775642-112775642 on Build GRCh38
rs925761509 Chr.5: 112775600-112775600 on Build GRCh38
rs750821213 Chr.5: 112775653-112775653 on Build GRCh38
rs915573166 Chr.5: 112775558-112775558 on Build GRCh38
rs863224820 Chr.5: 112775659-112775659 on Build GRCh38
rs774835466 Chr.5: 112775613-112775615 on Build GRCh38
rs771946819 Chr.5: 112775593-112775593 on Build GRCh38
rs587782293 Chr.5: 112775626-112775626 on Build GRCh38
rs558649487 Chr.5: 112775587-112775587 on Build GRCh38
rs967226219 Chr.5: 112775669-112775669 on Build GRCh38
rs769039873 Chr.5: 112775646-112775646 on Build GRCh38
rs786202822 Chr.5: 112775666-112775666 on Build GRCh38
rs730881230 Chr.5: 112775625-112775625 on Build GRCh38
rs397514031 Chr.5: 112775628-112775628 on Build GRCh38
rs762062860 Chr.5: 112775649-112775649 on Build GRCh38
rs371085910 Chr.5: 112775655-112775655 on Build GRCh38
rs769942791 Chr.5: 112775606-112775606 on Build GRCh38
rs763324377 Chr.5: 112775612-112775613 on Build GRCh38
rs878853444 Chr.5: 112775654-112775654 on Build GRCh38
rs534684461 Chr.5: 112775612-112775612 on Build GRCh38
rs764501133 Chr.5: 112775613-112775614 on Build GRCh38
rs778972617 Chr.5: 112775583-112775583 on Build GRCh38
rs863225354 Chr.5: 112775626-112775627 on Build GRCh38
rs774964663 Chr.5: 112775648-112775648 on Build GRCh38
rs876657408 Chr.5: 112775624-112775626 on Build GRCh38
rs116020626 Chr.5: 112775656-112775656 on Build GRCh38
rs863224458 Chr.5: 112775629-112775629 on Build GRCh38
rs536641097 Chr.5: 112775546-112775546 on Build GRCh38
rs587782557 Chr.5: 112775632-112775633 on Build GRCh38
rs35031194 Chr.5: 112775612-112775612 on Build GRCh38
rs201918368 Chr.5: 112775605-112775606 on Build GRCh38
rs78919815 Chr.5: 112775613-112775613 on Build GRCh38
rs1033648774 Chr.5: 112775608-112775608 on Build GRCh38
rs863225355 Chr.5: 112775656-112775659 on Build GRCh38
rs570467572 Chr.5: 112775601-112775601 on Build GRCh38
rs978697620 Chr.5: 112775597-112775597 on Build GRCh38
rs864622482 Chr.5: 112775665-112775665 on Build GRCh38
rs879254087 Chr.5: 112775627-112775627 on Build GRCh38
rs775651105 Chr.5: 112775639-112775639 on Build GRCh38
rs754553913 Chr.5: 112775664-112775664 on Build GRCh38
rs767875993 Chr.5: 112775651-112775651 on Build GRCh38
rs781151009 Chr.5: 112775550-112775550 on Build GRCh38
rs753336993 Chr.5: 112775662-112775662 on Build GRCh38
rs768759499 Chr.5: 112775562-112775562 on Build GRCh38

View Full Product Details