Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108317349-108317529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATM
Entrez Gene ID
472
Gene Name
ATM serine/threonine kinase
Gene Aliases
AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
UniGene
Hs.367437
Gene Chromosome Location
Chr.11: 108222484-108369102 on Build GRCh38
Transcripts
NM_000051.3 XM_005271561.4 XM_005271562.4 XM_006718843.3 XM_006718845.1 XM_011542840.2 XM_011542842.2 XM_011542843.2 XM_011542844.2 XM_011542845.1 XM_017017789.1 XM_017017790.1 XM_017017791.1 XM_017017249.1 XM_017017250.1

Gene Symbol
C11orf65
Entrez Gene ID
160140
Gene Name
chromosome 11 open reading frame 65
Gene Aliases
-
UniGene
Hs.653180
Gene Chromosome Location
Chr.11: 108307344-108469832 on Build GRCh38
Transcripts
NM_000051.3 XM_005271561.4 XM_005271562.4 XM_006718843.3 XM_006718845.1 XM_011542840.2 XM_011542842.2 XM_011542843.2 XM_011542844.2 XM_011542845.1 XM_017017789.1 XM_017017790.1 XM_017017791.1 XM_017017249.1 XM_017017250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6493987 c.6235G>A p.V2079I Chr.11: 108317409-108317409 on Build GRCh38
1183964 c.6247G>T p.G2083* Chr.11: 108317421-108317421 on Build GRCh38
5739442 c.6327G>T p.W2109C Chr.11: 108317501-108317501 on Build GRCh38
1739756 c.6311G>A p.W2104* Chr.11: 108317485-108317485 on Build GRCh38
3666003 c.6291A>T p.E2097D Chr.11: 108317465-108317465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs864622730 Chr.11: 108317492-108317492 on Build GRCh38
rs928129862 Chr.11: 108317385-108317385 on Build GRCh38
rs765763407 Chr.11: 108317495-108317495 on Build GRCh38
rs759029705 Chr.11: 108317504-108317504 on Build GRCh38
rs587779855 Chr.11: 108317413-108317413 on Build GRCh38
rs765592353 Chr.11: 108317381-108317382 on Build GRCh38
rs752478345 Chr.11: 108317393-108317393 on Build GRCh38
rs773891864 Chr.11: 108317497-108317497 on Build GRCh38
rs587780632 Chr.11: 108317489-108317489 on Build GRCh38
rs879253983 Chr.11: 108317487-108317487 on Build GRCh38
rs772608345 Chr.11: 108317424-108317424 on Build GRCh38
rs747372417 Chr.11: 108317438-108317438 on Build GRCh38
rs587779854 Chr.11: 108317406-108317406 on Build GRCh38
rs915524411 Chr.11: 108317442-108317442 on Build GRCh38
rs376898203 Chr.11: 108317429-108317429 on Build GRCh38
rs863224577 Chr.11: 108317441-108317441 on Build GRCh38
rs878853529 Chr.11: 108317413-108317414 on Build GRCh38
rs587780631 Chr.11: 108317467-108317467 on Build GRCh38
rs587782802 Chr.11: 108317470-108317470 on Build GRCh38
rs876658300 Chr.11: 108317506-108317506 on Build GRCh38
rs587782114 Chr.11: 108317500-108317500 on Build GRCh38
rs745977589 Chr.11: 108317420-108317420 on Build GRCh38
rs946942912 Chr.11: 108317451-108317451 on Build GRCh38
rs730881379 Chr.11: 108317427-108317427 on Build GRCh38
rs786203008 Chr.11: 108317402-108317402 on Build GRCh38
rs569483748 Chr.11: 108317408-108317408 on Build GRCh38
rs876658700 Chr.11: 108317426-108317426 on Build GRCh38
rs779082836 Chr.11: 108317410-108317410 on Build GRCh38
rs786204173 Chr.11: 108317407-108317407 on Build GRCh38
rs876660164 Chr.11: 108317475-108317475 on Build GRCh38
rs867760244 Chr.11: 108317501-108317501 on Build GRCh38
rs876659325 Chr.11: 108317477-108317477 on Build GRCh38
rs1032544874 Chr.11: 108317503-108317503 on Build GRCh38
rs141370828 Chr.11: 108317453-108317453 on Build GRCh38
rs587780633 Chr.11: 108317491-108317491 on Build GRCh38
rs730881380 Chr.11: 108317431-108317431 on Build GRCh38
rs755973863 Chr.11: 108317400-108317400 on Build GRCh38
rs767406075 Chr.11: 108317391-108317391 on Build GRCh38
rs1800060 Chr.11: 108317409-108317409 on Build GRCh38

View Full Product Details