Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73568066-73568613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT2
Entrez Gene ID
10965
Gene Name
acyl-CoA thioesterase 2
Gene Aliases
CTE-IA, CTE1A, MTE1, PTE2, PTE2A, ZAP128
UniGene
Hs.446685
Gene Chromosome Location
Chr.14: 73567620-73575658 on Build GRCh38
Transcripts
NR_046028.1 XM_017021289.1

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
UniGene
Hs.660774
Gene Chromosome Location
Chr.14: 73478484-73634418 on Build GRCh38
Transcripts
NR_046028.1 XM_017021289.1

SNP Detail

SNP ID Chromosome Location
rs1004815145 Chr.14: 73568315-73568315 on Build GRCh38
rs927616759 Chr.14: 73568555-73568555 on Build GRCh38
rs775704338 Chr.14: 73568324-73568324 on Build GRCh38
rs946339641 Chr.14: 73568109-73568109 on Build GRCh38
rs570168063 Chr.14: 73568261-73568261 on Build GRCh38
rs397732512 Chr.14: 73568363-73568364 on Build GRCh38
rs990295960 Chr.14: 73568441-73568441 on Build GRCh38
rs530533803 Chr.14: 73568563-73568563 on Build GRCh38
rs187386274 Chr.14: 73568191-73568191 on Build GRCh38
rs994658103 Chr.14: 73568225-73568225 on Build GRCh38
rs573950240 Chr.14: 73568375-73568375 on Build GRCh38
rs35487534 Chr.14: 73568566-73568566 on Build GRCh38
rs552834684 Chr.14: 73568455-73568455 on Build GRCh38
rs577457678 Chr.14: 73568486-73568486 on Build GRCh38
rs770811494 Chr.14: 73568450-73568450 on Build GRCh38
rs200631445 Chr.14: 73568566-73568567 on Build GRCh38
rs566311373 Chr.14: 73568196-73568196 on Build GRCh38
rs759742492 Chr.14: 73568158-73568161 on Build GRCh38
rs544802526 Chr.14: 73568498-73568498 on Build GRCh38
rs746966289 Chr.14: 73568421-73568421 on Build GRCh38
rs759228765 Chr.14: 73568566-73568566 on Build GRCh38
rs981718570 Chr.14: 73568533-73568533 on Build GRCh38
rs34209744 Chr.14: 73568352-73568353 on Build GRCh38
rs370100961 Chr.14: 73568363-73568363 on Build GRCh38
rs542676742 Chr.14: 73568581-73568581 on Build GRCh38
rs780589678 Chr.14: 73568166-73568166 on Build GRCh38
rs537512709 Chr.14: 73568266-73568266 on Build GRCh38
rs146058315 Chr.14: 73568212-73568212 on Build GRCh38
rs966111690 Chr.14: 73568395-73568395 on Build GRCh38
rs540998984 Chr.14: 73568431-73568431 on Build GRCh38
rs530157450 Chr.14: 73568171-73568171 on Build GRCh38
rs916354372 Chr.14: 73568567-73568567 on Build GRCh38
rs1054838669 Chr.14: 73568192-73568192 on Build GRCh38
rs777770539 Chr.14: 73568390-73568390 on Build GRCh38
rs776528942 Chr.14: 73568529-73568529 on Build GRCh38
rs906204632 Chr.14: 73568178-73568178 on Build GRCh38
rs544409574 Chr.14: 73568439-73568439 on Build GRCh38
rs996657332 Chr.14: 73568404-73568405 on Build GRCh38
rs140027599 Chr.14: 73568235-73568235 on Build GRCh38
rs776882421 Chr.14: 73568103-73568104 on Build GRCh38
rs543575680 Chr.14: 73568364-73568364 on Build GRCh38
rs181940651 Chr.14: 73568180-73568180 on Build GRCh38
rs116347091 Chr.14: 73568587-73568587 on Build GRCh38
rs75098542 Chr.14: 73568325-73568325 on Build GRCh38
rs773520592 Chr.14: 73568565-73568566 on Build GRCh38
rs968002678 Chr.14: 73568500-73568500 on Build GRCh38
rs886333926 Chr.14: 73568302-73568302 on Build GRCh38
rs191206858 Chr.14: 73568514-73568514 on Build GRCh38

View Full Product Details