Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 88630270-88630807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AEN
Entrez Gene ID
64782
Gene Name
apoptosis enhancing nuclease
Gene Aliases
ISG20L1, pp12744
UniGene
Hs.436102
Gene Chromosome Location
Chr.15: 88604601-88632281 on Build GRCh38
Transcripts
NM_022767.3 XM_005254966.1 XM_005254967.1 XM_006720645.3 XM_011521905.2 XM_017022489.1 XM_017022490.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6025642 c.909-4G>A p.? Chr.15: 88630379-88630379 on Build GRCh38
7341076 c.909+1G>A p.? Chr.15: 88630384-88630384 on Build GRCh38
4563400 c.971G>A p.R324K Chr.15: 88630287-88630287 on Build GRCh38
6555208 c.958G>T p.A320S Chr.15: 88630274-88630274 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1041753690 Chr.15: 88630690-88630690 on Build GRCh38
rs1033216186 Chr.15: 88630437-88630437 on Build GRCh38
rs1000355738 Chr.15: 88630436-88630436 on Build GRCh38
rs764732027 Chr.15: 88630297-88630297 on Build GRCh38
rs1001333505 Chr.15: 88630634-88630634 on Build GRCh38
rs1033167130 Chr.15: 88630733-88630733 on Build GRCh38
rs970820738 Chr.15: 88630472-88630472 on Build GRCh38
rs775345426 Chr.15: 88630364-88630365 on Build GRCh38
rs745506587 Chr.15: 88630301-88630313 on Build GRCh38
rs1020788016 Chr.15: 88630431-88630431 on Build GRCh38
rs958927218 Chr.15: 88630453-88630453 on Build GRCh38
rs759169829 Chr.15: 88630363-88630363 on Build GRCh38
rs916222551 Chr.15: 88630469-88630469 on Build GRCh38
rs942233016 Chr.15: 88630726-88630726 on Build GRCh38
rs931871010 Chr.15: 88630549-88630549 on Build GRCh38
rs773826202 Chr.15: 88630732-88630732 on Build GRCh38
rs918165251 Chr.15: 88630651-88630651 on Build GRCh38
rs890420227 Chr.15: 88630617-88630617 on Build GRCh38
rs370022294 Chr.15: 88630335-88630335 on Build GRCh38
rs781200131 Chr.15: 88630304-88630304 on Build GRCh38
rs114926971 Chr.15: 88630424-88630424 on Build GRCh38
rs899080269 Chr.15: 88630364-88630364 on Build GRCh38
rs3743474 Chr.15: 88630501-88630501 on Build GRCh38
rs970985667 Chr.15: 88630674-88630674 on Build GRCh38
rs1009319096 Chr.15: 88630653-88630653 on Build GRCh38
rs996072467 Chr.15: 88630387-88630387 on Build GRCh38
rs988323816 Chr.15: 88630358-88630358 on Build GRCh38
rs1008063022 Chr.15: 88630616-88630616 on Build GRCh38
rs919696665 Chr.15: 88630776-88630776 on Build GRCh38
rs981846077 Chr.15: 88630485-88630485 on Build GRCh38
rs551263710 Chr.15: 88630540-88630540 on Build GRCh38
rs757563706 Chr.15: 88630767-88630767 on Build GRCh38
rs771813576 Chr.15: 88630348-88630362 on Build GRCh38
rs67426590 Chr.15: 88630707-88630707 on Build GRCh38
rs756213497 Chr.15: 88630316-88630316 on Build GRCh38
rs929590764 Chr.15: 88630783-88630783 on Build GRCh38
rs140899692 Chr.15: 88630362-88630362 on Build GRCh38
rs757651205 Chr.15: 88630303-88630303 on Build GRCh38
rs769592866 Chr.15: 88630459-88630459 on Build GRCh38
rs1032440074 Chr.15: 88630637-88630637 on Build GRCh38
rs752664845 Chr.15: 88630507-88630507 on Build GRCh38
rs773216515 Chr.15: 88630334-88630334 on Build GRCh38
rs770791689 Chr.15: 88630352-88630352 on Build GRCh38
rs386786162 Chr.15: 88630350-88630364 on Build GRCh38
rs1045245593 Chr.15: 88630405-88630405 on Build GRCh38
rs1050322284 Chr.15: 88630608-88630608 on Build GRCh38
rs1018154909 Chr.15: 88630626-88630626 on Build GRCh38
rs949514466 Chr.15: 88630401-88630401 on Build GRCh38
rs186259223 Chr.15: 88630322-88630322 on Build GRCh38
rs748849975 Chr.15: 88630650-88630650 on Build GRCh38
rs373412213 Chr.15: 88630350-88630362 on Build GRCh38
rs1046587515 Chr.15: 88630339-88630339 on Build GRCh38
rs746120783 Chr.15: 88630305-88630305 on Build GRCh38
rs561628527 Chr.15: 88630425-88630425 on Build GRCh38
rs1037478779 Chr.15: 88630526-88630526 on Build GRCh38
rs757164209 Chr.15: 88630413-88630413 on Build GRCh38
rs920456347 Chr.15: 88630534-88630534 on Build GRCh38
rs764988881 Chr.15: 88630451-88630451 on Build GRCh38
rs751853313 Chr.15: 88630299-88630299 on Build GRCh38
rs752107672 Chr.15: 88630429-88630429 on Build GRCh38
rs147335655 Chr.15: 88630319-88630319 on Build GRCh38
rs964148397 Chr.15: 88630627-88630627 on Build GRCh38
rs565627249 Chr.15: 88630433-88630433 on Build GRCh38
rs898133781 Chr.15: 88630471-88630471 on Build GRCh38
rs956885158 Chr.15: 88630637-88630637 on Build GRCh38
rs988103147 Chr.15: 88630646-88630646 on Build GRCh38
rs3826038 Chr.15: 88630426-88630426 on Build GRCh38
rs17777460 Chr.15: 88630325-88630325 on Build GRCh38
rs780258779 Chr.15: 88630317-88630317 on Build GRCh38

View Full Product Details