Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35660277-35660804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF39
Entrez Gene ID
84904
Gene Name
Rho guanine nucleotide exchange factor 39
Gene Aliases
C9orf100
UniGene
Hs.534579
Gene Chromosome Location
Chr.9: 35658875-35665281 on Build GRCh38
Transcripts
NM_032818.2 XM_011518057.2 XM_017015225.1 XM_017015227.1 NM_001195200.1 NM_001195201.1 NM_001195217.1 NM_174923.2 XM_005251403.4

Gene Symbol
CCDC107
Entrez Gene ID
203260
Gene Name
coiled-coil domain containing 107
Gene Aliases
PSEC0222
UniGene
Hs.745107
Gene Chromosome Location
Chr.9: 35658282-35661503 on Build GRCh38
Transcripts
NM_032818.2 XM_011518057.2 XM_017015225.1 XM_017015227.1 NM_001195200.1 NM_001195201.1 NM_001195217.1 NM_174923.2 XM_005251403.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3367703 c.391C>T p.L131L Chr.9: 35660628-35660628 on Build GRCh38
6837410 c.461G>T p.W154L Chr.9: 35660796-35660796 on Build GRCh38
6115957 c.289G>C p.E97Q Chr.9: 35660431-35660431 on Build GRCh38
1624943 c.330C>A p.S110R Chr.9: 35660472-35660472 on Build GRCh38
1108682 c.363G>T p.E121D Chr.9: 35660600-35660600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148911211 Chr.9: 35660590-35660590 on Build GRCh38
rs147844073 Chr.9: 35660582-35660582 on Build GRCh38
rs776277883 Chr.9: 35660711-35660711 on Build GRCh38
rs768953113 Chr.9: 35660351-35660351 on Build GRCh38
rs769906088 Chr.9: 35660508-35660509 on Build GRCh38
rs200002081 Chr.9: 35660428-35660428 on Build GRCh38
rs762249879 Chr.9: 35660375-35660375 on Build GRCh38
rs776791852 Chr.9: 35660365-35660365 on Build GRCh38
rs561444311 Chr.9: 35660712-35660712 on Build GRCh38
rs762246010 Chr.9: 35660629-35660629 on Build GRCh38
rs749777866 Chr.9: 35660502-35660502 on Build GRCh38
rs746472349 Chr.9: 35660663-35660663 on Build GRCh38
rs780282380 Chr.9: 35660537-35660538 on Build GRCh38
rs142704657 Chr.9: 35660645-35660645 on Build GRCh38
rs747115708 Chr.9: 35660454-35660454 on Build GRCh38
rs776387476 Chr.9: 35660580-35660580 on Build GRCh38
rs903842511 Chr.9: 35660424-35660424 on Build GRCh38
rs41312840 Chr.9: 35660623-35660623 on Build GRCh38
rs756350610 Chr.9: 35660745-35660745 on Build GRCh38
rs78043355 Chr.9: 35660673-35660673 on Build GRCh38
rs764290730 Chr.9: 35660559-35660579 on Build GRCh38
rs767597591 Chr.9: 35660732-35660732 on Build GRCh38
rs748516738 Chr.9: 35660490-35660490 on Build GRCh38
rs945932818 Chr.9: 35660668-35660668 on Build GRCh38
rs759097940 Chr.9: 35660396-35660396 on Build GRCh38
rs774805907 Chr.9: 35660548-35660548 on Build GRCh38
rs777521919 Chr.9: 35660315-35660315 on Build GRCh38
rs772091722 Chr.9: 35660643-35660643 on Build GRCh38
rs183547206 Chr.9: 35660359-35660359 on Build GRCh38
rs754250887 Chr.9: 35660756-35660756 on Build GRCh38
rs895168551 Chr.9: 35660766-35660766 on Build GRCh38
rs778200263 Chr.9: 35660753-35660753 on Build GRCh38
rs757111794 Chr.9: 35660427-35660427 on Build GRCh38
rs748130756 Chr.9: 35660481-35660481 on Build GRCh38
rs758264689 Chr.9: 35660601-35660601 on Build GRCh38
rs1025125926 Chr.9: 35660569-35660569 on Build GRCh38
rs763230921 Chr.9: 35660559-35660559 on Build GRCh38
rs951079458 Chr.9: 35660317-35660318 on Build GRCh38
rs781532634 Chr.9: 35660477-35660477 on Build GRCh38
rs766349044 Chr.9: 35660606-35660606 on Build GRCh38
rs554125657 Chr.9: 35660657-35660657 on Build GRCh38
rs760576729 Chr.9: 35660647-35660647 on Build GRCh38
rs771573288 Chr.9: 35660520-35660520 on Build GRCh38
rs3174266 Chr.9: 35660719-35660719 on Build GRCh38
rs368340247 Chr.9: 35660491-35660491 on Build GRCh38
rs569771089 Chr.9: 35660314-35660314 on Build GRCh38
rs151241381 Chr.9: 35660646-35660646 on Build GRCh38
rs762791127 Chr.9: 35660699-35660699 on Build GRCh38
rs750249625 Chr.9: 35660596-35660596 on Build GRCh38
rs761413390 Chr.9: 35660688-35660688 on Build GRCh38
rs773919247 Chr.9: 35660384-35660384 on Build GRCh38
rs1018877615 Chr.9: 35660552-35660552 on Build GRCh38
rs145508148 Chr.9: 35660322-35660322 on Build GRCh38
rs763651309 Chr.9: 35660574-35660574 on Build GRCh38
rs750409092 Chr.9: 35660432-35660432 on Build GRCh38
rs766218821 Chr.9: 35660702-35660702 on Build GRCh38
rs376989197 Chr.9: 35660661-35660661 on Build GRCh38
rs760290547 Chr.9: 35660568-35660568 on Build GRCh38
rs773737305 Chr.9: 35660498-35660498 on Build GRCh38
rs753563292 Chr.9: 35660425-35660425 on Build GRCh38
rs768700325 Chr.9: 35660459-35660459 on Build GRCh38
rs760477591 Chr.9: 35660386-35660388 on Build GRCh38
rs779956527 Chr.9: 35660449-35660449 on Build GRCh38
rs540631282 Chr.9: 35660418-35660418 on Build GRCh38
rs909674270 Chr.9: 35660376-35660376 on Build GRCh38
rs374274041 Chr.9: 35660443-35660443 on Build GRCh38
rs752285646 Chr.9: 35660413-35660413 on Build GRCh38
rs751677353 Chr.9: 35660592-35660595 on Build GRCh38
rs772932958 Chr.9: 35660695-35660695 on Build GRCh38
rs746141126 Chr.9: 35660324-35660325 on Build GRCh38
rs1010088209 Chr.9: 35660360-35660360 on Build GRCh38
rs764873272 Chr.9: 35660691-35660691 on Build GRCh38
rs41304745 Chr.9: 35660677-35660677 on Build GRCh38
rs377011794 Chr.9: 35660367-35660367 on Build GRCh38
rs756933620 Chr.9: 35660339-35660339 on Build GRCh38
rs752832075 Chr.9: 35660740-35660740 on Build GRCh38
rs879125750 Chr.9: 35660445-35660445 on Build GRCh38
rs370759330 Chr.9: 35660378-35660378 on Build GRCh38
rs767735176 Chr.9: 35660633-35660634 on Build GRCh38
rs574840948 Chr.9: 35660408-35660408 on Build GRCh38
rs768872766 Chr.9: 35660301-35660301 on Build GRCh38
rs200901870 Chr.9: 35660422-35660422 on Build GRCh38
rs201836449 Chr.9: 35660417-35660417 on Build GRCh38
rs374048020 Chr.9: 35660655-35660655 on Build GRCh38
rs751573425 Chr.9: 35660609-35660609 on Build GRCh38

View Full Product Details