Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 50245348-50245856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAH2
Entrez Gene ID
56624
Gene Name
N-acylsphingosine amidohydrolase 2
Gene Aliases
BCDase, HNAC1, LCDase, N-CDase, NCDase
UniGene
Hs.512645
Gene Chromosome Location
Chr.10: 50184861-50251564 on Build GRCh38
Transcripts
NM_001143974.1 NM_019893.2 XM_011539970.2 XM_011539971.2 XM_011539972.2 XM_011539973.2 XM_017016435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1561305 c.174C>A p.T58T Chr.10: 50245408-50245408 on Build GRCh38
1561303 c.173C>A p.T58N Chr.10: 50245409-50245409 on Build GRCh38
6243661 c.98C>A p.T33N Chr.10: 50245427-50245427 on Build GRCh38
2146179 c.154A>T p.T52S Chr.10: 50245428-50245428 on Build GRCh38
6605415 c.227C>T p.T76I Chr.10: 50245355-50245355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898370448 Chr.10: 50245621-50245621 on Build GRCh38
rs56970000 Chr.10: 50245690-50245690 on Build GRCh38
rs774285066 Chr.10: 50245492-50245492 on Build GRCh38
rs748317315 Chr.10: 50245371-50245371 on Build GRCh38
rs538315030 Chr.10: 50245824-50245824 on Build GRCh38
rs891831347 Chr.10: 50245376-50245376 on Build GRCh38
rs943648867 Chr.10: 50245529-50245529 on Build GRCh38
rs530110148 Chr.10: 50245575-50245575 on Build GRCh38
rs1039249569 Chr.10: 50245531-50245531 on Build GRCh38
rs552646578 Chr.10: 50245624-50245624 on Build GRCh38
rs7077551 Chr.10: 50245593-50245593 on Build GRCh38
rs532626556 Chr.10: 50245608-50245608 on Build GRCh38
rs763608304 Chr.10: 50245432-50245432 on Build GRCh38
rs7090968 Chr.10: 50245590-50245590 on Build GRCh38
rs774832323 Chr.10: 50245427-50245427 on Build GRCh38
rs891887798 Chr.10: 50245553-50245553 on Build GRCh38
rs775450897 Chr.10: 50245797-50245797 on Build GRCh38
rs117126937 Chr.10: 50245755-50245755 on Build GRCh38
rs867264637 Chr.10: 50245394-50245394 on Build GRCh38
rs773491153 Chr.10: 50245433-50245434 on Build GRCh38
rs568448388 Chr.10: 50245729-50245729 on Build GRCh38
rs765745051 Chr.10: 50245445-50245445 on Build GRCh38
rs913366900 Chr.10: 50245795-50245795 on Build GRCh38
rs376958631 Chr.10: 50245572-50245572 on Build GRCh38
rs867954994 Chr.10: 50245449-50245449 on Build GRCh38
rs750983912 Chr.10: 50245446-50245446 on Build GRCh38
rs376981875 Chr.10: 50245505-50245505 on Build GRCh38
rs192860010 Chr.10: 50245438-50245438 on Build GRCh38
rs184411323 Chr.10: 50245732-50245732 on Build GRCh38
rs745842176 Chr.10: 50245497-50245497 on Build GRCh38
rs755732025 Chr.10: 50245510-50245510 on Build GRCh38
rs781230478 Chr.10: 50245469-50245469 on Build GRCh38
rs561284314 Chr.10: 50245571-50245571 on Build GRCh38
rs7097483 Chr.10: 50245543-50245543 on Build GRCh38
rs7097319 Chr.10: 50245393-50245393 on Build GRCh38
rs749098238 Chr.10: 50245481-50245481 on Build GRCh38
rs187718568 Chr.10: 50245746-50245746 on Build GRCh38
rs770909136 Chr.10: 50245485-50245485 on Build GRCh38
rs142364715 Chr.10: 50245403-50245403 on Build GRCh38
rs538113005 Chr.10: 50245387-50245387 on Build GRCh38
rs747036724 Chr.10: 50245465-50245465 on Build GRCh38
rs777106815 Chr.10: 50245775-50245776 on Build GRCh38
rs7067625 Chr.10: 50245431-50245431 on Build GRCh38
rs772181094 Chr.10: 50245434-50245434 on Build GRCh38
rs775981687 Chr.10: 50245528-50245528 on Build GRCh38
rs758862345 Chr.10: 50245413-50245413 on Build GRCh38
rs773108125 Chr.10: 50245389-50245389 on Build GRCh38
rs796741079 Chr.10: 50245788-50245788 on Build GRCh38
rs1039171197 Chr.10: 50245583-50245583 on Build GRCh38
rs141713369 Chr.10: 50245386-50245386 on Build GRCh38
rs963675343 Chr.10: 50245719-50245719 on Build GRCh38
rs755249924 Chr.10: 50245467-50245467 on Build GRCh38
rs569459344 Chr.10: 50245385-50245385 on Build GRCh38
rs1014120353 Chr.10: 50245646-50245646 on Build GRCh38
rs370340088 Chr.10: 50245379-50245379 on Build GRCh38
rs970270505 Chr.10: 50245673-50245673 on Build GRCh38
rs188096471 Chr.10: 50245429-50245429 on Build GRCh38
rs7077415 Chr.10: 50245533-50245533 on Build GRCh38
rs910858199 Chr.10: 50245525-50245525 on Build GRCh38
rs771100807 Chr.10: 50245610-50245610 on Build GRCh38
rs957478923 Chr.10: 50245720-50245720 on Build GRCh38
rs374685598 Chr.10: 50245408-50245408 on Build GRCh38
rs780218782 Chr.10: 50245464-50245464 on Build GRCh38
rs770121870 Chr.10: 50245789-50245789 on Build GRCh38
rs7067773 Chr.10: 50245511-50245511 on Build GRCh38
rs372733179 Chr.10: 50245461-50245461 on Build GRCh38
rs1026161935 Chr.10: 50245430-50245430 on Build GRCh38
rs994015959 Chr.10: 50245623-50245623 on Build GRCh38
rs988835035 Chr.10: 50245495-50245495 on Build GRCh38
rs146246912 Chr.10: 50245392-50245392 on Build GRCh38
rs7091001 Chr.10: 50245657-50245657 on Build GRCh38
rs905843837 Chr.10: 50245579-50245579 on Build GRCh38
rs566146974 Chr.10: 50245775-50245775 on Build GRCh38
rs754264015 Chr.10: 50245434-50245434 on Build GRCh38
rs575405935 Chr.10: 50245536-50245536 on Build GRCh38
rs377466392 Chr.10: 50245377-50245377 on Build GRCh38
rs1038545405 Chr.10: 50245823-50245823 on Build GRCh38
rs767204609 Chr.10: 50245415-50245415 on Build GRCh38
rs1053657357 Chr.10: 50245544-50245544 on Build GRCh38
rs913227467 Chr.10: 50245743-50245743 on Build GRCh38
rs1025942019 Chr.10: 50245598-50245598 on Build GRCh38
rs77391963 Chr.10: 50245697-50245697 on Build GRCh38

View Full Product Details