Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12752662-12753185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST2
Entrez Gene ID
54831
Gene Name
bestrophin 2
Gene Aliases
VMD2L1
UniGene
Hs.435611
Gene Chromosome Location
Chr.19: 12751691-12758458 on Build GRCh38
Transcripts
NM_017682.2 XM_005259963.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5854699 c.85T>A p.Y29N Chr.19: 12752677-12752677 on Build GRCh38
159365 c.101G>A p.R34Q Chr.19: 12752693-12752693 on Build GRCh38
6588311 c.111C>G p.L37L Chr.19: 12752703-12752703 on Build GRCh38
3796648 c.90A>T p.K30N Chr.19: 12752682-12752682 on Build GRCh38
4665633 c.143C>T p.A48V Chr.19: 12752735-12752735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767152406 Chr.19: 12753156-12753156 on Build GRCh38
rs754853705 Chr.19: 12753155-12753155 on Build GRCh38
rs749417306 Chr.19: 12752751-12752751 on Build GRCh38
rs904650540 Chr.19: 12752779-12752779 on Build GRCh38
rs778185946 Chr.19: 12752790-12752790 on Build GRCh38
rs755359302 Chr.19: 12752727-12752727 on Build GRCh38
rs369397456 Chr.19: 12752768-12752768 on Build GRCh38
rs200473398 Chr.19: 12752779-12752780 on Build GRCh38
rs745813177 Chr.19: 12752764-12752764 on Build GRCh38
rs924259210 Chr.19: 12752942-12752942 on Build GRCh38
rs977212592 Chr.19: 12752898-12752898 on Build GRCh38
rs997359394 Chr.19: 12752700-12752700 on Build GRCh38
rs553830005 Chr.19: 12752920-12752920 on Build GRCh38
rs751939642 Chr.19: 12752740-12752741 on Build GRCh38
rs1051630689 Chr.19: 12752951-12752951 on Build GRCh38
rs543618777 Chr.19: 12753051-12753051 on Build GRCh38
rs578207279 Chr.19: 12752780-12752780 on Build GRCh38
rs781620029 Chr.19: 12752730-12752730 on Build GRCh38
rs938734865 Chr.19: 12753095-12753104 on Build GRCh38
rs771016953 Chr.19: 12752754-12752754 on Build GRCh38
rs995981699 Chr.19: 12753035-12753035 on Build GRCh38
rs914748337 Chr.19: 12752804-12752804 on Build GRCh38
rs371908216 Chr.19: 12752705-12752705 on Build GRCh38
rs564916635 Chr.19: 12753076-12753076 on Build GRCh38
rs376542599 Chr.19: 12752744-12752744 on Build GRCh38
rs762207106 Chr.19: 12752776-12752777 on Build GRCh38
rs995610195 Chr.19: 12752745-12752745 on Build GRCh38
rs61737518 Chr.19: 12752712-12752712 on Build GRCh38
rs373298957 Chr.19: 12752776-12752776 on Build GRCh38
rs778405514 Chr.19: 12752793-12752794 on Build GRCh38
rs180970735 Chr.19: 12753146-12753146 on Build GRCh38
rs531279376 Chr.19: 12753050-12753050 on Build GRCh38
rs775071591 Chr.19: 12752689-12752689 on Build GRCh38
rs760341052 Chr.19: 12752771-12752771 on Build GRCh38
rs547288935 Chr.19: 12753116-12753116 on Build GRCh38
rs190553647 Chr.19: 12752726-12752726 on Build GRCh38
rs371836716 Chr.19: 12752739-12752739 on Build GRCh38
rs1014533352 Chr.19: 12752851-12752851 on Build GRCh38
rs756350492 Chr.19: 12752743-12752743 on Build GRCh38
rs532466975 Chr.19: 12753103-12753103 on Build GRCh38
rs973254218 Chr.19: 12752914-12752916 on Build GRCh38
rs752076555 Chr.19: 12752781-12752781 on Build GRCh38
rs763605475 Chr.19: 12752693-12752693 on Build GRCh38
rs767790140 Chr.19: 12752783-12752783 on Build GRCh38
rs185140574 Chr.19: 12753147-12753147 on Build GRCh38
rs758059504 Chr.19: 12753102-12753102 on Build GRCh38
rs751446630 Chr.19: 12752849-12752849 on Build GRCh38
rs938640661 Chr.19: 12752775-12752775 on Build GRCh38
rs889067267 Chr.19: 12752952-12752952 on Build GRCh38
rs1026034956 Chr.19: 12752753-12752753 on Build GRCh38
rs947101396 Chr.19: 12752805-12752805 on Build GRCh38
rs756575025 Chr.19: 12752780-12752780 on Build GRCh38
rs760251223 Chr.19: 12752692-12752692 on Build GRCh38
rs767329857 Chr.19: 12752685-12752685 on Build GRCh38
rs774209982 Chr.19: 12752761-12752761 on Build GRCh38
rs1037902812 Chr.19: 12753005-12753005 on Build GRCh38
rs147416801 Chr.19: 12752992-12752992 on Build GRCh38
rs753067576 Chr.19: 12752784-12752784 on Build GRCh38
rs369765091 Chr.19: 12752719-12752719 on Build GRCh38
rs776237135 Chr.19: 12752777-12752777 on Build GRCh38
rs1008543445 Chr.19: 12752832-12752832 on Build GRCh38
rs542784217 Chr.19: 12753007-12753007 on Build GRCh38
rs560975145 Chr.19: 12753010-12753010 on Build GRCh38
rs545699319 Chr.19: 12752786-12752786 on Build GRCh38
rs933013107 Chr.19: 12752944-12752944 on Build GRCh38

View Full Product Details