Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127050671-127051256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN1
Entrez Gene ID
274
Gene Name
bridging integrator 1
Gene Aliases
AMPH2, AMPHL, SH3P9
UniGene
Hs.193163
Gene Chromosome Location
Chr.2: 127048023-127107400 on Build GRCh38
Transcripts
NM_001320632.1 NM_001320633.1 NM_001320634.1 NM_001320640.1 NM_001320641.1 NM_001320642.1 NM_004305.3 NM_139343.2 NM_139344.2 NM_139345.2 NM_139346.2 NM_139347.2 NM_139348.2 NM_139349.2 NM_139350.2 NM_139351.2 XM_005263642.3 XM_005263643.3 XM_005263645.2 XM_006712425.3 XM_006712426.3 XM_006712427.2 XM_006712428.3 XM_006712429.3 XM_006712431.3 XM_006712432.3 XM_006712433.3 XM_006712434.3 XM_011510975.2 XM_017003819.1 XM_017003820.1 XM_017003821.1 XM_017003822.1 XM_017003823.1 XM_017003824.1 XM_017003825.1 XM_017003826.1 XM_017003827.1 XM_017003828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4393912 c.1261C>T p.L421L Chr.2: 127050825-127050825 on Build GRCh38
5860051 c.1462-1G>A p.? Chr.2: 127050913-127050913 on Build GRCh38
5483375 c.1252C>T p.R418C Chr.2: 127050834-127050834 on Build GRCh38
4665719 c.1477G>A p.V493I Chr.2: 127050897-127050897 on Build GRCh38
4084655 c.1461+1G>A p.? Chr.2: 127051153-127051153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372198581 Chr.2: 127051109-127051109 on Build GRCh38
rs905545802 Chr.2: 127051065-127051065 on Build GRCh38
rs932285344 Chr.2: 127050987-127050987 on Build GRCh38
rs371571307 Chr.2: 127050858-127050858 on Build GRCh38
rs759941022 Chr.2: 127050910-127050910 on Build GRCh38
rs768399754 Chr.2: 127050864-127050864 on Build GRCh38
rs567778570 Chr.2: 127051011-127051011 on Build GRCh38
rs770986090 Chr.2: 127051200-127051200 on Build GRCh38
rs547367134 Chr.2: 127051030-127051030 on Build GRCh38
rs767857710 Chr.2: 127051117-127051117 on Build GRCh38
rs140410496 Chr.2: 127051173-127051173 on Build GRCh38
rs200580275 Chr.2: 127051230-127051230 on Build GRCh38
rs115511091 Chr.2: 127051006-127051006 on Build GRCh38
rs532830644 Chr.2: 127050973-127050973 on Build GRCh38
rs765816262 Chr.2: 127051172-127051172 on Build GRCh38
rs748667501 Chr.2: 127050974-127050974 on Build GRCh38
rs560328063 Chr.2: 127050967-127050967 on Build GRCh38
rs780851819 Chr.2: 127050947-127050947 on Build GRCh38
rs757885823 Chr.2: 127050929-127050929 on Build GRCh38
rs1048747444 Chr.2: 127051097-127051097 on Build GRCh38
rs756816560 Chr.2: 127050840-127050840 on Build GRCh38
rs917067067 Chr.2: 127051036-127051036 on Build GRCh38
rs770606419 Chr.2: 127050763-127050763 on Build GRCh38
rs202120244 Chr.2: 127050770-127050770 on Build GRCh38
rs569414275 Chr.2: 127050999-127050999 on Build GRCh38
rs556315073 Chr.2: 127050713-127050713 on Build GRCh38
rs1044499407 Chr.2: 127051059-127051059 on Build GRCh38
rs766615886 Chr.2: 127050833-127050833 on Build GRCh38
rs376629216 Chr.2: 127050940-127050940 on Build GRCh38
rs772434419 Chr.2: 127051206-127051206 on Build GRCh38
rs777452575 Chr.2: 127050837-127050837 on Build GRCh38
rs757908634 Chr.2: 127051156-127051156 on Build GRCh38
rs377119796 Chr.2: 127050758-127050758 on Build GRCh38
rs748326046 Chr.2: 127050953-127050953 on Build GRCh38
rs371346104 Chr.2: 127051202-127051202 on Build GRCh38
rs755351031 Chr.2: 127050836-127050836 on Build GRCh38
rs1053291620 Chr.2: 127050792-127050792 on Build GRCh38
rs771128647 Chr.2: 127050774-127050789 on Build GRCh38
rs766890661 Chr.2: 127050908-127050908 on Build GRCh38
rs765354438 Chr.2: 127050816-127050816 on Build GRCh38
rs181369236 Chr.2: 127051004-127051004 on Build GRCh38
rs774764729 Chr.2: 127050909-127050909 on Build GRCh38
rs920972662 Chr.2: 127050986-127050986 on Build GRCh38
rs865803817 Chr.2: 127051180-127051180 on Build GRCh38
rs771936924 Chr.2: 127050768-127050768 on Build GRCh38
rs749338696 Chr.2: 127050961-127050961 on Build GRCh38
rs774704128 Chr.2: 127050790-127050791 on Build GRCh38
rs368910139 Chr.2: 127051190-127051190 on Build GRCh38
rs753612525 Chr.2: 127050838-127050838 on Build GRCh38
rs754740026 Chr.2: 127050941-127050941 on Build GRCh38
rs552627048 Chr.2: 127050989-127050989 on Build GRCh38
rs763458588 Chr.2: 127050829-127050829 on Build GRCh38
rs34647988 Chr.2: 127051154-127051154 on Build GRCh38
rs764951306 Chr.2: 127051153-127051153 on Build GRCh38
rs375583449 Chr.2: 127050859-127050859 on Build GRCh38
rs972060066 Chr.2: 127051148-127051148 on Build GRCh38
rs148422103 Chr.2: 127050834-127050834 on Build GRCh38
rs750869889 Chr.2: 127050826-127050826 on Build GRCh38
rs761117052 Chr.2: 127051209-127051209 on Build GRCh38
rs372650268 Chr.2: 127051170-127051170 on Build GRCh38
rs747918591 Chr.2: 127051184-127051184 on Build GRCh38
rs898294488 Chr.2: 127051084-127051084 on Build GRCh38
rs773477009 Chr.2: 127050960-127050960 on Build GRCh38
rs983554366 Chr.2: 127050697-127050697 on Build GRCh38
rs777971351 Chr.2: 127051195-127051195 on Build GRCh38
rs1024500063 Chr.2: 127050902-127050902 on Build GRCh38
rs35983514 Chr.2: 127051081-127051082 on Build GRCh38
rs570394977 Chr.2: 127051096-127051096 on Build GRCh38
rs995090564 Chr.2: 127050879-127050879 on Build GRCh38
rs2043798 Chr.2: 127050956-127050956 on Build GRCh38
rs773313892 Chr.2: 127050895-127050895 on Build GRCh38
rs770983720 Chr.2: 127051106-127051106 on Build GRCh38
rs776018720 Chr.2: 127051118-127051118 on Build GRCh38
rs767828834 Chr.2: 127050911-127050911 on Build GRCh38
rs78068615 Chr.2: 127050797-127050797 on Build GRCh38
rs745860741 Chr.2: 127050767-127050767 on Build GRCh38
rs768900274 Chr.2: 127050782-127050782 on Build GRCh38
rs779756862 Chr.2: 127050901-127050901 on Build GRCh38
rs188763849 Chr.2: 127050698-127050698 on Build GRCh38
rs368238742 Chr.2: 127050882-127050882 on Build GRCh38
rs745758782 Chr.2: 127050849-127050849 on Build GRCh38
rs375659793 Chr.2: 127051113-127051113 on Build GRCh38
rs748813574 Chr.2: 127050761-127050761 on Build GRCh38
rs754900286 Chr.2: 127051175-127051175 on Build GRCh38
rs759737221 Chr.2: 127051109-127051109 on Build GRCh38
rs1038397375 Chr.2: 127051120-127051120 on Build GRCh38
rs989924705 Chr.2: 127051181-127051181 on Build GRCh38
rs994477326 Chr.2: 127051091-127051091 on Build GRCh38
rs949865122 Chr.2: 127051038-127051038 on Build GRCh38
rs966514993 Chr.2: 127050723-127050723 on Build GRCh38
rs765870974 Chr.2: 127051220-127051220 on Build GRCh38
rs776757332 Chr.2: 127050787-127050787 on Build GRCh38
rs764789789 Chr.2: 127050924-127050924 on Build GRCh38
rs761873801 Chr.2: 127050791-127050791 on Build GRCh38
rs778580743 Chr.2: 127050844-127050844 on Build GRCh38
rs772939462 Chr.2: 127051216-127051216 on Build GRCh38
rs1052980508 Chr.2: 127051032-127051032 on Build GRCh38
rs147182000 Chr.2: 127050856-127050856 on Build GRCh38
rs746346952 Chr.2: 127051203-127051203 on Build GRCh38
rs749531373 Chr.2: 127051196-127051196 on Build GRCh38
rs780918654 Chr.2: 127051176-127051176 on Build GRCh38
rs146032225 Chr.2: 127051027-127051027 on Build GRCh38
rs760758221 Chr.2: 127050776-127050776 on Build GRCh38
rs368377494 Chr.2: 127051116-127051116 on Build GRCh38
rs200384643 Chr.2: 127050915-127050915 on Build GRCh38
rs35557593 Chr.2: 127051075-127051075 on Build GRCh38
rs751051376 Chr.2: 127051221-127051221 on Build GRCh38
rs144459969 Chr.2: 127050894-127050894 on Build GRCh38
rs769199881 Chr.2: 127051212-127051212 on Build GRCh38
rs139598933 Chr.2: 127051108-127051108 on Build GRCh38
rs761168323 Chr.2: 127051128-127051128 on Build GRCh38
rs778989524 Chr.2: 127050766-127050766 on Build GRCh38
rs974796988 Chr.2: 127050733-127050733 on Build GRCh38
rs772155930 Chr.2: 127050850-127050850 on Build GRCh38
rs372302632 Chr.2: 127050928-127050928 on Build GRCh38
rs776178572 Chr.2: 127051207-127051207 on Build GRCh38

View Full Product Details