Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101846634-101847182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf76
Entrez Gene ID
79591
Gene Name
chromosome 10 open reading frame 76
Gene Aliases
-
UniGene
Hs.16004
Gene Chromosome Location
Chr.10: 101845599-102056237 on Build GRCh38
Transcripts
NM_024541.2 XM_006717969.1 XM_011540151.2 XM_011540152.2 XM_011540153.2 XM_011540154.2 XM_011540156.2 XM_011540157.2

SNP Detail

SNP ID Chromosome Location
rs964649333 Chr.10: 101846875-101846875 on Build GRCh38
rs1001965702 Chr.10: 101846816-101846816 on Build GRCh38
rs896769813 Chr.10: 101847077-101847077 on Build GRCh38
rs558732208 Chr.10: 101846743-101846743 on Build GRCh38
rs545980820 Chr.10: 101847008-101847008 on Build GRCh38
rs763783820 Chr.10: 101846746-101846746 on Build GRCh38
rs184595314 Chr.10: 101847128-101847128 on Build GRCh38
rs556107330 Chr.10: 101846818-101846818 on Build GRCh38
rs1037054663 Chr.10: 101847136-101847136 on Build GRCh38
rs534119742 Chr.10: 101846687-101846687 on Build GRCh38
rs755571874 Chr.10: 101846939-101846939 on Build GRCh38
rs904834072 Chr.10: 101846798-101846798 on Build GRCh38
rs574728173 Chr.10: 101846842-101846842 on Build GRCh38
rs192703785 Chr.10: 101847033-101847033 on Build GRCh38
rs755822797 Chr.10: 101846700-101846700 on Build GRCh38
rs187169746 Chr.10: 101846681-101846681 on Build GRCh38
rs11817154 Chr.10: 101846979-101846979 on Build GRCh38
rs914281109 Chr.10: 101846940-101846940 on Build GRCh38
rs912500775 Chr.10: 101846898-101846898 on Build GRCh38
rs926770347 Chr.10: 101846989-101846989 on Build GRCh38
rs937620153 Chr.10: 101847025-101847025 on Build GRCh38
rs1016391407 Chr.10: 101846817-101846817 on Build GRCh38
rs987664388 Chr.10: 101846860-101846860 on Build GRCh38
rs995725331 Chr.10: 101846854-101846854 on Build GRCh38
rs1026922186 Chr.10: 101847125-101847125 on Build GRCh38
rs896486606 Chr.10: 101846822-101846822 on Build GRCh38
rs115859246 Chr.10: 101846909-101846909 on Build GRCh38
rs749723391 Chr.10: 101846868-101846868 on Build GRCh38
rs1056767473 Chr.10: 101846812-101846812 on Build GRCh38
rs1004241026 Chr.10: 101846831-101846831 on Build GRCh38
rs986673847 Chr.10: 101846891-101846891 on Build GRCh38
rs556471830 Chr.10: 101846699-101846699 on Build GRCh38
rs938199095 Chr.10: 101847005-101847005 on Build GRCh38
rs11191129 Chr.10: 101846786-101846786 on Build GRCh38
rs542238898 Chr.10: 101846852-101846852 on Build GRCh38
rs550083857 Chr.10: 101847146-101847146 on Build GRCh38
rs1004594254 Chr.10: 101847118-101847118 on Build GRCh38
rs988507451 Chr.10: 101846918-101846918 on Build GRCh38
rs777930619 Chr.10: 101847060-101847060 on Build GRCh38
rs1026474645 Chr.10: 101846853-101846853 on Build GRCh38
rs898536373 Chr.10: 101847158-101847158 on Build GRCh38
rs748775720 Chr.10: 101846986-101846986 on Build GRCh38
rs905403513 Chr.10: 101846766-101846766 on Build GRCh38
rs947159647 Chr.10: 101846958-101846958 on Build GRCh38
rs577072961 Chr.10: 101846769-101846769 on Build GRCh38
rs988410067 Chr.10: 101846910-101846910 on Build GRCh38
rs376839511 Chr.10: 101846892-101846892 on Build GRCh38
rs547153706 Chr.10: 101846670-101846670 on Build GRCh38
rs779235576 Chr.10: 101846955-101846955 on Build GRCh38
rs946844372 Chr.10: 101846733-101846733 on Build GRCh38
rs780801352 Chr.10: 101846863-101846863 on Build GRCh38
rs374532540 Chr.10: 101846726-101846726 on Build GRCh38
rs949271953 Chr.10: 101846770-101846770 on Build GRCh38

View Full Product Details