Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58131053-58131600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf37
Entrez Gene ID
145407
Gene Name
chromosome 14 open reading frame 37
Gene Aliases
c14_5376
UniGene
Hs.26766
Gene Chromosome Location
Chr.14: 57993536-58152305 on Build GRCh38
Transcripts
NM_001001872.3 NM_001320173.1 XM_011536471.2 XM_017021029.1 XM_017021030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198085 c.1781C>A p.T594N Chr.14: 58131562-58131562 on Build GRCh38
7285506 c.1810C>A p.L604M Chr.14: 58131533-58131533 on Build GRCh38
5946820 c.1804T>C p.Y602H Chr.14: 58131539-58131539 on Build GRCh38
5067937 c.1813G>A p.D605N Chr.14: 58131530-58131530 on Build GRCh38
1707385 c.1760G>A p.R587K Chr.14: 58131583-58131583 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs942309087 Chr.14: 58131395-58131395 on Build GRCh38
rs1013239508 Chr.14: 58131465-58131468 on Build GRCh38
rs758405561 Chr.14: 58131568-58131568 on Build GRCh38
rs754114106 Chr.14: 58131559-58131559 on Build GRCh38
rs994080236 Chr.14: 58131446-58131446 on Build GRCh38
rs764474782 Chr.14: 58131550-58131550 on Build GRCh38
rs760961554 Chr.14: 58131547-58131547 on Build GRCh38
rs886982478 Chr.14: 58131343-58131343 on Build GRCh38
rs748906388 Chr.14: 58131547-58131548 on Build GRCh38
rs567934232 Chr.14: 58131546-58131546 on Build GRCh38
rs45483691 Chr.14: 58131534-58131534 on Build GRCh38
rs533272417 Chr.14: 58131428-58131428 on Build GRCh38
rs1055532769 Chr.14: 58131234-58131234 on Build GRCh38
rs901158858 Chr.14: 58131418-58131418 on Build GRCh38
rs374832009 Chr.14: 58131490-58131490 on Build GRCh38
rs79566991 Chr.14: 58131300-58131300 on Build GRCh38
rs538125128 Chr.14: 58131564-58131564 on Build GRCh38
rs375929300 Chr.14: 58131562-58131562 on Build GRCh38
rs1023331129 Chr.14: 58131485-58131485 on Build GRCh38
rs1023550291 Chr.14: 58131178-58131178 on Build GRCh38
rs542984643 Chr.14: 58131188-58131188 on Build GRCh38
rs778192524 Chr.14: 58131403-58131403 on Build GRCh38
rs945636433 Chr.14: 58131109-58131109 on Build GRCh38
rs1028357845 Chr.14: 58131433-58131433 on Build GRCh38
rs577686406 Chr.14: 58131099-58131099 on Build GRCh38
rs910789182 Chr.14: 58131384-58131384 on Build GRCh38
rs543586129 Chr.14: 58131379-58131379 on Build GRCh38
rs969324595 Chr.14: 58131184-58131184 on Build GRCh38
rs553805623 Chr.14: 58131136-58131136 on Build GRCh38
rs745827775 Chr.14: 58131480-58131480 on Build GRCh38
rs201610059 Chr.14: 58131541-58131541 on Build GRCh38
rs1013709030 Chr.14: 58131165-58131165 on Build GRCh38
rs750290334 Chr.14: 58131566-58131566 on Build GRCh38
rs779647797 Chr.14: 58131502-58131502 on Build GRCh38
rs551411961 Chr.14: 58131505-58131505 on Build GRCh38
rs527666943 Chr.14: 58131538-58131538 on Build GRCh38
rs1025828710 Chr.14: 58131100-58131100 on Build GRCh38
rs779413486 Chr.14: 58131577-58131577 on Build GRCh38
rs776771661 Chr.14: 58131395-58131395 on Build GRCh38
rs141103194 Chr.14: 58131427-58131427 on Build GRCh38
rs925085399 Chr.14: 58131226-58131226 on Build GRCh38
rs1046809399 Chr.14: 58131454-58131454 on Build GRCh38
rs763375353 Chr.14: 58131510-58131510 on Build GRCh38
rs115309130 Chr.14: 58131329-58131329 on Build GRCh38
rs993907953 Chr.14: 58131094-58131094 on Build GRCh38
rs200455245 Chr.14: 58131497-58131497 on Build GRCh38
rs1020003310 Chr.14: 58131392-58131392 on Build GRCh38
rs896259079 Chr.14: 58131462-58131462 on Build GRCh38
rs771333577 Chr.14: 58131522-58131522 on Build GRCh38
rs778646906 Chr.14: 58131467-58131467 on Build GRCh38
rs956761822 Chr.14: 58131298-58131298 on Build GRCh38
rs894267649 Chr.14: 58131241-58131241 on Build GRCh38
rs938376599 Chr.14: 58131202-58131202 on Build GRCh38
rs371627713 Chr.14: 58131560-58131560 on Build GRCh38
rs375093886 Chr.14: 58131575-58131575 on Build GRCh38
rs369498926 Chr.14: 58131531-58131531 on Build GRCh38
rs1041752909 Chr.14: 58131152-58131152 on Build GRCh38
rs190285263 Chr.14: 58131186-58131186 on Build GRCh38
rs185334639 Chr.14: 58131101-58131101 on Build GRCh38
rs373371374 Chr.14: 58131507-58131507 on Build GRCh38

View Full Product Details