Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15587398-15587948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf45
Entrez Gene ID
89927
Gene Name
chromosome 16 open reading frame 45
Gene Aliases
MINP
UniGene
Hs.401798
Gene Chromosome Location
Chr.16: 15434468-15594521 on Build GRCh38
Transcripts
NM_001142469.1 NM_033201.2 XM_011522731.2 XM_017023824.1

SNP Detail

SNP ID Chromosome Location
rs939323048 Chr.16: 15587711-15587711 on Build GRCh38
rs1005352565 Chr.16: 15587497-15587497 on Build GRCh38
rs140697506 Chr.16: 15587862-15587863 on Build GRCh38
rs996545863 Chr.16: 15587553-15587553 on Build GRCh38
rs781459498 Chr.16: 15587614-15587614 on Build GRCh38
rs895050695 Chr.16: 15587803-15587803 on Build GRCh38
rs908426918 Chr.16: 15587737-15587737 on Build GRCh38
rs1016702835 Chr.16: 15587531-15587531 on Build GRCh38
rs897013565 Chr.16: 15587780-15587780 on Build GRCh38
rs1048364942 Chr.16: 15587799-15587799 on Build GRCh38
rs547629634 Chr.16: 15587916-15587916 on Build GRCh38
rs906494392 Chr.16: 15587861-15587861 on Build GRCh38
rs367641132 Chr.16: 15587520-15587520 on Build GRCh38
rs549468767 Chr.16: 15587703-15587703 on Build GRCh38
rs1047017932 Chr.16: 15587716-15587716 on Build GRCh38
rs537104423 Chr.16: 15587418-15587418 on Build GRCh38
rs886939719 Chr.16: 15587479-15587479 on Build GRCh38
rs912205950 Chr.16: 15587731-15587731 on Build GRCh38
rs990180439 Chr.16: 15587694-15587694 on Build GRCh38
rs182488871 Chr.16: 15587433-15587433 on Build GRCh38
rs998497221 Chr.16: 15587857-15587857 on Build GRCh38
rs116165738 Chr.16: 15587678-15587678 on Build GRCh38
rs1127272 Chr.16: 15587561-15587561 on Build GRCh38
rs61060358 Chr.16: 15587836-15587836 on Build GRCh38
rs755125829 Chr.16: 15587545-15587545 on Build GRCh38
rs770226162 Chr.16: 15587805-15587806 on Build GRCh38
rs562421597 Chr.16: 15587569-15587569 on Build GRCh38
rs922171980 Chr.16: 15587698-15587698 on Build GRCh38
rs11540893 Chr.16: 15587677-15587677 on Build GRCh38
rs780229670 Chr.16: 15587508-15587508 on Build GRCh38
rs941190305 Chr.16: 15587746-15587746 on Build GRCh38
rs1000024287 Chr.16: 15587867-15587867 on Build GRCh38
rs989579478 Chr.16: 15587653-15587653 on Build GRCh38
rs980824660 Chr.16: 15587691-15587691 on Build GRCh38
rs185880129 Chr.16: 15587655-15587655 on Build GRCh38
rs748447728 Chr.16: 15587761-15587761 on Build GRCh38
rs749624029 Chr.16: 15587638-15587638 on Build GRCh38
rs567321109 Chr.16: 15587748-15587748 on Build GRCh38
rs149272228 Chr.16: 15587742-15587742 on Build GRCh38
rs1007949177 Chr.16: 15587438-15587438 on Build GRCh38
rs932319269 Chr.16: 15587420-15587420 on Build GRCh38
rs544832960 Chr.16: 15587630-15587630 on Build GRCh38
rs577564855 Chr.16: 15587607-15587607 on Build GRCh38
rs959398631 Chr.16: 15587548-15587548 on Build GRCh38
rs758973857 Chr.16: 15587484-15587484 on Build GRCh38
rs756607603 Chr.16: 15587535-15587535 on Build GRCh38
rs182517019 Chr.16: 15587910-15587910 on Build GRCh38
rs756889950 Chr.16: 15587460-15587460 on Build GRCh38
rs746022812 Chr.16: 15587835-15587835 on Build GRCh38
rs1049399797 Chr.16: 15587724-15587724 on Build GRCh38
rs956929854 Chr.16: 15587870-15587870 on Build GRCh38
rs11540891 Chr.16: 15587754-15587754 on Build GRCh38
rs555036670 Chr.16: 15587419-15587419 on Build GRCh38
rs939252343 Chr.16: 15587438-15587438 on Build GRCh38
rs773374490 Chr.16: 15587647-15587647 on Build GRCh38
rs148101467 Chr.16: 15587825-15587826 on Build GRCh38
rs772347409 Chr.16: 15587875-15587875 on Build GRCh38
rs750000195 Chr.16: 15587424-15587424 on Build GRCh38
rs191078562 Chr.16: 15587757-15587757 on Build GRCh38
rs932350341 Chr.16: 15587723-15587723 on Build GRCh38
rs988123787 Chr.16: 15587637-15587639 on Build GRCh38
rs550244675 Chr.16: 15587791-15587791 on Build GRCh38
rs746532897 Chr.16: 15587478-15587478 on Build GRCh38
rs758088450 Chr.16: 15587432-15587432 on Build GRCh38
rs543842384 Chr.16: 15587468-15587468 on Build GRCh38
rs747409965 Chr.16: 15587512-15587512 on Build GRCh38
rs750936009 Chr.16: 15587476-15587476 on Build GRCh38
rs927947591 Chr.16: 15587708-15587708 on Build GRCh38
rs1031224612 Chr.16: 15587861-15587862 on Build GRCh38
rs3182975 Chr.16: 15587549-15587549 on Build GRCh38
rs765821189 Chr.16: 15587465-15587465 on Build GRCh38
rs995837948 Chr.16: 15587536-15587536 on Build GRCh38
rs748211459 Chr.16: 15587627-15587627 on Build GRCh38

View Full Product Details