Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13889069-13889623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf57
Entrez Gene ID
79173
Gene Name
chromosome 19 open reading frame 57
Gene Aliases
-
UniGene
Hs.143288
Gene Chromosome Location
Chr.19: 13882348-13906125 on Build GRCh38
Transcripts
NM_024323.3 XM_006722885.3 XM_011528286.2 XM_011528287.2 XM_017027300.1 XM_017027301.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4074709 c.1535A>G p.Q512R Chr.19: 13889321-13889321 on Build GRCh38
4074708 c.1634T>C p.L545P Chr.19: 13889222-13889222 on Build GRCh38
3281281 c.1609C>T p.L537L Chr.19: 13889247-13889247 on Build GRCh38
4074710 c.1431G>A p.P477P Chr.19: 13889425-13889425 on Build GRCh38
4000527 c.1499A>G p.Q500R Chr.19: 13889357-13889357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745870796 Chr.19: 13889496-13889496 on Build GRCh38
rs372491962 Chr.19: 13889187-13889187 on Build GRCh38
rs371139551 Chr.19: 13889181-13889181 on Build GRCh38
rs779598821 Chr.19: 13889312-13889312 on Build GRCh38
rs753230986 Chr.19: 13889214-13889214 on Build GRCh38
rs566278344 Chr.19: 13889205-13889205 on Build GRCh38
rs773214574 Chr.19: 13889259-13889259 on Build GRCh38
rs573407614 Chr.19: 13889316-13889316 on Build GRCh38
rs762118199 Chr.19: 13889450-13889450 on Build GRCh38
rs200553783 Chr.19: 13889468-13889468 on Build GRCh38
rs747839167 Chr.19: 13889334-13889334 on Build GRCh38
rs781023704 Chr.19: 13889479-13889479 on Build GRCh38
rs557641925 Chr.19: 13889223-13889223 on Build GRCh38
rs200031533 Chr.19: 13889212-13889212 on Build GRCh38
rs748425839 Chr.19: 13889591-13889591 on Build GRCh38
rs148291889 Chr.19: 13889307-13889307 on Build GRCh38
rs767965036 Chr.19: 13889453-13889453 on Build GRCh38
rs773205969 Chr.19: 13889198-13889198 on Build GRCh38
rs745772565 Chr.19: 13889537-13889537 on Build GRCh38
rs919367198 Chr.19: 13889166-13889166 on Build GRCh38
rs776976989 Chr.19: 13889288-13889288 on Build GRCh38
rs144452246 Chr.19: 13889435-13889435 on Build GRCh38
rs768428892 Chr.19: 13889231-13889231 on Build GRCh38
rs61733835 Chr.19: 13889358-13889358 on Build GRCh38
rs750366905 Chr.19: 13889570-13889570 on Build GRCh38
rs1031464238 Chr.19: 13889248-13889248 on Build GRCh38
rs78688381 Chr.19: 13889531-13889531 on Build GRCh38
rs569609896 Chr.19: 13889550-13889550 on Build GRCh38
rs143252964 Chr.19: 13889258-13889258 on Build GRCh38
rs761798434 Chr.19: 13889183-13889183 on Build GRCh38
rs376128927 Chr.19: 13889144-13889144 on Build GRCh38
rs532762540 Chr.19: 13889413-13889413 on Build GRCh38
rs757116463 Chr.19: 13889551-13889551 on Build GRCh38
rs759884121 Chr.19: 13889291-13889291 on Build GRCh38
rs923323435 Chr.19: 13889365-13889365 on Build GRCh38
rs778568972 Chr.19: 13889329-13889329 on Build GRCh38
rs751316867 Chr.19: 13889481-13889481 on Build GRCh38
rs1012529853 Chr.19: 13889407-13889407 on Build GRCh38
rs780233198 Chr.19: 13889502-13889502 on Build GRCh38
rs867271629 Chr.19: 13889588-13889588 on Build GRCh38
rs535747826 Chr.19: 13889218-13889218 on Build GRCh38
rs549086139 Chr.19: 13889454-13889454 on Build GRCh38
rs915365490 Chr.19: 13889518-13889518 on Build GRCh38
rs779887323 Chr.19: 13889583-13889583 on Build GRCh38
rs759585296 Chr.19: 13889499-13889499 on Build GRCh38
rs754666979 Chr.19: 13889318-13889318 on Build GRCh38
rs914436599 Chr.19: 13889323-13889323 on Build GRCh38
rs767482509 Chr.19: 13889559-13889559 on Build GRCh38
rs760603876 Chr.19: 13889260-13889260 on Build GRCh38
rs770000652 Chr.19: 13889436-13889436 on Build GRCh38
rs781297249 Chr.19: 13889431-13889431 on Build GRCh38
rs769178284 Chr.19: 13889300-13889300 on Build GRCh38
rs764590026 Chr.19: 13889373-13889373 on Build GRCh38
rs117615888 Chr.19: 13889211-13889211 on Build GRCh38
rs770089296 Chr.19: 13889367-13889367 on Build GRCh38
rs1054341527 Chr.19: 13889098-13889098 on Build GRCh38
rs781211818 Chr.19: 13889491-13889491 on Build GRCh38
rs1016558993 Chr.19: 13889170-13889170 on Build GRCh38
rs749862822 Chr.19: 13889230-13889230 on Build GRCh38
rs765661954 Chr.19: 13889293-13889293 on Build GRCh38
rs766546821 Chr.19: 13889287-13889287 on Build GRCh38
rs775234588 Chr.19: 13889152-13889152 on Build GRCh38
rs575661394 Chr.19: 13889294-13889294 on Build GRCh38
rs768883820 Chr.19: 13889513-13889513 on Build GRCh38
rs568528452 Chr.19: 13889528-13889528 on Build GRCh38
rs539885338 Chr.19: 13889295-13889295 on Build GRCh38
rs774183720 Chr.19: 13889232-13889232 on Build GRCh38
rs765845203 Chr.19: 13889387-13889387 on Build GRCh38
rs886598321 Chr.19: 13889382-13889382 on Build GRCh38
rs749441204 Chr.19: 13889503-13889503 on Build GRCh38
rs973381881 Chr.19: 13889165-13889165 on Build GRCh38
rs753679280 Chr.19: 13889493-13889493 on Build GRCh38
rs551002745 Chr.19: 13889425-13889425 on Build GRCh38
rs149161110 Chr.19: 13889251-13889251 on Build GRCh38
rs752461358 Chr.19: 13889412-13889412 on Build GRCh38
rs199786719 Chr.19: 13889159-13889159 on Build GRCh38
rs200969478 Chr.19: 13889206-13889206 on Build GRCh38
rs943332549 Chr.19: 13889114-13889114 on Build GRCh38
rs1037875772 Chr.19: 13889386-13889386 on Build GRCh38
rs201514767 Chr.19: 13889157-13889157 on Build GRCh38
rs3803891 Chr.19: 13889587-13889587 on Build GRCh38
rs771903764 Chr.19: 13889344-13889344 on Build GRCh38
rs754115879 Chr.19: 13889210-13889210 on Build GRCh38
rs758864820 Chr.19: 13889215-13889215 on Build GRCh38
rs138545136 Chr.19: 13889396-13889396 on Build GRCh38
rs778468764 Chr.19: 13889397-13889397 on Build GRCh38
rs763163182 Chr.19: 13889371-13889371 on Build GRCh38
rs774819622 Chr.19: 13889448-13889448 on Build GRCh38
rs147705428 Chr.19: 13889217-13889217 on Build GRCh38
rs372100801 Chr.19: 13889581-13889581 on Build GRCh38
rs149264968 Chr.19: 13889426-13889426 on Build GRCh38
rs1006474996 Chr.19: 13889158-13889158 on Build GRCh38
rs779617536 Chr.19: 13889224-13889224 on Build GRCh38
rs73922796 Chr.19: 13889537-13889537 on Build GRCh38
rs770089859 Chr.19: 13889113-13889113 on Build GRCh38
rs777317871 Chr.19: 13889479-13889479 on Build GRCh38
rs773018050 Chr.19: 13889350-13889350 on Build GRCh38
rs769295047 Chr.19: 13889145-13889145 on Build GRCh38
rs145301854 Chr.19: 13889469-13889469 on Build GRCh38
rs200491827 Chr.19: 13889393-13889393 on Build GRCh38
rs573794640 Chr.19: 13889363-13889363 on Build GRCh38
rs3803892 Chr.19: 13889357-13889357 on Build GRCh38
rs140619176 Chr.19: 13889308-13889308 on Build GRCh38
rs749341329 Chr.19: 13889586-13889586 on Build GRCh38
rs148392972 Chr.19: 13889460-13889460 on Build GRCh38
rs777610998 Chr.19: 13889211-13889212 on Build GRCh38
rs747023999 Chr.19: 13889352-13889352 on Build GRCh38
rs763741668 Chr.19: 13889546-13889546 on Build GRCh38
rs368580390 Chr.19: 13889446-13889446 on Build GRCh38
rs934561496 Chr.19: 13889093-13889093 on Build GRCh38
rs779190129 Chr.19: 13889589-13889589 on Build GRCh38
rs748456410 Chr.19: 13889516-13889516 on Build GRCh38
rs367919307 Chr.19: 13889543-13889543 on Build GRCh38
rs527426427 Chr.19: 13889445-13889445 on Build GRCh38
rs759040859 Chr.19: 13889457-13889457 on Build GRCh38
rs61732721 Chr.19: 13889383-13889383 on Build GRCh38
rs1047910489 Chr.19: 13889154-13889154 on Build GRCh38
rs751325635 Chr.19: 13889270-13889271 on Build GRCh38
rs369147070 Chr.19: 13889140-13889140 on Build GRCh38
rs765173677 Chr.19: 13889580-13889580 on Build GRCh38
rs147206426 Chr.19: 13889490-13889490 on Build GRCh38
rs369132888 Chr.19: 13889240-13889240 on Build GRCh38
rs1047880608 Chr.19: 13889370-13889370 on Build GRCh38
rs777669384 Chr.19: 13889424-13889424 on Build GRCh38
rs1039039669 Chr.19: 13889136-13889136 on Build GRCh38
rs761219639 Chr.19: 13889536-13889536 on Build GRCh38
rs762389939 Chr.19: 13889545-13889545 on Build GRCh38
rs140606837 Chr.19: 13889514-13889514 on Build GRCh38
rs757010360 Chr.19: 13889332-13889333 on Build GRCh38
rs143008219 Chr.19: 13889368-13889368 on Build GRCh38

View Full Product Details