Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 119296348-119296918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf3
Entrez Gene ID
401152
Gene Name
chromosome 4 open reading frame 3
Gene Aliases
-
UniGene
Hs.173705
Gene Chromosome Location
Chr.4: 119296419-119304445 on Build GRCh38
Transcripts
NM_001001701.3 NM_001170330.1

SNP Detail

SNP ID Chromosome Location
rs925557816 Chr.4: 119296688-119296688 on Build GRCh38
rs1001821173 Chr.4: 119296565-119296565 on Build GRCh38
rs993783998 Chr.4: 119296494-119296494 on Build GRCh38
rs563431739 Chr.4: 119296761-119296761 on Build GRCh38
rs1013712178 Chr.4: 119296843-119296843 on Build GRCh38
rs944134258 Chr.4: 119296379-119296379 on Build GRCh38
rs935860502 Chr.4: 119296896-119296896 on Build GRCh38
rs537667873 Chr.4: 119296405-119296405 on Build GRCh38
rs1040362302 Chr.4: 119296695-119296695 on Build GRCh38
rs1040583628 Chr.4: 119296388-119296388 on Build GRCh38
rs780824499 Chr.4: 119296581-119296581 on Build GRCh38
rs927887770 Chr.4: 119296528-119296543 on Build GRCh38
rs370774982 Chr.4: 119296847-119296847 on Build GRCh38
rs1044642806 Chr.4: 119296504-119296504 on Build GRCh38
rs77923072 Chr.4: 119296613-119296613 on Build GRCh38
rs916902284 Chr.4: 119296844-119296844 on Build GRCh38
rs1006430117 Chr.4: 119296731-119296731 on Build GRCh38
rs375907055 Chr.4: 119296759-119296759 on Build GRCh38
rs762853150 Chr.4: 119296804-119296804 on Build GRCh38
rs552195990 Chr.4: 119296838-119296838 on Build GRCh38
rs771433756 Chr.4: 119296433-119296433 on Build GRCh38
rs909558972 Chr.4: 119296425-119296425 on Build GRCh38
rs565642855 Chr.4: 119296857-119296857 on Build GRCh38
rs891716313 Chr.4: 119296588-119296588 on Build GRCh38
rs927115797 Chr.4: 119296876-119296876 on Build GRCh38
rs960989235 Chr.4: 119296440-119296440 on Build GRCh38
rs369734964 Chr.4: 119296884-119296884 on Build GRCh38
rs561875082 Chr.4: 119296716-119296716 on Build GRCh38
rs1010689414 Chr.4: 119296651-119296651 on Build GRCh38
rs1054335990 Chr.4: 119296560-119296560 on Build GRCh38
rs901947811 Chr.4: 119296708-119296708 on Build GRCh38
rs954302689 Chr.4: 119296707-119296707 on Build GRCh38
rs917018079 Chr.4: 119296484-119296484 on Build GRCh38
rs979845984 Chr.4: 119296873-119296873 on Build GRCh38
rs537997112 Chr.4: 119296757-119296757 on Build GRCh38
rs768350335 Chr.4: 119296444-119296444 on Build GRCh38
rs972424064 Chr.4: 119296458-119296458 on Build GRCh38
rs966913346 Chr.4: 119296628-119296628 on Build GRCh38
rs1033329386 Chr.4: 119296571-119296571 on Build GRCh38
rs919050424 Chr.4: 119296753-119296753 on Build GRCh38
rs184385954 Chr.4: 119296518-119296518 on Build GRCh38
rs76922238 Chr.4: 119296712-119296712 on Build GRCh38
rs141609811 Chr.4: 119296784-119296784 on Build GRCh38
rs980044612 Chr.4: 119296660-119296660 on Build GRCh38
rs952178733 Chr.4: 119296721-119296721 on Build GRCh38
rs116599747 Chr.4: 119296711-119296711 on Build GRCh38
rs887725563 Chr.4: 119296561-119296561 on Build GRCh38
rs555638057 Chr.4: 119296740-119296740 on Build GRCh38
rs557364328 Chr.4: 119296488-119296488 on Build GRCh38
rs77377547 Chr.4: 119296556-119296556 on Build GRCh38
rs763933472 Chr.4: 119296805-119296805 on Build GRCh38
rs1013298563 Chr.4: 119296608-119296608 on Build GRCh38

View Full Product Details