Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120953277-120953822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5
Entrez Gene ID
727
Gene Name
complement component 5
Gene Aliases
C5D, C5a, C5b, CPAMD4, ECLZB
UniGene
Hs.494997
Gene Chromosome Location
Chr.9: 120952335-121075174 on Build GRCh38
Transcripts
NM_001317163.1 NM_001735.2 XM_011518980.2 XM_017015102.1 XM_017015103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616196 c.4858A>G p.M1620V Chr.9: 120953773-120953773 on Build GRCh38
1104704 c.4879A>G p.I1627V Chr.9: 120953752-120953752 on Build GRCh38
6208333 c.4824C>A p.N1608K Chr.9: 120953807-120953807 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1011827080 Chr.9: 120953657-120953657 on Build GRCh38
rs41313637 Chr.9: 120953673-120953673 on Build GRCh38
rs772799880 Chr.9: 120953715-120953716 on Build GRCh38
rs1034723789 Chr.9: 120953349-120953349 on Build GRCh38
rs564009235 Chr.9: 120953623-120953623 on Build GRCh38
rs909958572 Chr.9: 120953476-120953476 on Build GRCh38
rs956133496 Chr.9: 120953337-120953337 on Build GRCh38
rs748416336 Chr.9: 120953707-120953707 on Build GRCh38
rs376359892 Chr.9: 120953711-120953711 on Build GRCh38
rs529745339 Chr.9: 120953495-120953495 on Build GRCh38
rs559464915 Chr.9: 120953446-120953446 on Build GRCh38
rs558475548 Chr.9: 120953345-120953345 on Build GRCh38
rs920821420 Chr.9: 120953516-120953516 on Build GRCh38
rs943390228 Chr.9: 120953437-120953437 on Build GRCh38
rs543650571 Chr.9: 120953336-120953336 on Build GRCh38
rs761806894 Chr.9: 120953663-120953663 on Build GRCh38
rs886629911 Chr.9: 120953593-120953593 on Build GRCh38
rs961466723 Chr.9: 120953554-120953554 on Build GRCh38
rs772173761 Chr.9: 120953545-120953545 on Build GRCh38
rs777352661 Chr.9: 120953680-120953680 on Build GRCh38
rs540879797 Chr.9: 120953405-120953405 on Build GRCh38
rs774236417 Chr.9: 120953729-120953729 on Build GRCh38
rs1056174897 Chr.9: 120953640-120953640 on Build GRCh38
rs373869052 Chr.9: 120953694-120953694 on Build GRCh38
rs367883502 Chr.9: 120953696-120953696 on Build GRCh38
rs972703207 Chr.9: 120953566-120953566 on Build GRCh38
rs371658863 Chr.9: 120953700-120953700 on Build GRCh38
rs753463804 Chr.9: 120953684-120953684 on Build GRCh38
rs149147126 Chr.9: 120953732-120953732 on Build GRCh38
rs767316313 Chr.9: 120953754-120953754 on Build GRCh38
rs760651497 Chr.9: 120953794-120953794 on Build GRCh38
rs934209869 Chr.9: 120953642-120953642 on Build GRCh38
rs531706595 Chr.9: 120953617-120953617 on Build GRCh38
rs955421511 Chr.9: 120953703-120953703 on Build GRCh38
rs576240486 Chr.9: 120953304-120953304 on Build GRCh38
rs576929119 Chr.9: 120953381-120953381 on Build GRCh38
rs560786653 Chr.9: 120953471-120953471 on Build GRCh38
rs775519377 Chr.9: 120953755-120953755 on Build GRCh38
rs149090883 Chr.9: 120953717-120953717 on Build GRCh38
rs770261017 Chr.9: 120953710-120953710 on Build GRCh38
rs760419832 Chr.9: 120953791-120953792 on Build GRCh38
rs781739988 Chr.9: 120953706-120953706 on Build GRCh38
rs894809550 Chr.9: 120953646-120953646 on Build GRCh38
rs771338438 Chr.9: 120953720-120953720 on Build GRCh38
rs987583253 Chr.9: 120953412-120953412 on Build GRCh38
rs373402882 Chr.9: 120953324-120953324 on Build GRCh38
rs759733181 Chr.9: 120953460-120953460 on Build GRCh38
rs977924415 Chr.9: 120953308-120953308 on Build GRCh38
rs960606306 Chr.9: 120953380-120953380 on Build GRCh38
rs41313639 Chr.9: 120953426-120953426 on Build GRCh38
rs1031272974 Chr.9: 120953319-120953319 on Build GRCh38
rs984584882 Chr.9: 120953472-120953472 on Build GRCh38
rs776493316 Chr.9: 120953570-120953570 on Build GRCh38
rs387906554 Chr.9: 120953758-120953759 on Build GRCh38
rs552786127 Chr.9: 120953658-120953658 on Build GRCh38
rs748980959 Chr.9: 120953650-120953650 on Build GRCh38
rs41258384 Chr.9: 120953628-120953628 on Build GRCh38
rs768980521 Chr.9: 120953704-120953704 on Build GRCh38
rs541833503 Chr.9: 120953513-120953513 on Build GRCh38

View Full Product Details