Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137250199-137250777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf173-AS1
Entrez Gene ID
100129722
Gene Name
C9orf173 antisense RNA 1
Gene Aliases
-
UniGene
Hs.656347
Gene Chromosome Location
Chr.9: 137250219-137253497 on Build GRCh38
Transcripts
NR_038389.1 XM_006717108.2 XM_011518694.1 XM_011518695.1 XM_011518696.2 XM_011518697.1 XM_017014718.1

Gene Symbol
FAM166A
Entrez Gene ID
401565
Gene Name
family with sequence similarity 166 member A
Gene Aliases
HSD46
UniGene
Hs.522530
Gene Chromosome Location
Chr.9: 137243583-137251193 on Build GRCh38
Transcripts
NR_038389.1 XM_006717108.2 XM_011518694.1 XM_011518695.1 XM_011518696.2 XM_011518697.1 XM_017014718.1

SNP Detail

SNP ID Chromosome Location
rs556206640 Chr.9: 137250534-137250534 on Build GRCh38
rs73668203 Chr.9: 137250386-137250386 on Build GRCh38
rs950349828 Chr.9: 137250527-137250527 on Build GRCh38
rs950059370 Chr.9: 137250749-137250749 on Build GRCh38
rs1039768827 Chr.9: 137250234-137250234 on Build GRCh38
rs553295059 Chr.9: 137250247-137250247 on Build GRCh38
rs999077829 Chr.9: 137250564-137250564 on Build GRCh38
rs903464853 Chr.9: 137250295-137250295 on Build GRCh38
rs111972181 Chr.9: 137250626-137250626 on Build GRCh38
rs3195220 Chr.9: 137250243-137250243 on Build GRCh38
rs1014866225 Chr.9: 137250381-137250381 on Build GRCh38
rs921111368 Chr.9: 137250747-137250747 on Build GRCh38
rs191017492 Chr.9: 137250275-137250275 on Build GRCh38
rs918442457 Chr.9: 137250460-137250460 on Build GRCh38
rs989195664 Chr.9: 137250356-137250356 on Build GRCh38
rs577906274 Chr.9: 137250390-137250390 on Build GRCh38
rs985121090 Chr.9: 137250417-137250417 on Build GRCh38
rs767514379 Chr.9: 137250745-137250745 on Build GRCh38
rs73565763 Chr.9: 137250380-137250380 on Build GRCh38
rs999528614 Chr.9: 137250313-137250313 on Build GRCh38
rs893302549 Chr.9: 137250594-137250594 on Build GRCh38
rs569723175 Chr.9: 137250664-137250664 on Build GRCh38
rs543414456 Chr.9: 137250317-137250317 on Build GRCh38
rs183323285 Chr.9: 137250489-137250489 on Build GRCh38
rs1055124368 Chr.9: 137250568-137250568 on Build GRCh38
rs962497831 Chr.9: 137250706-137250706 on Build GRCh38
rs934056260 Chr.9: 137250750-137250750 on Build GRCh38
rs572934371 Chr.9: 137250432-137250432 on Build GRCh38
rs556999620 Chr.9: 137250324-137250324 on Build GRCh38
rs998179080 Chr.9: 137250278-137250278 on Build GRCh38
rs1133438 Chr.9: 137250625-137250625 on Build GRCh38
rs921373767 Chr.9: 137250507-137250507 on Build GRCh38
rs910001170 Chr.9: 137250667-137250667 on Build GRCh38
rs770605203 Chr.9: 137250510-137250510 on Build GRCh38
rs930072336 Chr.9: 137250310-137250310 on Build GRCh38
rs1017952188 Chr.9: 137250622-137250622 on Build GRCh38
rs918694122 Chr.9: 137250301-137250301 on Build GRCh38
rs1023375399 Chr.9: 137250603-137250603 on Build GRCh38
rs1133439 Chr.9: 137250280-137250280 on Build GRCh38
rs560939564 Chr.9: 137250619-137250619 on Build GRCh38
rs957725719 Chr.9: 137250416-137250416 on Build GRCh38
rs1056490925 Chr.9: 137250375-137250375 on Build GRCh38
rs550524058 Chr.9: 137250615-137250615 on Build GRCh38
rs909680343 Chr.9: 137250462-137250462 on Build GRCh38
rs768867589 Chr.9: 137250616-137250616 on Build GRCh38
rs977043962 Chr.9: 137250458-137250458 on Build GRCh38
rs373198322 Chr.9: 137250640-137250641 on Build GRCh38
rs929769606 Chr.9: 137250463-137250463 on Build GRCh38
rs139650234 Chr.9: 137250249-137250249 on Build GRCh38
rs761999234 Chr.9: 137250731-137250731 on Build GRCh38
rs559626573 Chr.9: 137250391-137250391 on Build GRCh38
rs976592436 Chr.9: 137250287-137250287 on Build GRCh38
rs1025977600 Chr.9: 137250643-137250643 on Build GRCh38
rs116869306 Chr.9: 137250665-137250665 on Build GRCh38
rs937336903 Chr.9: 137250358-137250358 on Build GRCh38
rs957528837 Chr.9: 137250344-137250344 on Build GRCh38
rs544241325 Chr.9: 137250433-137250433 on Build GRCh38
rs909970177 Chr.9: 137250490-137250490 on Build GRCh38
rs951306200 Chr.9: 137250403-137250403 on Build GRCh38
rs937367907 Chr.9: 137250491-137250491 on Build GRCh38
rs1013131116 Chr.9: 137250601-137250601 on Build GRCh38
rs969096823 Chr.9: 137250609-137250609 on Build GRCh38
rs991960743 Chr.9: 137250680-137250680 on Build GRCh38
rs539250740 Chr.9: 137250270-137250270 on Build GRCh38
rs541219795 Chr.9: 137250240-137250240 on Build GRCh38
rs1051079220 Chr.9: 137250513-137250513 on Build GRCh38
rs117626986 Chr.9: 137250506-137250506 on Build GRCh38
rs975247167 Chr.9: 137250743-137250743 on Build GRCh38
rs79974358 Chr.9: 137250694-137250694 on Build GRCh38
rs745601171 Chr.9: 137250279-137250279 on Build GRCh38
rs1000357207 Chr.9: 137250582-137250582 on Build GRCh38
rs927631750 Chr.9: 137250401-137250401 on Build GRCh38
rs965391304 Chr.9: 137250623-137250623 on Build GRCh38
rs933981153 Chr.9: 137250510-137250510 on Build GRCh38
rs1042323092 Chr.9: 137250537-137250537 on Build GRCh38
rs911133245 Chr.9: 137250250-137250250 on Build GRCh38

View Full Product Details