Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114626676-114627224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM268
Entrez Gene ID
203197
Gene Name
transmembrane protein 268
Gene Aliases
C9orf91
UniGene
Hs.522357
Gene Chromosome Location
Chr.9: 114603897-114646423 on Build GRCh38
Transcripts
NM_153045.3 XM_006716997.1 XM_011518350.1 XM_011518351.2 XM_011518352.1 XM_011518353.1 XM_011518354.1 XM_011518355.1 XM_017014429.1 XM_017014430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4668221 c.245C>T p.A82V Chr.9: 114626927-114626927 on Build GRCh38
3903410 c.222G>A p.P74P Chr.9: 114626904-114626904 on Build GRCh38
3745789 c.296C>T p.S99L Chr.9: 114626978-114626978 on Build GRCh38
6617038 c.309G>A p.R103R Chr.9: 114626991-114626991 on Build GRCh38
1104425 c.277A>G p.R93G Chr.9: 114626959-114626959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556374931 Chr.9: 114626764-114626764 on Build GRCh38
rs528956798 Chr.9: 114627153-114627153 on Build GRCh38
rs574974608 Chr.9: 114626809-114626809 on Build GRCh38
rs747876890 Chr.9: 114626863-114626863 on Build GRCh38
rs769171646 Chr.9: 114627049-114627049 on Build GRCh38
rs949532788 Chr.9: 114627137-114627137 on Build GRCh38
rs949744707 Chr.9: 114626720-114626720 on Build GRCh38
rs764517200 Chr.9: 114626979-114626979 on Build GRCh38
rs201893905 Chr.9: 114626905-114626905 on Build GRCh38
rs565525215 Chr.9: 114627183-114627183 on Build GRCh38
rs747607201 Chr.9: 114626938-114626938 on Build GRCh38
rs752184747 Chr.9: 114627024-114627024 on Build GRCh38
rs996118158 Chr.9: 114626989-114626989 on Build GRCh38
rs549709965 Chr.9: 114627121-114627121 on Build GRCh38
rs750206413 Chr.9: 114626904-114626904 on Build GRCh38
rs577148052 Chr.9: 114626722-114626722 on Build GRCh38
rs900196844 Chr.9: 114627197-114627197 on Build GRCh38
rs762205864 Chr.9: 114627057-114627057 on Build GRCh38
rs569554280 Chr.9: 114627138-114627138 on Build GRCh38
rs767944193 Chr.9: 114626883-114626883 on Build GRCh38
rs765408245 Chr.9: 114626986-114626986 on Build GRCh38
rs754963234 Chr.9: 114626937-114626937 on Build GRCh38
rs1030228128 Chr.9: 114627201-114627201 on Build GRCh38
rs974565409 Chr.9: 114626796-114626796 on Build GRCh38
rs377294473 Chr.9: 114627005-114627005 on Build GRCh38
rs999909021 Chr.9: 114626775-114626775 on Build GRCh38
rs201950258 Chr.9: 114626901-114626901 on Build GRCh38
rs769358495 Chr.9: 114626948-114626948 on Build GRCh38
rs772531072 Chr.9: 114626964-114626964 on Build GRCh38
rs751334203 Chr.9: 114627012-114627012 on Build GRCh38
rs747315485 Chr.9: 114627040-114627040 on Build GRCh38
rs200051807 Chr.9: 114627026-114627026 on Build GRCh38
rs923137744 Chr.9: 114626808-114626808 on Build GRCh38
rs765442550 Chr.9: 114626903-114626903 on Build GRCh38
rs186322980 Chr.9: 114626752-114626752 on Build GRCh38
rs372811324 Chr.9: 114626978-114626978 on Build GRCh38
rs548685613 Chr.9: 114627180-114627180 on Build GRCh38
rs775816921 Chr.9: 114626976-114626976 on Build GRCh38
rs769423690 Chr.9: 114626864-114626864 on Build GRCh38
rs146386481 Chr.9: 114626963-114626963 on Build GRCh38
rs897863293 Chr.9: 114627171-114627171 on Build GRCh38
rs754983428 Chr.9: 114627013-114627013 on Build GRCh38
rs967201690 Chr.9: 114627058-114627058 on Build GRCh38
rs932397354 Chr.9: 114627131-114627131 on Build GRCh38
rs758536206 Chr.9: 114626991-114626991 on Build GRCh38
rs770482908 Chr.9: 114627035-114627035 on Build GRCh38
rs1055333861 Chr.9: 114627086-114627086 on Build GRCh38
rs1047445637 Chr.9: 114626965-114626965 on Build GRCh38
rs938248666 Chr.9: 114627074-114627074 on Build GRCh38
rs182231069 Chr.9: 114626702-114626702 on Build GRCh38
rs913986467 Chr.9: 114626836-114626836 on Build GRCh38
rs142772339 Chr.9: 114626985-114626985 on Build GRCh38
rs955423494 Chr.9: 114626812-114626812 on Build GRCh38
rs749024118 Chr.9: 114627028-114627028 on Build GRCh38
rs959060787 Chr.9: 114627190-114627190 on Build GRCh38
rs1030156092 Chr.9: 114627014-114627014 on Build GRCh38
rs541958711 Chr.9: 114626888-114626888 on Build GRCh38
rs777082214 Chr.9: 114627052-114627052 on Build GRCh38
rs925316936 Chr.9: 114627073-114627073 on Build GRCh38
rs964988686 Chr.9: 114626773-114626773 on Build GRCh38
rs532787740 Chr.9: 114627085-114627085 on Build GRCh38
rs374290052 Chr.9: 114626892-114626892 on Build GRCh38
rs1006422879 Chr.9: 114627159-114627159 on Build GRCh38
rs777042057 Chr.9: 114627069-114627069 on Build GRCh38
rs534537717 Chr.9: 114627186-114627186 on Build GRCh38
rs139126699 Chr.9: 114626987-114626987 on Build GRCh38
rs371081937 Chr.9: 114626861-114626861 on Build GRCh38
rs115599938 Chr.9: 114626770-114626770 on Build GRCh38
rs1015517621 Chr.9: 114627147-114627147 on Build GRCh38
rs918183353 Chr.9: 114627122-114627122 on Build GRCh38
rs760061669 Chr.9: 114627143-114627143 on Build GRCh38
rs772884544 Chr.9: 114626950-114626950 on Build GRCh38
rs370281778 Chr.9: 114626865-114626865 on Build GRCh38
rs764214627 Chr.9: 114626890-114626890 on Build GRCh38
rs368180091 Chr.9: 114626873-114626873 on Build GRCh38
rs747002507 Chr.9: 114626927-114626927 on Build GRCh38
rs376099024 Chr.9: 114627036-114627036 on Build GRCh38
rs201504427 Chr.9: 114627001-114627001 on Build GRCh38
rs779691240 Chr.9: 114626917-114626917 on Build GRCh38
rs1536737 Chr.9: 114627198-114627198 on Build GRCh38
rs1054242982 Chr.9: 114627092-114627092 on Build GRCh38
rs370551840 Chr.9: 114627022-114627022 on Build GRCh38
rs761258746 Chr.9: 114626982-114626982 on Build GRCh38
rs935665498 Chr.9: 114627070-114627070 on Build GRCh38
rs986909875 Chr.9: 114626813-114626813 on Build GRCh38
rs950441209 Chr.9: 114626741-114626741 on Build GRCh38
rs776013367 Chr.9: 114626884-114626884 on Build GRCh38
rs368743512 Chr.9: 114627025-114627025 on Build GRCh38

View Full Product Details