Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114644602-114645182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM268
Entrez Gene ID
203197
Gene Name
transmembrane protein 268
Gene Aliases
C9orf91
UniGene
Hs.522357
Gene Chromosome Location
Chr.9: 114603897-114646423 on Build GRCh38
Transcripts
NM_153045.3 XM_006716997.1 XM_011518350.1 XM_011518351.2 XM_011518352.1 XM_011518353.1 XM_011518354.1 XM_011518355.1 XM_017014429.1 XM_017014430.1

SNP Detail

SNP ID Chromosome Location
rs771335470 Chr.9: 114644955-114644955 on Build GRCh38
rs141547701 Chr.9: 114645005-114645005 on Build GRCh38
rs974147636 Chr.9: 114644922-114644922 on Build GRCh38
rs386737864 Chr.9: 114644683-114644685 on Build GRCh38
rs142453550 Chr.9: 114644885-114644885 on Build GRCh38
rs868146445 Chr.9: 114645079-114645079 on Build GRCh38
rs769978160 Chr.9: 114645016-114645016 on Build GRCh38
rs111530413 Chr.9: 114644684-114644684 on Build GRCh38
rs886708043 Chr.9: 114645041-114645041 on Build GRCh38
rs1008426300 Chr.9: 114644778-114644778 on Build GRCh38
rs10759727 Chr.9: 114644683-114644683 on Build GRCh38
rs944182178 Chr.9: 114644971-114644971 on Build GRCh38
rs940021871 Chr.9: 114644657-114644657 on Build GRCh38
rs919892765 Chr.9: 114644948-114644948 on Build GRCh38
rs908656223 Chr.9: 114644653-114644653 on Build GRCh38
rs58090448 Chr.9: 114644886-114644886 on Build GRCh38
rs908625000 Chr.9: 114644888-114644888 on Build GRCh38
rs902684696 Chr.9: 114644980-114644980 on Build GRCh38
rs907018807 Chr.9: 114645077-114645077 on Build GRCh38
rs989413434 Chr.9: 114644890-114644890 on Build GRCh38
rs1053550994 Chr.9: 114644993-114644993 on Build GRCh38
rs773795680 Chr.9: 114644901-114644901 on Build GRCh38
rs386737865 Chr.9: 114644885-114644886 on Build GRCh38
rs920046108 Chr.9: 114645059-114645059 on Build GRCh38
rs932873497 Chr.9: 114644727-114644727 on Build GRCh38
rs1031602749 Chr.9: 114645098-114645098 on Build GRCh38
rs11537999 Chr.9: 114644819-114644819 on Build GRCh38
rs546515669 Chr.9: 114644879-114644879 on Build GRCh38
rs1010867241 Chr.9: 114645129-114645129 on Build GRCh38
rs891276443 Chr.9: 114644776-114644776 on Build GRCh38
rs747422636 Chr.9: 114644921-114644921 on Build GRCh38
rs554389302 Chr.9: 114644757-114644757 on Build GRCh38
rs765536467 Chr.9: 114644995-114644995 on Build GRCh38
rs986736642 Chr.9: 114644960-114644961 on Build GRCh38
rs548252518 Chr.9: 114645078-114645078 on Build GRCh38
rs541411850 Chr.9: 114644911-114644911 on Build GRCh38
rs944234090 Chr.9: 114645123-114645123 on Build GRCh38
rs147029035 Chr.9: 114645044-114645044 on Build GRCh38
rs569811636 Chr.9: 114645049-114645049 on Build GRCh38
rs971754325 Chr.9: 114644841-114644841 on Build GRCh38
rs902299735 Chr.9: 114644805-114644805 on Build GRCh38
rs974550802 Chr.9: 114645050-114645050 on Build GRCh38
rs1032383052 Chr.9: 114644858-114644858 on Build GRCh38
rs373777839 Chr.9: 114644859-114644859 on Build GRCh38
rs191223130 Chr.9: 114644994-114644994 on Build GRCh38
rs1024917794 Chr.9: 114644882-114644882 on Build GRCh38
rs932698322 Chr.9: 114644956-114644956 on Build GRCh38
rs1042678492 Chr.9: 114645157-114645157 on Build GRCh38
rs527362352 Chr.9: 114644963-114644963 on Build GRCh38
rs138406200 Chr.9: 114645061-114645061 on Build GRCh38
rs539520661 Chr.9: 114645031-114645031 on Build GRCh38
rs577538758 Chr.9: 114644834-114644834 on Build GRCh38
rs1000592606 Chr.9: 114644986-114644986 on Build GRCh38
rs568224106 Chr.9: 114644753-114644753 on Build GRCh38
rs915336842 Chr.9: 114644904-114644904 on Build GRCh38
rs147946774 Chr.9: 114644973-114644973 on Build GRCh38
rs1012757619 Chr.9: 114644701-114644701 on Build GRCh38
rs1000627047 Chr.9: 114644857-114644857 on Build GRCh38
rs1046005642 Chr.9: 114645074-114645074 on Build GRCh38
rs1011938691 Chr.9: 114644998-114644998 on Build GRCh38
rs1020715605 Chr.9: 114644800-114644800 on Build GRCh38
rs1050413053 Chr.9: 114644962-114644962 on Build GRCh38
rs567255550 Chr.9: 114645028-114645028 on Build GRCh38
rs10759728 Chr.9: 114644685-114644685 on Build GRCh38
rs561746922 Chr.9: 114644916-114644916 on Build GRCh38
rs374759844 Chr.9: 114644797-114644798 on Build GRCh38

View Full Product Details