Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15785665-15786213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA5B
Entrez Gene ID
11238
Gene Name
carbonic anhydrase 5B
Gene Aliases
CA-VB
UniGene
Hs.653287
Gene Chromosome Location
Chr.X: 15738281-15787625 on Build GRCh38
Transcripts
NM_007220.3 NR_002725.2

Gene Symbol
INE2
Entrez Gene ID
8551
Gene Name
inactivation escape 2 (non-protein coding)
Gene Aliases
NCRNA00011
UniGene
Hs.653287
Gene Chromosome Location
Chr.X: 15785716-15787589 on Build GRCh38
Transcripts
NM_007220.3 NR_002725.2

SNP Detail

SNP ID Chromosome Location
rs1048062187 Chr.X: 15786110-15786110 on Build GRCh38
rs911230469 Chr.X: 15785990-15785990 on Build GRCh38
rs12850450 Chr.X: 15785714-15785714 on Build GRCh38
rs201342194 Chr.X: 15785867-15785867 on Build GRCh38
rs758864620 Chr.X: 15786087-15786087 on Build GRCh38
rs376588764 Chr.X: 15786180-15786180 on Build GRCh38
rs181633991 Chr.X: 15785902-15785902 on Build GRCh38
rs964944354 Chr.X: 15786028-15786028 on Build GRCh38
rs989090277 Chr.X: 15785692-15785692 on Build GRCh38
rs759870103 Chr.X: 15785780-15785780 on Build GRCh38
rs67321658 Chr.X: 15785868-15785869 on Build GRCh38
rs751692722 Chr.X: 15785901-15785901 on Build GRCh38
rs933675971 Chr.X: 15785804-15785805 on Build GRCh38
rs1033566304 Chr.X: 15785956-15785956 on Build GRCh38
rs755882511 Chr.X: 15786019-15786019 on Build GRCh38
rs933986658 Chr.X: 15785823-15785823 on Build GRCh38
rs12851914 Chr.X: 15785707-15785707 on Build GRCh38
rs12851913 Chr.X: 15785703-15785703 on Build GRCh38
rs371010730 Chr.X: 15785793-15785793 on Build GRCh38
rs921009304 Chr.X: 15785733-15785734 on Build GRCh38
rs746190710 Chr.X: 15785807-15785807 on Build GRCh38
rs61247855 Chr.X: 15785867-15785868 on Build GRCh38
rs6632734 Chr.X: 15785883-15785883 on Build GRCh38
rs986471468 Chr.X: 15785845-15785845 on Build GRCh38
rs6632735 Chr.X: 15785884-15785884 on Build GRCh38
rs6632732 Chr.X: 15785874-15785874 on Build GRCh38
rs1038469188 Chr.X: 15786020-15786020 on Build GRCh38
rs969740082 Chr.X: 15785948-15785948 on Build GRCh38
rs922873480 Chr.X: 15785967-15785967 on Build GRCh38
rs780580456 Chr.X: 15786090-15786090 on Build GRCh38
rs546436017 Chr.X: 15786173-15786173 on Build GRCh38
rs924251871 Chr.X: 15785846-15785846 on Build GRCh38
rs999918834 Chr.X: 15785952-15785952 on Build GRCh38
rs375796113 Chr.X: 15785792-15785792 on Build GRCh38
rs779692348 Chr.X: 15786037-15786037 on Build GRCh38
rs918697626 Chr.X: 15786071-15786071 on Build GRCh38
rs6632733 Chr.X: 15785875-15785875 on Build GRCh38
rs941403274 Chr.X: 15785992-15785992 on Build GRCh38
rs907305815 Chr.X: 15785934-15785934 on Build GRCh38
rs989406222 Chr.X: 15785963-15785963 on Build GRCh38
rs899018900 Chr.X: 15785899-15785899 on Build GRCh38
rs903443190 Chr.X: 15786015-15786015 on Build GRCh38
rs767482209 Chr.X: 15785930-15785931 on Build GRCh38
rs955109011 Chr.X: 15786113-15786113 on Build GRCh38
rs778649083 Chr.X: 15786164-15786167 on Build GRCh38
rs371910788 Chr.X: 15785961-15785961 on Build GRCh38
rs28633488 Chr.X: 15785873-15785873 on Build GRCh38
rs1049795435 Chr.X: 15785837-15785837 on Build GRCh38
rs1047627402 Chr.X: 15785903-15785903 on Build GRCh38
rs1013873860 Chr.X: 15785931-15785931 on Build GRCh38
rs755860818 Chr.X: 15785865-15785870 on Build GRCh38
rs1022621018 Chr.X: 15785933-15785933 on Build GRCh38
rs895745252 Chr.X: 15785914-15785914 on Build GRCh38
rs6632731 Chr.X: 15785868-15785868 on Build GRCh38
rs748757429 Chr.X: 15785864-15785865 on Build GRCh38
rs753448078 Chr.X: 15786091-15786091 on Build GRCh38
rs767745106 Chr.X: 15785870-15785871 on Build GRCh38
rs986520925 Chr.X: 15786172-15786172 on Build GRCh38
rs368348669 Chr.X: 15785873-15785874 on Build GRCh38
rs955640180 Chr.X: 15785972-15785972 on Build GRCh38
rs186530382 Chr.X: 15785983-15785983 on Build GRCh38
rs1016909172 Chr.X: 15786025-15786025 on Build GRCh38
rs865794718 Chr.X: 15785863-15785863 on Build GRCh38
rs892995534 Chr.X: 15785914-15785914 on Build GRCh38
rs775603375 Chr.X: 15785864-15785864 on Build GRCh38
rs1054770005 Chr.X: 15785896-15785897 on Build GRCh38

View Full Product Details