Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2884291-2884831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN1C
Entrez Gene ID
1028
Gene Name
cyclin dependent kinase inhibitor 1C
Gene Aliases
BWCR, BWS, KIP2, WBS, p57, p57Kip2
UniGene
Hs.106070
Gene Chromosome Location
Chr.11: 2883218-2885804 on Build GRCh38
Transcripts
NM_000076.2 NM_001122630.1 NM_001122631.1 XM_005252732.4 XM_017017088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6354783 c.819C>G p.S273S Chr.11: 2884671-2884671 on Build GRCh38
3666257 c.776C>A p.A259D Chr.11: 2884714-2884714 on Build GRCh38
6352320 c.708G>A p.E236E Chr.11: 2884782-2884782 on Build GRCh38
3666256 c.805G>A p.G269R Chr.11: 2884685-2884685 on Build GRCh38
7213283 c.734_735delGG p.G245fs*40 Chr.11: 2884755-2884756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761539484 Chr.11: 2884806-2884806 on Build GRCh38
rs483352991 Chr.11: 2884769-2884769 on Build GRCh38
rs113214469 Chr.11: 2884510-2884510 on Build GRCh38
rs779262119 Chr.11: 2884641-2884641 on Build GRCh38
rs111273511 Chr.11: 2884609-2884609 on Build GRCh38
rs748889587 Chr.11: 2884645-2884654 on Build GRCh38
rs1052164294 Chr.11: 2884513-2884513 on Build GRCh38
rs1016927638 Chr.11: 2884567-2884567 on Build GRCh38
rs483352990 Chr.11: 2884784-2884784 on Build GRCh38
rs960837503 Chr.11: 2884588-2884588 on Build GRCh38
rs552952278 Chr.11: 2884577-2884577 on Build GRCh38
rs368106038 Chr.11: 2884597-2884597 on Build GRCh38
rs394513 Chr.11: 2884383-2884383 on Build GRCh38
rs570525745 Chr.11: 2884473-2884473 on Build GRCh38
rs546016935 Chr.11: 2884788-2884788 on Build GRCh38
rs1000373562 Chr.11: 2884316-2884316 on Build GRCh38
rs1056815146 Chr.11: 2884470-2884470 on Build GRCh38
rs483352989 Chr.11: 2884787-2884787 on Build GRCh38
rs762619956 Chr.11: 2884707-2884707 on Build GRCh38
rs954200255 Chr.11: 2884365-2884365 on Build GRCh38
rs773932111 Chr.11: 2884793-2884793 on Build GRCh38
rs771502215 Chr.11: 2884811-2884811 on Build GRCh38
rs745815931 Chr.11: 2884689-2884689 on Build GRCh38
rs797045445 Chr.11: 2884796-2884796 on Build GRCh38
rs1031878233 Chr.11: 2884339-2884340 on Build GRCh38
rs558762873 Chr.11: 2884404-2884404 on Build GRCh38
rs766858608 Chr.11: 2884717-2884717 on Build GRCh38
rs758620755 Chr.11: 2884743-2884743 on Build GRCh38
rs1007312392 Chr.11: 2884544-2884544 on Build GRCh38
rs394514 Chr.11: 2884390-2884390 on Build GRCh38
rs3741341 Chr.11: 2884782-2884782 on Build GRCh38
rs998865853 Chr.11: 2884340-2884340 on Build GRCh38
rs747104008 Chr.11: 2884686-2884686 on Build GRCh38
rs483352994 Chr.11: 2884729-2884733 on Build GRCh38
rs879220376 Chr.11: 2884565-2884565 on Build GRCh38
rs779445226 Chr.11: 2884631-2884640 on Build GRCh38
rs483352992 Chr.11: 2884761-2884761 on Build GRCh38
rs534934487 Chr.11: 2884528-2884528 on Build GRCh38
rs483352993 Chr.11: 2884759-2884759 on Build GRCh38
rs781340171 Chr.11: 2884688-2884688 on Build GRCh38
rs773881541 Chr.11: 2884715-2884715 on Build GRCh38
rs753022994 Chr.11: 2884741-2884741 on Build GRCh38
rs891995652 Chr.11: 2884516-2884516 on Build GRCh38
rs972784801 Chr.11: 2884382-2884382 on Build GRCh38
rs918659334 Chr.11: 2884582-2884582 on Build GRCh38
rs754283907 Chr.11: 2884721-2884721 on Build GRCh38
rs962683501 Chr.11: 2884569-2884569 on Build GRCh38
rs387907225 Chr.11: 2884670-2884670 on Build GRCh38
rs564726941 Chr.11: 2884792-2884792 on Build GRCh38
rs962737302 Chr.11: 2884372-2884372 on Build GRCh38
rs754196288 Chr.11: 2884634-2884634 on Build GRCh38
rs999313274 Chr.11: 2884508-2884508 on Build GRCh38
rs777855717 Chr.11: 2884660-2884660 on Build GRCh38
rs956188423 Chr.11: 2884339-2884339 on Build GRCh38
rs985456061 Chr.11: 2884369-2884369 on Build GRCh38
rs879939998 Chr.11: 2884377-2884377 on Build GRCh38
rs998334945 Chr.11: 2884519-2884519 on Build GRCh38
rs761296394 Chr.11: 2884716-2884716 on Build GRCh38
rs989415252 Chr.11: 2884468-2884468 on Build GRCh38
rs541422709 Chr.11: 2884621-2884621 on Build GRCh38
rs758576724 Chr.11: 2884655-2884655 on Build GRCh38
rs751650132 Chr.11: 2884758-2884758 on Build GRCh38
rs947713247 Chr.11: 2884474-2884474 on Build GRCh38
rs934968787 Chr.11: 2884633-2884633 on Build GRCh38
rs779975908 Chr.11: 2884791-2884791 on Build GRCh38
rs760038657 Chr.11: 2884727-2884727 on Build GRCh38
rs878853640 Chr.11: 2884778-2884778 on Build GRCh38
rs757359164 Chr.11: 2884773-2884773 on Build GRCh38
rs752846343 Chr.11: 2884651-2884651 on Build GRCh38
rs991965444 Chr.11: 2884604-2884604 on Build GRCh38
rs775458829 Chr.11: 2884704-2884704 on Build GRCh38
rs1030347802 Chr.11: 2884527-2884527 on Build GRCh38
rs878853641 Chr.11: 2884745-2884745 on Build GRCh38
rs398331 Chr.11: 2884486-2884486 on Build GRCh38
rs924962401 Chr.11: 2884630-2884631 on Build GRCh38
rs483352988 Chr.11: 2884802-2884802 on Build GRCh38
rs556682082 Chr.11: 2884755-2884755 on Build GRCh38
rs1043378194 Chr.11: 2884490-2884490 on Build GRCh38
rs371264 Chr.11: 2884638-2884638 on Build GRCh38
rs948030376 Chr.11: 2884612-2884612 on Build GRCh38
rs1029900408 Chr.11: 2884349-2884349 on Build GRCh38
rs515726203 Chr.11: 2884675-2884675 on Build GRCh38
rs781428134 Chr.11: 2884775-2884775 on Build GRCh38
rs903513294 Chr.11: 2884495-2884495 on Build GRCh38
rs755391541 Chr.11: 2884635-2884635 on Build GRCh38
rs878853642 Chr.11: 2884720-2884720 on Build GRCh38
rs916492470 Chr.11: 2884607-2884607 on Build GRCh38
rs768242627 Chr.11: 2884709-2884709 on Build GRCh38
rs104894200 Chr.11: 2884750-2884750 on Build GRCh38
rs765542916 Chr.11: 2884738-2884738 on Build GRCh38
rs901558295 Chr.11: 2884701-2884724 on Build GRCh38

View Full Product Details