Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2884696-2885221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN1C
Entrez Gene ID
1028
Gene Name
cyclin dependent kinase inhibitor 1C
Gene Aliases
BWCR, BWS, KIP2, WBS, p57, p57Kip2
UniGene
Hs.106070
Gene Chromosome Location
Chr.11: 2883218-2885804 on Build GRCh38
Transcripts
NM_000076.2 NM_001122630.1 NM_001122631.1 XM_005252732.4 XM_017017088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3943620 c.303C>T p.R101R Chr.11: 2885187-2885187 on Build GRCh38
5606257 c.512_523del12 p.A171_A174delAPVA Chr.11: 2884967-2884978 on Build GRCh38
5469108 c.627C>A p.A209A Chr.11: 2884863-2884863 on Build GRCh38
7447779 c.618_629del12 p.A213_P216delAPAP Chr.11: 2884861-2884872 on Build GRCh38
3666257 c.776C>A p.A259D Chr.11: 2884714-2884714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113374868 Chr.11: 2884951-2884951 on Build GRCh38
rs483352981 Chr.11: 2885018-2885018 on Build GRCh38
rs483352974 Chr.11: 2885182-2885183 on Build GRCh38
rs878853638 Chr.11: 2884841-2884870 on Build GRCh38
rs878853635 Chr.11: 2884878-2884879 on Build GRCh38
rs878853629 Chr.11: 2884936-2884947 on Build GRCh38
rs483352990 Chr.11: 2884784-2884784 on Build GRCh38
rs786205241 Chr.11: 2884860-2884861 on Build GRCh38
rs932963247 Chr.11: 2885163-2885163 on Build GRCh38
rs384713 Chr.11: 2884929-2884929 on Build GRCh38
rs763525000 Chr.11: 2885170-2885170 on Build GRCh38
rs483352987 Chr.11: 2884814-2884814 on Build GRCh38
rs483352978 Chr.11: 2885030-2885051 on Build GRCh38
rs570636789 Chr.11: 2885141-2885141 on Build GRCh38
rs551863674 Chr.11: 2885126-2885126 on Build GRCh38
rs533485167 Chr.11: 2884962-2884962 on Build GRCh38
rs387906399 Chr.11: 2885179-2885180 on Build GRCh38
rs878853627 Chr.11: 2884991-2884996 on Build GRCh38
rs751669088 Chr.11: 2884839-2884839 on Build GRCh38
rs773932111 Chr.11: 2884793-2884793 on Build GRCh38
rs771502215 Chr.11: 2884811-2884811 on Build GRCh38
rs778772076 Chr.11: 2885100-2885100 on Build GRCh38
rs766858608 Chr.11: 2884717-2884717 on Build GRCh38
rs758620755 Chr.11: 2884743-2884743 on Build GRCh38
rs3741342 Chr.11: 2884840-2884840 on Build GRCh38
rs763289302 Chr.11: 2884831-2884831 on Build GRCh38
rs1182342 Chr.11: 2884873-2884873 on Build GRCh38
rs483352994 Chr.11: 2884729-2884733 on Build GRCh38
rs771731330 Chr.11: 2885137-2885137 on Build GRCh38
rs878853630 Chr.11: 2884909-2884932 on Build GRCh38
rs540923047 Chr.11: 2884821-2884821 on Build GRCh38
rs528634940 Chr.11: 2884820-2884820 on Build GRCh38
rs945890937 Chr.11: 2885140-2885140 on Build GRCh38
rs904430375 Chr.11: 2885151-2885151 on Build GRCh38
rs483352992 Chr.11: 2884761-2884761 on Build GRCh38
rs773881541 Chr.11: 2884715-2884715 on Build GRCh38
rs753022994 Chr.11: 2884741-2884741 on Build GRCh38
rs878853631 Chr.11: 2884929-2884930 on Build GRCh38
rs931922290 Chr.11: 2885154-2885154 on Build GRCh38
rs534471786 Chr.11: 2885166-2885166 on Build GRCh38
rs191294997 Chr.11: 2884935-2884935 on Build GRCh38
rs774805080 Chr.11: 2885185-2885185 on Build GRCh38
rs2583439 Chr.11: 2884897-2884897 on Build GRCh38
rs747757242 Chr.11: 2885127-2885127 on Build GRCh38
rs550978405 Chr.11: 2884920-2884920 on Build GRCh38
rs751650132 Chr.11: 2884758-2884758 on Build GRCh38
rs772704243 Chr.11: 2884840-2884841 on Build GRCh38
rs786205235 Chr.11: 2885156-2885157 on Build GRCh38
rs878853633 Chr.11: 2884902-2884902 on Build GRCh38
rs878853639 Chr.11: 2884861-2884866 on Build GRCh38
rs779975908 Chr.11: 2884791-2884791 on Build GRCh38
rs760038657 Chr.11: 2884727-2884727 on Build GRCh38
rs772684721 Chr.11: 2885138-2885138 on Build GRCh38
rs786205237 Chr.11: 2884855-2884855 on Build GRCh38
rs878853641 Chr.11: 2884745-2884745 on Build GRCh38
rs483352975 Chr.11: 2885169-2885169 on Build GRCh38
rs556682082 Chr.11: 2884755-2884755 on Build GRCh38
rs562237921 Chr.11: 2884850-2884850 on Build GRCh38
rs878853636 Chr.11: 2884873-2884890 on Build GRCh38
rs786205240 Chr.11: 2884846-2884849 on Build GRCh38
rs781428134 Chr.11: 2884775-2884775 on Build GRCh38
rs794726872 Chr.11: 2884878-2884878 on Build GRCh38
rs889984547 Chr.11: 2884959-2884964 on Build GRCh38
rs770277810 Chr.11: 2885145-2885145 on Build GRCh38
rs104894200 Chr.11: 2884750-2884750 on Build GRCh38
rs529326848 Chr.11: 2884890-2884890 on Build GRCh38
rs923004197 Chr.11: 2885160-2885160 on Build GRCh38
rs786205239 Chr.11: 2884859-2884859 on Build GRCh38
rs761539484 Chr.11: 2884806-2884806 on Build GRCh38
rs1028702668 Chr.11: 2884918-2884935 on Build GRCh38
rs483352991 Chr.11: 2884769-2884769 on Build GRCh38
rs896475967 Chr.11: 2885062-2885062 on Build GRCh38
rs764328739 Chr.11: 2885181-2885181 on Build GRCh38
rs767656648 Chr.11: 2884860-2884860 on Build GRCh38
rs399771 Chr.11: 2884971-2884971 on Build GRCh38
rs775889163 Chr.11: 2884826-2884826 on Build GRCh38
rs546016935 Chr.11: 2884788-2884788 on Build GRCh38
rs878853628 Chr.11: 2884965-2884965 on Build GRCh38
rs483352980 Chr.11: 2885023-2885024 on Build GRCh38
rs764536797 Chr.11: 2884833-2884833 on Build GRCh38
rs778468310 Chr.11: 2884841-2884864 on Build GRCh38
rs754671425 Chr.11: 2885047-2885047 on Build GRCh38
rs483352989 Chr.11: 2884787-2884787 on Build GRCh38
rs385101 Chr.11: 2885038-2885038 on Build GRCh38
rs997267634 Chr.11: 2884909-2884920 on Build GRCh38
rs898729531 Chr.11: 2885007-2885007 on Build GRCh38
rs797045445 Chr.11: 2884796-2884796 on Build GRCh38
rs771450542 Chr.11: 2884857-2884858 on Build GRCh38
rs878853626 Chr.11: 2885034-2885034 on Build GRCh38
rs1050598447 Chr.11: 2884939-2884939 on Build GRCh38
rs3741341 Chr.11: 2884782-2884782 on Build GRCh38
rs565544512 Chr.11: 2884967-2884978 on Build GRCh38
rs112196492 Chr.11: 2884923-2884923 on Build GRCh38
rs753342439 Chr.11: 2885039-2885039 on Build GRCh38
rs483352993 Chr.11: 2884759-2884759 on Build GRCh38
rs483352985 Chr.11: 2884849-2884849 on Build GRCh38
rs1038604691 Chr.11: 2884991-2884991 on Build GRCh38
rs754283907 Chr.11: 2884721-2884721 on Build GRCh38
rs564726941 Chr.11: 2884792-2884792 on Build GRCh38
rs878853632 Chr.11: 2884918-2884923 on Build GRCh38
rs786205238 Chr.11: 2884854-2884855 on Build GRCh38
rs761296394 Chr.11: 2884716-2884716 on Build GRCh38
rs759134767 Chr.11: 2884861-2884872 on Build GRCh38
rs786205234 Chr.11: 2885041-2885041 on Build GRCh38
rs757359164 Chr.11: 2884773-2884773 on Build GRCh38
rs878853640 Chr.11: 2884778-2884778 on Build GRCh38
rs878853625 Chr.11: 2885046-2885046 on Build GRCh38
rs878853634 Chr.11: 2884879-2884890 on Build GRCh38
rs750581456 Chr.11: 2884865-2884865 on Build GRCh38
rs483352988 Chr.11: 2884802-2884802 on Build GRCh38
rs2583440 Chr.11: 2884879-2884879 on Build GRCh38
rs786205236 Chr.11: 2885089-2885090 on Build GRCh38
rs878853637 Chr.11: 2884867-2884890 on Build GRCh38
rs878853642 Chr.11: 2884720-2884720 on Build GRCh38
rs747360016 Chr.11: 2884843-2884860 on Build GRCh38
rs757586907 Chr.11: 2884851-2884851 on Build GRCh38
rs1013695050 Chr.11: 2885090-2885090 on Build GRCh38
rs372633 Chr.11: 2885014-2885014 on Build GRCh38
rs765542916 Chr.11: 2884738-2884738 on Build GRCh38

View Full Product Details