Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48748344-48748853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP152
Entrez Gene ID
22995
Gene Name
centrosomal protein 152
Gene Aliases
MCPH4, MCPH9, SCKL5
UniGene
Hs.443005
Gene Chromosome Location
Chr.15: 48662534-48811901 on Build GRCh38
Transcripts
NM_001194998.1 NM_014985.3 XM_006720437.3 XM_011521373.2 XM_011521374.2 XM_011521375.2 XM_011521378.2 XM_011521379.2 XM_011521381.2 XM_017022014.1 XM_017022015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7204307 c.3520G>A p.E1174K Chr.15: 48748557-48748557 on Build GRCh38
6278514 c.3559G>A p.G1187R Chr.15: 48748518-48748518 on Build GRCh38
5802817 c.3629T>C p.I1210T Chr.15: 48748448-48748448 on Build GRCh38
5808151 c.3505G>T p.G1169* Chr.15: 48748572-48748572 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs911905122 Chr.15: 48748712-48748712 on Build GRCh38
rs767248208 Chr.15: 48748554-48748554 on Build GRCh38
rs751912220 Chr.15: 48748539-48748539 on Build GRCh38
rs398027188 Chr.15: 48748683-48748683 on Build GRCh38
rs759788211 Chr.15: 48748547-48748547 on Build GRCh38
rs941054950 Chr.15: 48748666-48748666 on Build GRCh38
rs1032950585 Chr.15: 48748426-48748426 on Build GRCh38
rs759405983 Chr.15: 48748446-48748446 on Build GRCh38
rs972146111 Chr.15: 48748744-48748744 on Build GRCh38
rs1037278523 Chr.15: 48748713-48748713 on Build GRCh38
rs865900311 Chr.15: 48748542-48748542 on Build GRCh38
rs117061285 Chr.15: 48748727-48748727 on Build GRCh38
rs573740782 Chr.15: 48748669-48748669 on Build GRCh38
rs906058301 Chr.15: 48748784-48748784 on Build GRCh38
rs767740955 Chr.15: 48748635-48748636 on Build GRCh38
rs146078987 Chr.15: 48748540-48748540 on Build GRCh38
rs369867192 Chr.15: 48748566-48748566 on Build GRCh38
rs373282905 Chr.15: 48748635-48748635 on Build GRCh38
rs932363651 Chr.15: 48748610-48748610 on Build GRCh38
rs955334640 Chr.15: 48748600-48748600 on Build GRCh38
rs892094392 Chr.15: 48748445-48748445 on Build GRCh38
rs922435112 Chr.15: 48748568-48748568 on Build GRCh38
rs755542727 Chr.15: 48748767-48748767 on Build GRCh38
rs570388512 Chr.15: 48748380-48748380 on Build GRCh38
rs752277098 Chr.15: 48748640-48748640 on Build GRCh38
rs761865621 Chr.15: 48748601-48748601 on Build GRCh38
rs1001554333 Chr.15: 48748808-48748808 on Build GRCh38
rs1045228353 Chr.15: 48748811-48748811 on Build GRCh38
rs34352957 Chr.15: 48748668-48748668 on Build GRCh38
rs980165206 Chr.15: 48748393-48748393 on Build GRCh38
rs957623896 Chr.15: 48748429-48748429 on Build GRCh38
rs1014463831 Chr.15: 48748757-48748757 on Build GRCh38
rs374227858 Chr.15: 48748728-48748728 on Build GRCh38
rs1028129886 Chr.15: 48748599-48748599 on Build GRCh38
rs754336519 Chr.15: 48748428-48748428 on Build GRCh38
rs989399178 Chr.15: 48748469-48748469 on Build GRCh38
rs987081522 Chr.15: 48748709-48748709 on Build GRCh38
rs930700504 Chr.15: 48748810-48748810 on Build GRCh38
rs1049524715 Chr.15: 48748627-48748627 on Build GRCh38
rs139926728 Chr.15: 48748758-48748758 on Build GRCh38

View Full Product Details