Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 165378832-165379375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPE
Entrez Gene ID
1363
Gene Name
carboxypeptidase E
Gene Aliases
CPH
UniGene
Hs.75360
Gene Chromosome Location
Chr.4: 165378942-165498547 on Build GRCh38
Transcripts
NM_001873.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5355937 c.120C>A p.R40R Chr.4: 165379341-165379341 on Build GRCh38
6545903 c.7G>T p.G3W Chr.4: 165379228-165379228 on Build GRCh38
5769941 c.143G>A p.G48D Chr.4: 165379364-165379364 on Build GRCh38
4171314 c.23C>G p.A8G Chr.4: 165379244-165379244 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1001192073 Chr.4: 165379071-165379071 on Build GRCh38
rs888306541 Chr.4: 165378869-165378869 on Build GRCh38
rs548739022 Chr.4: 165379039-165379039 on Build GRCh38
rs976331601 Chr.4: 165379145-165379145 on Build GRCh38
rs768504422 Chr.4: 165379290-165379312 on Build GRCh38
rs925715158 Chr.4: 165378947-165378947 on Build GRCh38
rs771957496 Chr.4: 165379316-165379316 on Build GRCh38
rs1001536008 Chr.4: 165378987-165378987 on Build GRCh38
rs961274941 Chr.4: 165378882-165378882 on Build GRCh38
rs944919219 Chr.4: 165378986-165378986 on Build GRCh38
rs887865654 Chr.4: 165379021-165379021 on Build GRCh38
rs147957257 Chr.4: 165378932-165378932 on Build GRCh38
rs1034805944 Chr.4: 165378989-165378989 on Build GRCh38
rs1033217112 Chr.4: 165379096-165379096 on Build GRCh38
rs933881993 Chr.4: 165379100-165379100 on Build GRCh38
rs565869186 Chr.4: 165379249-165379249 on Build GRCh38
rs947321276 Chr.4: 165379114-165379114 on Build GRCh38
rs545073973 Chr.4: 165378938-165378938 on Build GRCh38
rs1049050392 Chr.4: 165379180-165379180 on Build GRCh38
rs921906422 Chr.4: 165379005-165379005 on Build GRCh38
rs543990252 Chr.4: 165378872-165378872 on Build GRCh38
rs988992374 Chr.4: 165378968-165378968 on Build GRCh38
rs940744414 Chr.4: 165379236-165379236 on Build GRCh38
rs538449558 Chr.4: 165379234-165379234 on Build GRCh38
rs1014157674 Chr.4: 165379045-165379045 on Build GRCh38
rs757266004 Chr.4: 165379266-165379266 on Build GRCh38
rs1039990844 Chr.4: 165378920-165378920 on Build GRCh38
rs909514067 Chr.4: 165378886-165378886 on Build GRCh38
rs531555316 Chr.4: 165379044-165379044 on Build GRCh38
rs537253101 Chr.4: 165379034-165379034 on Build GRCh38
rs966277433 Chr.4: 165379141-165379141 on Build GRCh38
rs896963496 Chr.4: 165379284-165379284 on Build GRCh38
rs909639157 Chr.4: 165379041-165379041 on Build GRCh38
rs1040370772 Chr.4: 165378991-165378991 on Build GRCh38
rs949851535 Chr.4: 165379309-165379309 on Build GRCh38
rs1013934967 Chr.4: 165378906-165378906 on Build GRCh38
rs1039369067 Chr.4: 165379090-165379090 on Build GRCh38
rs922285057 Chr.4: 165379093-165379093 on Build GRCh38
rs11552516 Chr.4: 165379172-165379172 on Build GRCh38
rs913521818 Chr.4: 165378969-165378969 on Build GRCh38
rs560313170 Chr.4: 165378945-165378945 on Build GRCh38
rs1010753915 Chr.4: 165379220-165379220 on Build GRCh38
rs1049635844 Chr.4: 165379017-165379017 on Build GRCh38
rs932014557 Chr.4: 165379164-165379164 on Build GRCh38
rs931986257 Chr.4: 165379015-165379015 on Build GRCh38
rs1036885508 Chr.4: 165379271-165379271 on Build GRCh38
rs1023990575 Chr.4: 165379064-165379064 on Build GRCh38
rs909295102 Chr.4: 165379204-165379204 on Build GRCh38
rs745682954 Chr.4: 165379315-165379315 on Build GRCh38
rs111586791 Chr.4: 165378935-165378935 on Build GRCh38
rs571068220 Chr.4: 165379113-165379113 on Build GRCh38
rs993070553 Chr.4: 165378994-165378994 on Build GRCh38
rs536627016 Chr.4: 165379298-165379298 on Build GRCh38
rs778940259 Chr.4: 165379308-165379308 on Build GRCh38
rs145384396 Chr.4: 165378957-165378957 on Build GRCh38
rs554021167 Chr.4: 165379040-165379040 on Build GRCh38
rs957138370 Chr.4: 165379098-165379098 on Build GRCh38
rs922167029 Chr.4: 165379162-165379162 on Build GRCh38
rs1019562151 Chr.4: 165378962-165378980 on Build GRCh38
rs1049594014 Chr.4: 165378862-165378862 on Build GRCh38
rs553814573 Chr.4: 165379248-165379248 on Build GRCh38
rs549940521 Chr.4: 165379107-165379107 on Build GRCh38
rs1015964900 Chr.4: 165378881-165378881 on Build GRCh38
rs141853034 Chr.4: 165378933-165378933 on Build GRCh38
rs756315983 Chr.4: 165379068-165379068 on Build GRCh38

View Full Product Details