Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36929191-36929727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF2RB
Entrez Gene ID
1439
Gene Name
colony stimulating factor 2 receptor beta common subunit
Gene Aliases
CD131, CDw131, IL3RB, IL5RB, SMDP5
UniGene
Hs.592192
Gene Chromosome Location
Chr.22: 36913574-36940449 on Build GRCh38
Transcripts
NM_000395.2 XM_005261340.3 XM_011529903.2 XM_011529904.2 XM_011529905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6253595 c.530G>T p.R177L Chr.22: 36929540-36929540 on Build GRCh38
444946 c.631C>T p.R211* Chr.22: 36929720-36929720 on Build GRCh38
1033898 c.528G>A p.K176K Chr.22: 36929538-36929538 on Build GRCh38
3554020 c.455G>A p.W152* Chr.22: 36929465-36929465 on Build GRCh38
5529262 c.445C>T p.L149L Chr.22: 36929455-36929455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953725230 Chr.22: 36929700-36929700 on Build GRCh38
rs1042340585 Chr.22: 36929265-36929265 on Build GRCh38
rs774995639 Chr.22: 36929475-36929475 on Build GRCh38
rs61737702 Chr.22: 36929642-36929642 on Build GRCh38
rs201329664 Chr.22: 36929354-36929354 on Build GRCh38
rs539179276 Chr.22: 36929532-36929532 on Build GRCh38
rs751990782 Chr.22: 36929366-36929366 on Build GRCh38
rs139583249 Chr.22: 36929701-36929701 on Build GRCh38
rs370781346 Chr.22: 36929551-36929551 on Build GRCh38
rs771979424 Chr.22: 36929415-36929415 on Build GRCh38
rs749562198 Chr.22: 36929399-36929399 on Build GRCh38
rs547060760 Chr.22: 36929493-36929493 on Build GRCh38
rs757739081 Chr.22: 36929375-36929375 on Build GRCh38
rs768091118 Chr.22: 36929610-36929610 on Build GRCh38
rs773022268 Chr.22: 36929352-36929352 on Build GRCh38
rs991635780 Chr.22: 36929301-36929301 on Build GRCh38
rs952276881 Chr.22: 36929339-36929339 on Build GRCh38
rs370120556 Chr.22: 36929673-36929673 on Build GRCh38
rs529939805 Chr.22: 36929571-36929571 on Build GRCh38
rs1012347639 Chr.22: 36929315-36929315 on Build GRCh38
rs767911516 Chr.22: 36929380-36929380 on Build GRCh38
rs966332244 Chr.22: 36929672-36929672 on Build GRCh38
rs200604301 Chr.22: 36929358-36929358 on Build GRCh38
rs540559131 Chr.22: 36929598-36929598 on Build GRCh38
rs959459178 Chr.22: 36929295-36929295 on Build GRCh38
rs773038230 Chr.22: 36929416-36929416 on Build GRCh38
rs376547021 Chr.22: 36929612-36929612 on Build GRCh38
rs35082017 Chr.22: 36929462-36929462 on Build GRCh38
rs150587930 Chr.22: 36929684-36929684 on Build GRCh38
rs768051950 Chr.22: 36929496-36929496 on Build GRCh38
rs1018533401 Chr.22: 36929230-36929230 on Build GRCh38
rs777336576 Chr.22: 36929690-36929690 on Build GRCh38
rs762165060 Chr.22: 36929584-36929614 on Build GRCh38
rs766424551 Chr.22: 36929675-36929675 on Build GRCh38
rs765484350 Chr.22: 36929641-36929641 on Build GRCh38
rs1027762796 Chr.22: 36929317-36929317 on Build GRCh38
rs199604353 Chr.22: 36929540-36929540 on Build GRCh38
rs76327427 Chr.22: 36929363-36929363 on Build GRCh38
rs922323586 Chr.22: 36929413-36929413 on Build GRCh38
rs755188718 Chr.22: 36929539-36929539 on Build GRCh38
rs748209868 Chr.22: 36929405-36929405 on Build GRCh38
rs902508678 Chr.22: 36929272-36929272 on Build GRCh38
rs759239121 Chr.22: 36929443-36929443 on Build GRCh38
rs963930043 Chr.22: 36929231-36929231 on Build GRCh38
rs977899700 Chr.22: 36929293-36929293 on Build GRCh38
rs1035031141 Chr.22: 36929294-36929294 on Build GRCh38
rs776237333 Chr.22: 36929440-36929440 on Build GRCh38
rs1045985309 Chr.22: 36929314-36929314 on Build GRCh38
rs1003547219 Chr.22: 36929287-36929287 on Build GRCh38
rs998825916 Chr.22: 36929463-36929463 on Build GRCh38
rs916092508 Chr.22: 36929304-36929304 on Build GRCh38
rs746992696 Chr.22: 36929570-36929570 on Build GRCh38
rs770782977 Chr.22: 36929439-36929439 on Build GRCh38
rs764825237 Chr.22: 36929455-36929455 on Build GRCh38
rs776552254 Chr.22: 36929307-36929307 on Build GRCh38
rs367633751 Chr.22: 36929665-36929665 on Build GRCh38
rs779000979 Chr.22: 36929404-36929404 on Build GRCh38
rs113156338 Chr.22: 36929661-36929661 on Build GRCh38
rs754189724 Chr.22: 36929621-36929621 on Build GRCh38
rs777765485 Chr.22: 36929563-36929563 on Build GRCh38
rs189318720 Chr.22: 36929264-36929264 on Build GRCh38
rs893664049 Chr.22: 36929256-36929256 on Build GRCh38
rs775059195 Chr.22: 36929602-36929602 on Build GRCh38
rs748941609 Chr.22: 36929676-36929676 on Build GRCh38
rs899691848 Chr.22: 36929436-36929436 on Build GRCh38
rs750755016 Chr.22: 36929387-36929387 on Build GRCh38
rs866537612 Chr.22: 36929699-36929699 on Build GRCh38
rs950279218 Chr.22: 36929313-36929313 on Build GRCh38
rs759047033 Chr.22: 36929362-36929362 on Build GRCh38
rs762515992 Chr.22: 36929603-36929603 on Build GRCh38
rs761414020 Chr.22: 36929685-36929685 on Build GRCh38
rs780918629 Chr.22: 36929572-36929572 on Build GRCh38
rs140679574 Chr.22: 36929662-36929662 on Build GRCh38
rs771509643 Chr.22: 36929686-36929686 on Build GRCh38
rs781118627 Chr.22: 36929666-36929666 on Build GRCh38
rs758466490 Chr.22: 36929557-36929557 on Build GRCh38
rs367927682 Chr.22: 36929702-36929702 on Build GRCh38
rs780418241 Chr.22: 36929395-36929395 on Build GRCh38
rs774857861 Chr.22: 36929354-36929354 on Build GRCh38
rs372998096 Chr.22: 36929565-36929565 on Build GRCh38
rs1049607959 Chr.22: 36929220-36929220 on Build GRCh38
rs765958619 Chr.22: 36929357-36929357 on Build GRCh38
rs773833290 Chr.22: 36929611-36929611 on Build GRCh38
rs746797033 Chr.22: 36929428-36929428 on Build GRCh38
rs745658011 Chr.22: 36929667-36929667 on Build GRCh38
rs376405266 Chr.22: 36929390-36929390 on Build GRCh38
rs374827848 Chr.22: 36929683-36929683 on Build GRCh38
rs756516150 Chr.22: 36929512-36929512 on Build GRCh38
rs745663251 Chr.22: 36929591-36929591 on Build GRCh38
rs759942643 Chr.22: 36929693-36929693 on Build GRCh38
rs902433304 Chr.22: 36929590-36929590 on Build GRCh38
rs767022834 Chr.22: 36929619-36929619 on Build GRCh38
rs565413898 Chr.22: 36929509-36929509 on Build GRCh38
rs766898007 Chr.22: 36929524-36929524 on Build GRCh38
rs112325735 Chr.22: 36929648-36929648 on Build GRCh38
rs755084001 Chr.22: 36929401-36929401 on Build GRCh38
rs759831547 Chr.22: 36929632-36929632 on Build GRCh38

View Full Product Details