Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 83871302-83871838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYLC1
Entrez Gene ID
1538
Gene Name
cylicin 1
Gene Aliases
CYCL1
UniGene
Hs.444230
Gene Chromosome Location
Chr.X: 83861126-83886700 on Build GRCh38
Transcripts
NM_001271680.1 NM_021118.2 XM_005262086.3

SNP Detail

SNP ID Chromosome Location
rs898260448 Chr.X: 83871674-83871674 on Build GRCh38
rs757475394 Chr.X: 83871619-83871619 on Build GRCh38
rs930014123 Chr.X: 83871641-83871641 on Build GRCh38
rs185258122 Chr.X: 83871601-83871601 on Build GRCh38
rs757061367 Chr.X: 83871688-83871688 on Build GRCh38
rs751741613 Chr.X: 83871609-83871609 on Build GRCh38
rs775346829 Chr.X: 83871603-83871603 on Build GRCh38
rs746789514 Chr.X: 83871681-83871681 on Build GRCh38
rs770520850 Chr.X: 83871549-83871549 on Build GRCh38
rs368876224 Chr.X: 83871449-83871449 on Build GRCh38
rs375772815 Chr.X: 83871444-83871444 on Build GRCh38
rs759286065 Chr.X: 83871541-83871541 on Build GRCh38
rs753775166 Chr.X: 83871429-83871429 on Build GRCh38
rs922483205 Chr.X: 83871636-83871636 on Build GRCh38
rs771596731 Chr.X: 83871519-83871519 on Build GRCh38
rs909997018 Chr.X: 83871736-83871736 on Build GRCh38
rs191928176 Chr.X: 83871514-83871514 on Build GRCh38
rs995180048 Chr.X: 83871726-83871726 on Build GRCh38
rs1057147989 Chr.X: 83871614-83871614 on Build GRCh38
rs1019513078 Chr.X: 83871390-83871390 on Build GRCh38
rs140363451 Chr.X: 83871535-83871535 on Build GRCh38
rs770350213 Chr.X: 83871496-83871496 on Build GRCh38
rs1024475551 Chr.X: 83871347-83871347 on Build GRCh38
rs749514414 Chr.X: 83871581-83871582 on Build GRCh38
rs768934395 Chr.X: 83871590-83871590 on Build GRCh38
rs1014460687 Chr.X: 83871332-83871332 on Build GRCh38
rs754971866 Chr.X: 83871432-83871432 on Build GRCh38
rs759330900 Chr.X: 83871595-83871595 on Build GRCh38
rs939722181 Chr.X: 83871625-83871625 on Build GRCh38
rs757279410 Chr.X: 83871454-83871454 on Build GRCh38
rs746296402 Chr.X: 83871491-83871491 on Build GRCh38
rs765275569 Chr.X: 83871600-83871600 on Build GRCh38
rs953993195 Chr.X: 83871329-83871329 on Build GRCh38
rs781282634 Chr.X: 83871489-83871489 on Build GRCh38
rs749731888 Chr.X: 83871517-83871517 on Build GRCh38
rs966462650 Chr.X: 83871394-83871394 on Build GRCh38
rs773052157 Chr.X: 83871524-83871524 on Build GRCh38
rs758534227 Chr.X: 83871445-83871445 on Build GRCh38
rs764870173 Chr.X: 83871441-83871441 on Build GRCh38
rs868072234 Chr.X: 83871673-83871673 on Build GRCh38
rs941443572 Chr.X: 83871766-83871766 on Build GRCh38
rs894653959 Chr.X: 83871807-83871807 on Build GRCh38
rs375585605 Chr.X: 83871788-83871788 on Build GRCh38
rs776282972 Chr.X: 83871562-83871562 on Build GRCh38
rs777497473 Chr.X: 83871664-83871664 on Build GRCh38
rs987091201 Chr.X: 83871378-83871378 on Build GRCh38

View Full Product Details