Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46935365-46935860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4A11
Entrez Gene ID
1579
Gene Name
cytochrome P450 family 4 subfamily A member 11
Gene Aliases
CP4Y, CYP4A2, CYP4AII
UniGene
Hs.1645
Gene Chromosome Location
Chr.1: 46929174-46941484 on Build GRCh38
Transcripts
NM_000778.3 NM_001319155.1 NR_134988.1 NR_134989.1 NR_134990.1 NR_134991.1 NR_134992.1 NR_134993.1 NR_134994.1 XM_005270539.1 XM_017000464.1 XM_017000465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
140358 c.532G>T p.G178C Chr.1: 46935626-46935626 on Build GRCh38
117420 c.553G>T p.V185F Chr.1: 46935605-46935605 on Build GRCh38
118161 c.570C>T p.S190S Chr.1: 46935588-46935588 on Build GRCh38
3865701 c.545C>T p.P182L Chr.1: 46935613-46935613 on Build GRCh38
5669472 c.619A>G p.S207G Chr.1: 46935539-46935539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574055681 Chr.1: 46935656-46935656 on Build GRCh38
rs143018352 Chr.1: 46935633-46935633 on Build GRCh38
rs376943575 Chr.1: 46935573-46935573 on Build GRCh38
rs777489918 Chr.1: 46935582-46935582 on Build GRCh38
rs764147276 Chr.1: 46935616-46935616 on Build GRCh38
rs878902402 Chr.1: 46935795-46935795 on Build GRCh38
rs769649161 Chr.1: 46935748-46935748 on Build GRCh38
rs766773893 Chr.1: 46935674-46935674 on Build GRCh38
rs935405877 Chr.1: 46935829-46935829 on Build GRCh38
rs974945201 Chr.1: 46935785-46935785 on Build GRCh38
rs924023293 Chr.1: 46935825-46935825 on Build GRCh38
rs539428854 Chr.1: 46935527-46935527 on Build GRCh38
rs756378325 Chr.1: 46935666-46935666 on Build GRCh38
rs755679629 Chr.1: 46935707-46935733 on Build GRCh38
rs35763495 Chr.1: 46935810-46935810 on Build GRCh38
rs770751722 Chr.1: 46935589-46935589 on Build GRCh38
rs1023449092 Chr.1: 46935444-46935444 on Build GRCh38
rs9333006 Chr.1: 46935410-46935410 on Build GRCh38
rs577649417 Chr.1: 46935780-46935780 on Build GRCh38
rs768647899 Chr.1: 46935836-46935836 on Build GRCh38
rs551730881 Chr.1: 46935536-46935536 on Build GRCh38
rs754293457 Chr.1: 46935675-46935675 on Build GRCh38
rs561643197 Chr.1: 46935415-46935415 on Build GRCh38
rs34497654 Chr.1: 46935776-46935776 on Build GRCh38
rs902501132 Chr.1: 46935394-46935394 on Build GRCh38
rs571502120 Chr.1: 46935798-46935798 on Build GRCh38
rs2269231 Chr.1: 46935752-46935752 on Build GRCh38
rs777392421 Chr.1: 46935681-46935681 on Build GRCh38
rs756859338 Chr.1: 46935692-46935692 on Build GRCh38
rs199768508 Chr.1: 46935472-46935472 on Build GRCh38
rs577613992 Chr.1: 46935388-46935388 on Build GRCh38
rs755563711 Chr.1: 46935676-46935676 on Build GRCh38
rs34710862 Chr.1: 46935727-46935728 on Build GRCh38
rs759491535 Chr.1: 46935530-46935530 on Build GRCh38
rs776305178 Chr.1: 46935557-46935557 on Build GRCh38
rs370325040 Chr.1: 46935651-46935651 on Build GRCh38
rs749817925 Chr.1: 46935568-46935568 on Build GRCh38
rs767846632 Chr.1: 46935555-46935555 on Build GRCh38
rs780712696 Chr.1: 46935493-46935493 on Build GRCh38
rs535169313 Chr.1: 46935735-46935735 on Build GRCh38
rs746361673 Chr.1: 46935482-46935482 on Build GRCh38
rs148545122 Chr.1: 46935541-46935541 on Build GRCh38
rs140345183 Chr.1: 46935593-46935593 on Build GRCh38
rs780943219 Chr.1: 46935696-46935696 on Build GRCh38
rs143503396 Chr.1: 46935559-46935559 on Build GRCh38
rs549527801 Chr.1: 46935411-46935411 on Build GRCh38
rs545335072 Chr.1: 46935404-46935404 on Build GRCh38
rs563624307 Chr.1: 46935405-46935405 on Build GRCh38
rs757553372 Chr.1: 46935474-46935474 on Build GRCh38
rs769652273 Chr.1: 46935595-46935595 on Build GRCh38
rs747575936 Chr.1: 46935507-46935507 on Build GRCh38
rs868672389 Chr.1: 46935538-46935538 on Build GRCh38
rs775527556 Chr.1: 46935537-46935537 on Build GRCh38
rs745608540 Chr.1: 46935594-46935594 on Build GRCh38
rs544981813 Chr.1: 46935790-46935790 on Build GRCh38
rs62618709 Chr.1: 46935605-46935605 on Build GRCh38
rs780379911 Chr.1: 46935560-46935560 on Build GRCh38
rs200852244 Chr.1: 46935497-46935497 on Build GRCh38
rs536870781 Chr.1: 46935623-46935623 on Build GRCh38
rs995295558 Chr.1: 46935708-46935708 on Build GRCh38
rs746642707 Chr.1: 46935587-46935587 on Build GRCh38
rs12725784 Chr.1: 46935387-46935387 on Build GRCh38
rs770253753 Chr.1: 46935495-46935495 on Build GRCh38
rs542861156 Chr.1: 46935830-46935830 on Build GRCh38
rs2269232 Chr.1: 46935794-46935794 on Build GRCh38
rs1044948725 Chr.1: 46935659-46935659 on Build GRCh38
rs750669737 Chr.1: 46935653-46935653 on Build GRCh38
rs143061381 Chr.1: 46935540-46935540 on Build GRCh38
rs896368187 Chr.1: 46935430-46935430 on Build GRCh38
rs867076279 Chr.1: 46935704-46935704 on Build GRCh38
rs151044753 Chr.1: 46935542-46935542 on Build GRCh38
rs1025391467 Chr.1: 46935714-46935714 on Build GRCh38
rs144945108 Chr.1: 46935610-46935610 on Build GRCh38
rs528940696 Chr.1: 46935431-46935431 on Build GRCh38
rs565726317 Chr.1: 46935510-46935510 on Build GRCh38
rs1012496266 Chr.1: 46935443-46935443 on Build GRCh38
rs760405439 Chr.1: 46935520-46935520 on Build GRCh38
rs373527802 Chr.1: 46935519-46935519 on Build GRCh38
rs757534980 Chr.1: 46935544-46935544 on Build GRCh38
rs746554456 Chr.1: 46935688-46935688 on Build GRCh38

View Full Product Details