Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70325199-70325700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX19B
Entrez Gene ID
11269
Gene Name
DEAD-box helicase 19B
Gene Aliases
DBP5, DDX19, RNAh
UniGene
Hs.221761
Gene Chromosome Location
Chr.16: 70289767-70335213 on Build GRCh38
Transcripts
NM_001014449.2 NM_001014451.2 NM_001257172.1 NM_001257173.1 NM_001257174.1 NM_001257175.1 NM_007242.5 XM_006721127.2 XM_011522832.2 XM_011522833.2 XM_011522834.2 XM_017022890.1 NR_039997.1

Gene Symbol
LOC100506083
Entrez Gene ID
100506083
Gene Name
uncharacterized LOC100506083
Gene Aliases
-
UniGene
Hs.635008
Gene Chromosome Location
Chr.16: 70315640-70346747 on Build GRCh38
Transcripts
NM_001014449.2 NM_001014451.2 NM_001257172.1 NM_001257173.1 NM_001257174.1 NM_001257175.1 NM_007242.5 XM_006721127.2 XM_011522832.2 XM_011522833.2 XM_011522834.2 XM_017022890.1 NR_039997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4062328 c.523G>A p.A175T Chr.16: 70325604-70325604 on Build GRCh38
4062326 c.512C>T p.T171M Chr.16: 70325593-70325593 on Build GRCh38
3782982 c.599G>A p.G200D Chr.16: 70325680-70325680 on Build GRCh38
6344487 c.607+5G>T p.? Chr.16: 70325693-70325693 on Build GRCh38
3691309 c.595C>T p.R199* Chr.16: 70325676-70325676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140554740 Chr.16: 70325616-70325616 on Build GRCh38
rs549447766 Chr.16: 70325415-70325415 on Build GRCh38
rs939022853 Chr.16: 70325390-70325390 on Build GRCh38
rs541720606 Chr.16: 70325288-70325288 on Build GRCh38
rs1016512766 Chr.16: 70325492-70325492 on Build GRCh38
rs756280078 Chr.16: 70325622-70325622 on Build GRCh38
rs779949295 Chr.16: 70325665-70325665 on Build GRCh38
rs1008911119 Chr.16: 70325609-70325609 on Build GRCh38
rs777893383 Chr.16: 70325626-70325626 on Build GRCh38
rs879553911 Chr.16: 70325269-70325269 on Build GRCh38
rs1055177066 Chr.16: 70325443-70325443 on Build GRCh38
rs530906449 Chr.16: 70325356-70325356 on Build GRCh38
rs762357250 Chr.16: 70325597-70325597 on Build GRCh38
rs899365515 Chr.16: 70325505-70325505 on Build GRCh38
rs939357360 Chr.16: 70325223-70325223 on Build GRCh38
rs894741098 Chr.16: 70325446-70325446 on Build GRCh38
rs777160984 Chr.16: 70325555-70325555 on Build GRCh38
rs767204262 Chr.16: 70325628-70325628 on Build GRCh38
rs1038475951 Chr.16: 70325299-70325299 on Build GRCh38
rs1018049283 Chr.16: 70325657-70325657 on Build GRCh38
rs967819207 Chr.16: 70325509-70325509 on Build GRCh38
rs751106898 Chr.16: 70325604-70325604 on Build GRCh38
rs372080640 Chr.16: 70325544-70325544 on Build GRCh38
rs752890711 Chr.16: 70325552-70325552 on Build GRCh38
rs1008547740 Chr.16: 70325420-70325420 on Build GRCh38
rs776132389 Chr.16: 70325592-70325592 on Build GRCh38
rs1026400851 Chr.16: 70325575-70325575 on Build GRCh38
rs563807930 Chr.16: 70325345-70325345 on Build GRCh38
rs749257884 Chr.16: 70325667-70325667 on Build GRCh38
rs555637154 Chr.16: 70325247-70325247 on Build GRCh38
rs992605289 Chr.16: 70325238-70325238 on Build GRCh38
rs562007663 Chr.16: 70325419-70325419 on Build GRCh38
rs543332261 Chr.16: 70325478-70325478 on Build GRCh38
rs574291903 Chr.16: 70325280-70325280 on Build GRCh38
rs375444457 Chr.16: 70325671-70325671 on Build GRCh38
rs930144708 Chr.16: 70325330-70325330 on Build GRCh38
rs760953575 Chr.16: 70325549-70325549 on Build GRCh38
rs900191593 Chr.16: 70325469-70325469 on Build GRCh38
rs111815770 Chr.16: 70325669-70325669 on Build GRCh38
rs578141004 Chr.16: 70325321-70325321 on Build GRCh38
rs765811423 Chr.16: 70325588-70325588 on Build GRCh38
rs150416272 Chr.16: 70325654-70325654 on Build GRCh38
rs529138715 Chr.16: 70325503-70325503 on Build GRCh38
rs891411733 Chr.16: 70325343-70325343 on Build GRCh38
rs1022001742 Chr.16: 70325498-70325498 on Build GRCh38
rs772247442 Chr.16: 70325535-70325535 on Build GRCh38
rs767678734 Chr.16: 70325461-70325461 on Build GRCh38
rs150883787 Chr.16: 70325336-70325336 on Build GRCh38
rs559643559 Chr.16: 70325308-70325308 on Build GRCh38
rs995859922 Chr.16: 70325485-70325485 on Build GRCh38
rs763676230 Chr.16: 70325600-70325600 on Build GRCh38
rs760031392 Chr.16: 70325558-70325558 on Build GRCh38
rs529449578 Chr.16: 70325334-70325334 on Build GRCh38
rs945212351 Chr.16: 70325287-70325287 on Build GRCh38

View Full Product Details