Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 36284533-36285064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTHD1
Entrez Gene ID
401124
Gene Name
death domain containing 1
Gene Aliases
-
UniGene
Hs.363407
Gene Chromosome Location
Chr.4: 36281593-36347511 on Build GRCh38
Transcripts
NM_001136536.4 NM_001170700.2 XM_006714014.3 XM_011513693.2 XM_011513694.2 XM_011513695.2 XM_011513696.2 XM_017008191.1 XM_017008192.1 XM_017008193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6365776 c.510G>T p.K170N Chr.4: 36284589-36284589 on Build GRCh38
7318977 c.495T>C p.N165N Chr.4: 36284574-36284574 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs963851805 Chr.4: 36285029-36285029 on Build GRCh38
rs1018489319 Chr.4: 36284685-36284685 on Build GRCh38
rs1030607968 Chr.4: 36284969-36284969 on Build GRCh38
rs922294209 Chr.4: 36284735-36284735 on Build GRCh38
rs375425907 Chr.4: 36285006-36285006 on Build GRCh38
rs539474057 Chr.4: 36284621-36284621 on Build GRCh38
rs1046450738 Chr.4: 36284956-36284956 on Build GRCh38
rs1052355420 Chr.4: 36284954-36284954 on Build GRCh38
rs1030138971 Chr.4: 36284617-36284617 on Build GRCh38
rs748842866 Chr.4: 36284961-36284961 on Build GRCh38
rs936885476 Chr.4: 36284844-36284844 on Build GRCh38
rs780441183 Chr.4: 36284968-36284968 on Build GRCh38
rs868043239 Chr.4: 36285035-36285035 on Build GRCh38
rs888376664 Chr.4: 36284985-36284985 on Build GRCh38
rs956587496 Chr.4: 36284580-36284580 on Build GRCh38
rs73117663 Chr.4: 36284605-36284605 on Build GRCh38
rs553296725 Chr.4: 36284838-36284838 on Build GRCh38
rs779461866 Chr.4: 36284994-36284994 on Build GRCh38
rs574780779 Chr.4: 36284880-36284880 on Build GRCh38
rs1025571782 Chr.4: 36284655-36284655 on Build GRCh38
rs557492738 Chr.4: 36284656-36284656 on Build GRCh38
rs755749715 Chr.4: 36284720-36284720 on Build GRCh38
rs868175646 Chr.4: 36284859-36284859 on Build GRCh38
rs566100344 Chr.4: 36284706-36284706 on Build GRCh38
rs895240671 Chr.4: 36284590-36284590 on Build GRCh38
rs745479579 Chr.4: 36284616-36284616 on Build GRCh38
rs778344672 Chr.4: 36284609-36284609 on Build GRCh38
rs977469348 Chr.4: 36284829-36284829 on Build GRCh38
rs971690831 Chr.4: 36284751-36284751 on Build GRCh38
rs181566786 Chr.4: 36284796-36284796 on Build GRCh38
rs1005385222 Chr.4: 36284677-36284677 on Build GRCh38
rs918080849 Chr.4: 36284826-36284826 on Build GRCh38
rs753725419 Chr.4: 36284603-36284603 on Build GRCh38
rs1047466165 Chr.4: 36284975-36284975 on Build GRCh38
rs952713232 Chr.4: 36284667-36284667 on Build GRCh38
rs144824956 Chr.4: 36284999-36284999 on Build GRCh38
rs547399661 Chr.4: 36284813-36284813 on Build GRCh38
rs985885465 Chr.4: 36284651-36284651 on Build GRCh38
rs1030877579 Chr.4: 36284578-36284578 on Build GRCh38
rs779581581 Chr.4: 36284875-36284875 on Build GRCh38
rs756744728 Chr.4: 36284660-36284660 on Build GRCh38
rs948172788 Chr.4: 36284839-36284839 on Build GRCh38
rs930220385 Chr.4: 36284816-36284816 on Build GRCh38
rs766301788 Chr.4: 36284560-36284560 on Build GRCh38
rs916745013 Chr.4: 36284901-36284901 on Build GRCh38
rs997992799 Chr.4: 36285032-36285032 on Build GRCh38
rs936681380 Chr.4: 36284887-36284887 on Build GRCh38
rs963095843 Chr.4: 36285024-36285024 on Build GRCh38
rs994074170 Chr.4: 36284632-36284632 on Build GRCh38
rs1015970698 Chr.4: 36284693-36284693 on Build GRCh38
rs1000778156 Chr.4: 36284573-36284573 on Build GRCh38
rs992759812 Chr.4: 36284823-36284823 on Build GRCh38
rs867785502 Chr.4: 36284628-36284628 on Build GRCh38
rs1046421888 Chr.4: 36284607-36284607 on Build GRCh38

View Full Product Details