Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 183096802-183097351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP19
Entrez Gene ID
142679
Gene Name
dual specificity phosphatase 19
Gene Aliases
DUSP17, LMWDSP3, SKRP1, TS-DSP1
UniGene
Hs.132237
Gene Chromosome Location
Chr.2: 183078559-183100008 on Build GRCh38
Transcripts
NM_001142314.1 NM_001321519.1 NM_080876.3 NR_135688.1

SNP Detail

SNP ID Chromosome Location
rs745737895 Chr.2: 183097175-183097175 on Build GRCh38
rs34475351 Chr.2: 183096869-183096870 on Build GRCh38
rs371279686 Chr.2: 183097009-183097009 on Build GRCh38
rs905642546 Chr.2: 183097004-183097004 on Build GRCh38
rs536408631 Chr.2: 183096965-183096965 on Build GRCh38
rs942489603 Chr.2: 183097296-183097296 on Build GRCh38
rs990857581 Chr.2: 183096861-183096861 on Build GRCh38
rs942459772 Chr.2: 183096979-183096979 on Build GRCh38
rs994046374 Chr.2: 183097139-183097139 on Build GRCh38
rs765595639 Chr.2: 183097043-183097043 on Build GRCh38
rs979921685 Chr.2: 183097023-183097023 on Build GRCh38
rs566742364 Chr.2: 183097140-183097140 on Build GRCh38
rs4363996 Chr.2: 183097212-183097212 on Build GRCh38
rs1029425415 Chr.2: 183097166-183097166 on Build GRCh38
rs879163120 Chr.2: 183096862-183096862 on Build GRCh38
rs968199812 Chr.2: 183097230-183097230 on Build GRCh38
rs751562380 Chr.2: 183097319-183097319 on Build GRCh38
rs548999532 Chr.2: 183097029-183097030 on Build GRCh38
rs1044552607 Chr.2: 183096999-183096999 on Build GRCh38
rs763257807 Chr.2: 183096990-183096995 on Build GRCh38
rs911278507 Chr.2: 183097195-183097195 on Build GRCh38
rs935530897 Chr.2: 183097283-183097283 on Build GRCh38
rs974109178 Chr.2: 183097182-183097182 on Build GRCh38
rs1039456380 Chr.2: 183096986-183096986 on Build GRCh38
rs918523975 Chr.2: 183096915-183096915 on Build GRCh38
rs1035185675 Chr.2: 183097098-183097098 on Build GRCh38
rs902767947 Chr.2: 183097016-183097016 on Build GRCh38
rs553306019 Chr.2: 183097097-183097097 on Build GRCh38
rs558306157 Chr.2: 183097275-183097275 on Build GRCh38
rs150141445 Chr.2: 183097174-183097174 on Build GRCh38
rs192136678 Chr.2: 183096908-183096908 on Build GRCh38
rs918542492 Chr.2: 183097268-183097268 on Build GRCh38
rs758612082 Chr.2: 183096981-183096981 on Build GRCh38
rs2675069 Chr.2: 183096922-183096922 on Build GRCh38
rs916598876 Chr.2: 183096876-183096876 on Build GRCh38
rs963088723 Chr.2: 183097180-183097180 on Build GRCh38
rs975218394 Chr.2: 183097313-183097313 on Build GRCh38
rs183208873 Chr.2: 183097276-183097276 on Build GRCh38
rs912427555 Chr.2: 183097292-183097292 on Build GRCh38
rs999732476 Chr.2: 183097032-183097032 on Build GRCh38
rs985338404 Chr.2: 183097177-183097177 on Build GRCh38
rs755206837 Chr.2: 183096914-183096914 on Build GRCh38
rs1011256970 Chr.2: 183097006-183097006 on Build GRCh38
rs34201127 Chr.2: 183097042-183097042 on Build GRCh38
rs764293383 Chr.2: 183097029-183097029 on Build GRCh38
rs138566038 Chr.2: 183097320-183097320 on Build GRCh38
rs985965003 Chr.2: 183096907-183096907 on Build GRCh38
rs892815127 Chr.2: 183096989-183096989 on Build GRCh38
rs201935643 Chr.2: 183096833-183096833 on Build GRCh38
rs896660063 Chr.2: 183096994-183096997 on Build GRCh38
rs770148384 Chr.2: 183096905-183096905 on Build GRCh38
rs370148048 Chr.2: 183097303-183097303 on Build GRCh38
rs6759305 Chr.2: 183097075-183097075 on Build GRCh38
rs1013981891 Chr.2: 183096995-183096995 on Build GRCh38
rs573186521 Chr.2: 183096852-183096852 on Build GRCh38
rs912396243 Chr.2: 183096966-183096966 on Build GRCh38
rs1056612327 Chr.2: 183096982-183096982 on Build GRCh38
rs2368335 Chr.2: 183096940-183096940 on Build GRCh38
rs1016596792 Chr.2: 183097019-183097023 on Build GRCh38
rs571600087 Chr.2: 183096853-183096853 on Build GRCh38
rs960848723 Chr.2: 183097163-183097163 on Build GRCh38

View Full Product Details