Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57153532-57154076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUXA
Entrez Gene ID
503835
Gene Name
double homeobox A
Gene Aliases
-
UniGene
Hs.585857
Gene Chromosome Location
Chr.19: 57151726-57167488 on Build GRCh38
Transcripts
NM_001012729.1

SNP Detail

SNP ID Chromosome Location
rs931700205 Chr.19: 57153938-57153938 on Build GRCh38
rs945891766 Chr.19: 57153949-57153949 on Build GRCh38
rs1000149076 Chr.19: 57153750-57153750 on Build GRCh38
rs1014784666 Chr.19: 57154029-57154029 on Build GRCh38
rs202000984 Chr.19: 57153887-57153887 on Build GRCh38
rs1037497558 Chr.19: 57153598-57153598 on Build GRCh38
rs906837402 Chr.19: 57153768-57153768 on Build GRCh38
rs1010591330 Chr.19: 57153706-57153706 on Build GRCh38
rs530232823 Chr.19: 57153739-57153739 on Build GRCh38
rs1047292943 Chr.19: 57153582-57153582 on Build GRCh38
rs1050995134 Chr.19: 57153614-57153614 on Build GRCh38
rs904564537 Chr.19: 57153742-57153742 on Build GRCh38
rs763545262 Chr.19: 57153885-57153885 on Build GRCh38
rs1052006819 Chr.19: 57153667-57153667 on Build GRCh38
rs958903757 Chr.19: 57153557-57153557 on Build GRCh38
rs954700116 Chr.19: 57153899-57153899 on Build GRCh38
rs1053233607 Chr.19: 57154031-57154031 on Build GRCh38
rs907569324 Chr.19: 57153584-57153584 on Build GRCh38
rs539976095 Chr.19: 57153981-57153981 on Build GRCh38
rs553532050 Chr.19: 57154021-57154021 on Build GRCh38
rs898234693 Chr.19: 57153687-57153687 on Build GRCh38
rs549484286 Chr.19: 57153902-57153902 on Build GRCh38
rs1024386300 Chr.19: 57154047-57154047 on Build GRCh38
rs868426238 Chr.19: 57153808-57153808 on Build GRCh38
rs1000158882 Chr.19: 57153978-57153978 on Build GRCh38
rs10416425 Chr.19: 57153977-57153977 on Build GRCh38
rs114702643 Chr.19: 57153821-57153821 on Build GRCh38
rs948545676 Chr.19: 57153980-57153980 on Build GRCh38
rs749221536 Chr.19: 57153695-57153695 on Build GRCh38
rs114163951 Chr.19: 57153581-57153581 on Build GRCh38
rs1044115317 Chr.19: 57153995-57153995 on Build GRCh38
rs184307856 Chr.19: 57153898-57153898 on Build GRCh38
rs951566577 Chr.19: 57153882-57153882 on Build GRCh38
rs1021779288 Chr.19: 57153883-57153883 on Build GRCh38
rs921695291 Chr.19: 57153934-57153934 on Build GRCh38
rs1051502612 Chr.19: 57153939-57153939 on Build GRCh38
rs1020184809 Chr.19: 57153728-57153728 on Build GRCh38
rs996019732 Chr.19: 57153651-57153651 on Build GRCh38
rs567082571 Chr.19: 57153829-57153829 on Build GRCh38
rs773539734 Chr.19: 57153872-57153872 on Build GRCh38
rs906946640 Chr.19: 57154006-57154006 on Build GRCh38
rs565133094 Chr.19: 57153565-57153565 on Build GRCh38
rs1024918211 Chr.19: 57153852-57153852 on Build GRCh38
rs1034399374 Chr.19: 57153761-57153761 on Build GRCh38
rs191320556 Chr.19: 57153842-57153842 on Build GRCh38
rs35665114 Chr.19: 57153887-57153887 on Build GRCh38
rs1041656305 Chr.19: 57153953-57153953 on Build GRCh38
rs908447916 Chr.19: 57153925-57153925 on Build GRCh38
rs1002958833 Chr.19: 57153783-57153783 on Build GRCh38
rs567732450 Chr.19: 57153653-57153653 on Build GRCh38
rs939832579 Chr.19: 57153639-57153639 on Build GRCh38
rs1012693918 Chr.19: 57153736-57153736 on Build GRCh38
rs917515031 Chr.19: 57153569-57153569 on Build GRCh38
rs890395390 Chr.19: 57153689-57153689 on Build GRCh38
rs146529276 Chr.19: 57153948-57153948 on Build GRCh38
rs780054501 Chr.19: 57153592-57153592 on Build GRCh38
rs11882189 Chr.19: 57153905-57153905 on Build GRCh38
rs189149771 Chr.19: 57153587-57153587 on Build GRCh38
rs974775641 Chr.19: 57153889-57153889 on Build GRCh38
rs570038545 Chr.19: 57153637-57153637 on Build GRCh38
rs554167571 Chr.19: 57153959-57153959 on Build GRCh38

View Full Product Details