Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 103115141-103115522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC2H1
Entrez Gene ID
79659
Gene Name
dynein cytoplasmic 2 heavy chain 1
Gene Aliases
ATD3, DHC1b, DHC2, DNCH2, DYH1B, SRPS2B, SRTD3, hdhc11
UniGene
Hs.503721
Gene Chromosome Location
Chr.11: 103109431-103479863 on Build GRCh38
Transcripts
NM_001080463.1 NM_001377.2 XM_006718903.2 XM_017018291.1 XM_017018292.1 XM_017018293.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4679819 c.503G>A p.G168D Chr.11: 103115177-103115177 on Build GRCh38
922337 c.598G>T p.E200* Chr.11: 103115272-103115272 on Build GRCh38
200024 c.520G>A p.D174N Chr.11: 103115194-103115194 on Build GRCh38
5341367 c.554G>A p.R185H Chr.11: 103115228-103115228 on Build GRCh38
4017310 c.620G>T p.R207I Chr.11: 103115294-103115294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777097742 Chr.11: 103115255-103115255 on Build GRCh38
rs901694037 Chr.11: 103115397-103115397 on Build GRCh38
rs759256438 Chr.11: 103115492-103115492 on Build GRCh38
rs939511073 Chr.11: 103115213-103115213 on Build GRCh38
rs964198131 Chr.11: 103115252-103115252 on Build GRCh38
rs574334723 Chr.11: 103115440-103115440 on Build GRCh38
rs35318378 Chr.11: 103115215-103115216 on Build GRCh38
rs781152098 Chr.11: 103115183-103115183 on Build GRCh38
rs1046768712 Chr.11: 103115313-103115313 on Build GRCh38
rs772844062 Chr.11: 103115265-103115265 on Build GRCh38
rs555291565 Chr.11: 103115410-103115410 on Build GRCh38
rs772243087 Chr.11: 103115451-103115451 on Build GRCh38
rs775008223 Chr.11: 103115304-103115304 on Build GRCh38
rs759095555 Chr.11: 103115177-103115177 on Build GRCh38
rs529740957 Chr.11: 103115232-103115232 on Build GRCh38
rs752637620 Chr.11: 103115387-103115387 on Build GRCh38
rs770473454 Chr.11: 103115266-103115266 on Build GRCh38
rs532267144 Chr.11: 103115318-103115318 on Build GRCh38
rs550735709 Chr.11: 103115182-103115182 on Build GRCh38
rs1008157528 Chr.11: 103115442-103115442 on Build GRCh38
rs964228869 Chr.11: 103115493-103115493 on Build GRCh38
rs566324506 Chr.11: 103115227-103115227 on Build GRCh38
rs962701652 Chr.11: 103115435-103115435 on Build GRCh38
rs1031029720 Chr.11: 103115436-103115436 on Build GRCh38
rs201567316 Chr.11: 103115258-103115258 on Build GRCh38
rs11225550 Chr.11: 103115437-103115437 on Build GRCh38
rs12226466 Chr.11: 103115415-103115415 on Build GRCh38
rs769294289 Chr.11: 103115262-103115265 on Build GRCh38
rs7932257 Chr.11: 103115323-103115323 on Build GRCh38
rs745983021 Chr.11: 103115218-103115218 on Build GRCh38
rs557070449 Chr.11: 103115444-103115444 on Build GRCh38
rs190830113 Chr.11: 103115403-103115403 on Build GRCh38
rs919696586 Chr.11: 103115289-103115289 on Build GRCh38
rs903975397 Chr.11: 103115412-103115412 on Build GRCh38
rs186288567 Chr.11: 103115324-103115324 on Build GRCh38
rs928273249 Chr.11: 103115314-103115314 on Build GRCh38
rs753678228 Chr.11: 103115336-103115336 on Build GRCh38
rs372939881 Chr.11: 103115214-103115214 on Build GRCh38
rs757134411 Chr.11: 103115194-103115194 on Build GRCh38
rs755872520 Chr.11: 103115184-103115184 on Build GRCh38
rs566776071 Chr.11: 103115281-103115281 on Build GRCh38
rs368561058 Chr.11: 103115174-103115174 on Build GRCh38
rs368933119 Chr.11: 103115277-103115277 on Build GRCh38
rs769891971 Chr.11: 103115359-103115359 on Build GRCh38
rs939542929 Chr.11: 103115480-103115480 on Build GRCh38
rs759173558 Chr.11: 103115288-103115288 on Build GRCh38
rs777555139 Chr.11: 103115192-103115192 on Build GRCh38
rs11225551 Chr.11: 103115443-103115443 on Build GRCh38
rs753800897 Chr.11: 103115193-103115193 on Build GRCh38
rs763419649 Chr.11: 103115273-103115273 on Build GRCh38
rs766020059 Chr.11: 103115166-103115166 on Build GRCh38
rs770481238 Chr.11: 103115426-103115426 on Build GRCh38
rs534999214 Chr.11: 103115399-103115399 on Build GRCh38
rs766992862 Chr.11: 103115296-103115296 on Build GRCh38
rs199960819 Chr.11: 103115228-103115228 on Build GRCh38
rs748058835 Chr.11: 103115186-103115186 on Build GRCh38
rs886047559 Chr.11: 103115213-103115213 on Build GRCh38

View Full Product Details