Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244969761-244970250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB2
Entrez Gene ID
84288
Gene Name
EF-hand calcium binding domain 2
Gene Aliases
CFAP200, DRC8
UniGene
Hs.134857
Gene Chromosome Location
Chr.1: 244969739-245125228 on Build GRCh38
Transcripts
NM_032328.3 XM_011544301.1 NR_111907.1

Gene Symbol
LOC101928068
Entrez Gene ID
101928068
Gene Name
uncharacterized LOC101928068
Gene Aliases
-
UniGene
Hs.134857
Gene Chromosome Location
Chr.1: 244965204-244970869 on Build GRCh38
Transcripts
NM_032328.3 XM_011544301.1 NR_111907.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4143605 c.65T>G p.V22G Chr.1: 244970187-244970187 on Build GRCh38
4143606 c.115A>G p.T39A Chr.1: 244970237-244970237 on Build GRCh38
4589958 c.126_127delGC p.L45fs*32 Chr.1: 244970248-244970249 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1030957129 Chr.1: 244970145-244970145 on Build GRCh38
rs1011952814 Chr.1: 244969926-244969926 on Build GRCh38
rs922747607 Chr.1: 244970001-244970001 on Build GRCh38
rs758458658 Chr.1: 244969813-244969813 on Build GRCh38
rs979263825 Chr.1: 244969792-244969792 on Build GRCh38
rs560898311 Chr.1: 244970030-244970030 on Build GRCh38
rs774437968 Chr.1: 244970173-244970173 on Build GRCh38
rs918023850 Chr.1: 244970098-244970098 on Build GRCh38
rs375439510 Chr.1: 244969812-244969812 on Build GRCh38
rs909905808 Chr.1: 244970038-244970038 on Build GRCh38
rs909871308 Chr.1: 244970202-244970202 on Build GRCh38
rs888666671 Chr.1: 244969836-244969836 on Build GRCh38
rs538966017 Chr.1: 244970088-244970088 on Build GRCh38
rs941494364 Chr.1: 244969887-244969887 on Build GRCh38
rs550418691 Chr.1: 244970020-244970020 on Build GRCh38
rs967918403 Chr.1: 244969932-244969932 on Build GRCh38
rs963075997 Chr.1: 244970228-244970228 on Build GRCh38
rs894860653 Chr.1: 244970056-244970056 on Build GRCh38
rs903794321 Chr.1: 244969976-244969976 on Build GRCh38
rs938219604 Chr.1: 244970092-244970092 on Build GRCh38
rs922576230 Chr.1: 244969797-244969797 on Build GRCh38
rs757874379 Chr.1: 244970051-244970051 on Build GRCh38
rs143358867 Chr.1: 244969839-244969839 on Build GRCh38
rs778809858 Chr.1: 244970041-244970041 on Build GRCh38
rs557180113 Chr.1: 244970151-244970151 on Build GRCh38
rs900329896 Chr.1: 244970168-244970168 on Build GRCh38
rs759547450 Chr.1: 244970183-244970183 on Build GRCh38
rs766825394 Chr.1: 244970141-244970141 on Build GRCh38
rs530600166 Chr.1: 244969943-244969943 on Build GRCh38
rs537514480 Chr.1: 244970139-244970139 on Build GRCh38
rs573243329 Chr.1: 244969866-244969866 on Build GRCh38
rs779579650 Chr.1: 244970079-244970079 on Build GRCh38
rs529660425 Chr.1: 244970042-244970042 on Build GRCh38
rs573179823 Chr.1: 244970222-244970222 on Build GRCh38
rs923758022 Chr.1: 244969965-244969965 on Build GRCh38
rs565165061 Chr.1: 244969924-244969924 on Build GRCh38
rs1034797545 Chr.1: 244969881-244969881 on Build GRCh38
rs983549678 Chr.1: 244970052-244970052 on Build GRCh38
rs1000744796 Chr.1: 244970165-244970165 on Build GRCh38
rs780690443 Chr.1: 244970208-244970208 on Build GRCh38
rs748514967 Chr.1: 244970114-244970114 on Build GRCh38
rs897333428 Chr.1: 244969869-244969869 on Build GRCh38
rs374870956 Chr.1: 244969789-244969789 on Build GRCh38
rs1000632475 Chr.1: 244970013-244970013 on Build GRCh38
rs767655461 Chr.1: 244969968-244969968 on Build GRCh38
rs1010007027 Chr.1: 244969964-244969964 on Build GRCh38
rs977239935 Chr.1: 244969979-244969979 on Build GRCh38
rs1004526424 Chr.1: 244970192-244970192 on Build GRCh38
rs768301304 Chr.1: 244970097-244970097 on Build GRCh38
rs997782810 Chr.1: 244970181-244970181 on Build GRCh38
rs776155626 Chr.1: 244970108-244970108 on Build GRCh38
rs941387809 Chr.1: 244970219-244970219 on Build GRCh38
rs1056986525 Chr.1: 244969935-244969935 on Build GRCh38
rs985524124 Chr.1: 244970201-244970201 on Build GRCh38
rs975909496 Chr.1: 244970175-244970175 on Build GRCh38
rs1038560207 Chr.1: 244969903-244969903 on Build GRCh38
rs371167848 Chr.1: 244970188-244970188 on Build GRCh38
rs552376569 Chr.1: 244970096-244970096 on Build GRCh38
rs568975131 Chr.1: 244970136-244970136 on Build GRCh38
rs773831150 Chr.1: 244970118-244970118 on Build GRCh38
rs552984249 Chr.1: 244970210-244970210 on Build GRCh38
rs546662742 Chr.1: 244970045-244970045 on Build GRCh38
rs755186356 Chr.1: 244969957-244969957 on Build GRCh38
rs770336509 Chr.1: 244970116-244970116 on Build GRCh38
rs1015938128 Chr.1: 244970213-244970213 on Build GRCh38
rs1003689015 Chr.1: 244969880-244969880 on Build GRCh38
rs932748752 Chr.1: 244970012-244970012 on Build GRCh38
rs1012005011 Chr.1: 244970070-244970070 on Build GRCh38
rs753484664 Chr.1: 244970043-244970045 on Build GRCh38
rs903550975 Chr.1: 244970127-244970127 on Build GRCh38
rs976113875 Chr.1: 244969822-244969822 on Build GRCh38
rs959293174 Chr.1: 244969889-244969889 on Build GRCh38
rs766596007 Chr.1: 244969831-244969831 on Build GRCh38
rs548539510 Chr.1: 244969845-244969866 on Build GRCh38
rs750507322 Chr.1: 244969794-244969794 on Build GRCh38
rs1038961066 Chr.1: 244970166-244970166 on Build GRCh38
rs945368482 Chr.1: 244970126-244970126 on Build GRCh38
rs765058175 Chr.1: 244970227-244970227 on Build GRCh38
rs757111908 Chr.1: 244970036-244970036 on Build GRCh38
rs191269150 Chr.1: 244969877-244969877 on Build GRCh38
rs897294551 Chr.1: 244970049-244970049 on Build GRCh38
rs1021975822 Chr.1: 244969928-244969928 on Build GRCh38
rs1037139447 Chr.1: 244969861-244969861 on Build GRCh38
rs983051295 Chr.1: 244969868-244969868 on Build GRCh38
rs566761007 Chr.1: 244970073-244970073 on Build GRCh38
rs962972234 Chr.1: 244970086-244970086 on Build GRCh38
rs372434795 Chr.1: 244970050-244970050 on Build GRCh38
rs7545078 Chr.1: 244970187-244970187 on Build GRCh38

View Full Product Details