Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23360560-23361069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFS
Entrez Gene ID
10278
Gene Name
embryonal Fyn-associated substrate
Gene Aliases
CAS3, CASS3, EFS1, EFS2, HEFS, SIN
UniGene
Hs.24587
Gene Chromosome Location
Chr.14: 23356400-23365633 on Build GRCh38
Transcripts
NM_001277174.1 NM_005864.3 NM_032459.2 XM_005267256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7247849 c.103C>T p.L35L Chr.14: 23360749-23360749 on Build GRCh38
5587705 c.137G>A p.W46* Chr.14: 23360715-23360715 on Build GRCh38
6396213 c.106C>T p.Q36* Chr.14: 23360746-23360746 on Build GRCh38
5606508 c.78C>T p.F26F Chr.14: 23360774-23360774 on Build GRCh38
3495140 c.144C>T p.L48L Chr.14: 23360708-23360708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768106724 Chr.14: 23360738-23360738 on Build GRCh38
rs537555000 Chr.14: 23360666-23360666 on Build GRCh38
rs750247067 Chr.14: 23360684-23360684 on Build GRCh38
rs778056626 Chr.14: 23360675-23360675 on Build GRCh38
rs926366419 Chr.14: 23361045-23361045 on Build GRCh38
rs865948821 Chr.14: 23360702-23360702 on Build GRCh38
rs1052070471 Chr.14: 23360767-23360767 on Build GRCh38
rs776081551 Chr.14: 23360818-23360818 on Build GRCh38
rs975360586 Chr.14: 23360582-23360582 on Build GRCh38
rs918092361 Chr.14: 23360701-23360701 on Build GRCh38
rs767158899 Chr.14: 23360846-23360846 on Build GRCh38
rs747569514 Chr.14: 23360817-23360822 on Build GRCh38
rs566332564 Chr.14: 23360622-23360622 on Build GRCh38
rs367826175 Chr.14: 23360863-23360863 on Build GRCh38
rs2231798 Chr.14: 23360833-23360833 on Build GRCh38
rs959988036 Chr.14: 23360915-23360915 on Build GRCh38
rs1026926731 Chr.14: 23360862-23360862 on Build GRCh38
rs764268978 Chr.14: 23360821-23360821 on Build GRCh38
rs200734043 Chr.14: 23360801-23360801 on Build GRCh38
rs752787769 Chr.14: 23360742-23360742 on Build GRCh38
rs149177500 Chr.14: 23360744-23360744 on Build GRCh38
rs375046962 Chr.14: 23360720-23360720 on Build GRCh38
rs756312267 Chr.14: 23360616-23360616 on Build GRCh38
rs183593168 Chr.14: 23360919-23360919 on Build GRCh38
rs981009183 Chr.14: 23360995-23360995 on Build GRCh38
rs535021372 Chr.14: 23360637-23360637 on Build GRCh38
rs571494366 Chr.14: 23360963-23360963 on Build GRCh38
rs780275370 Chr.14: 23360714-23360721 on Build GRCh38
rs762453443 Chr.14: 23360731-23360731 on Build GRCh38
rs188466425 Chr.14: 23360669-23360669 on Build GRCh38
rs918079992 Chr.14: 23360952-23360952 on Build GRCh38
rs747668326 Chr.14: 23360772-23360772 on Build GRCh38
rs777393805 Chr.14: 23360853-23360853 on Build GRCh38
rs563397066 Chr.14: 23360887-23360887 on Build GRCh38
rs769399321 Chr.14: 23360723-23360723 on Build GRCh38
rs772905581 Chr.14: 23360724-23360724 on Build GRCh38
rs199828925 Chr.14: 23360674-23360674 on Build GRCh38
rs746894866 Chr.14: 23360641-23360641 on Build GRCh38
rs760082284 Chr.14: 23360673-23360673 on Build GRCh38
rs1007939844 Chr.14: 23360817-23360817 on Build GRCh38
rs551830253 Chr.14: 23360930-23360930 on Build GRCh38
rs752123181 Chr.14: 23360670-23360670 on Build GRCh38
rs755094116 Chr.14: 23360601-23360601 on Build GRCh38
rs779760788 Chr.14: 23360696-23360696 on Build GRCh38
rs768529687 Chr.14: 23360712-23360712 on Build GRCh38
rs949579821 Chr.14: 23360703-23360703 on Build GRCh38
rs191828624 Chr.14: 23360918-23360918 on Build GRCh38
rs982666847 Chr.14: 23360886-23360886 on Build GRCh38
rs746231709 Chr.14: 23360800-23360800 on Build GRCh38
rs747555440 Chr.14: 23360820-23360820 on Build GRCh38
rs993600542 Chr.14: 23360764-23360764 on Build GRCh38
rs759753072 Chr.14: 23360988-23360988 on Build GRCh38
rs780726112 Chr.14: 23360719-23360719 on Build GRCh38
rs74413096 Chr.14: 23360653-23360653 on Build GRCh38
rs754754512 Chr.14: 23360845-23360845 on Build GRCh38
rs373577270 Chr.14: 23360773-23360773 on Build GRCh38
rs755964602 Chr.14: 23360852-23360852 on Build GRCh38
rs948944834 Chr.14: 23360990-23360990 on Build GRCh38
rs141340036 Chr.14: 23360615-23360615 on Build GRCh38
rs571795917 Chr.14: 23360654-23360654 on Build GRCh38
rs904761706 Chr.14: 23360750-23360750 on Build GRCh38
rs763371688 Chr.14: 23360658-23360658 on Build GRCh38
rs542572292 Chr.14: 23360910-23360910 on Build GRCh38
rs550783926 Chr.14: 23360992-23360992 on Build GRCh38
rs934927079 Chr.14: 23361030-23361030 on Build GRCh38
rs749998161 Chr.14: 23360615-23360616 on Build GRCh38
rs926834300 Chr.14: 23361002-23361002 on Build GRCh38
rs777078722 Chr.14: 23360774-23360774 on Build GRCh38
rs774808490 Chr.14: 23360735-23360735 on Build GRCh38
rs770687956 Chr.14: 23360787-23360787 on Build GRCh38
rs765508825 Chr.14: 23361046-23361046 on Build GRCh38
rs752682270 Chr.14: 23360848-23360848 on Build GRCh38
rs372971469 Chr.14: 23360695-23360695 on Build GRCh38
rs745645493 Chr.14: 23360631-23360631 on Build GRCh38
rs145243595 Chr.14: 23360657-23360657 on Build GRCh38
rs200301658 Chr.14: 23360734-23360734 on Build GRCh38
rs754943844 Chr.14: 23360756-23360756 on Build GRCh38
rs529016504 Chr.14: 23360905-23360905 on Build GRCh38
rs71413982 Chr.14: 23360592-23360592 on Build GRCh38
rs372118594 Chr.14: 23360838-23360838 on Build GRCh38
rs751418618 Chr.14: 23360841-23360841 on Build GRCh38
rs758835459 Chr.14: 23360706-23360713 on Build GRCh38
rs748778404 Chr.14: 23360859-23360859 on Build GRCh38
rs764027598 Chr.14: 23360746-23360746 on Build GRCh38
rs531041222 Chr.14: 23360968-23360968 on Build GRCh38
rs992596065 Chr.14: 23360929-23360929 on Build GRCh38
rs776076766 Chr.14: 23360645-23360645 on Build GRCh38
rs2231797 Chr.14: 23360839-23360839 on Build GRCh38
rs768161511 Chr.14: 23360643-23360643 on Build GRCh38
rs781360690 Chr.14: 23360609-23360609 on Build GRCh38
rs2231800 Chr.14: 23360749-23360749 on Build GRCh38
rs751330577 Chr.14: 23360755-23360755 on Build GRCh38
rs148307786 Chr.14: 23360752-23360752 on Build GRCh38
rs781181918 Chr.14: 23360770-23360770 on Build GRCh38
rs2231799 Chr.14: 23360786-23360786 on Build GRCh38
rs926121349 Chr.14: 23360996-23360996 on Build GRCh38
rs760172126 Chr.14: 23360737-23360737 on Build GRCh38
rs142212759 Chr.14: 23360754-23360754 on Build GRCh38
rs201669687 Chr.14: 23360793-23360793 on Build GRCh38

View Full Product Details