Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130065576-130066081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELF4
Entrez Gene ID
2000
Gene Name
E74 like ETS transcription factor 4
Gene Aliases
ELFR, MEF
UniGene
Hs.271940
Gene Chromosome Location
Chr.X: 130064920-130110713 on Build GRCh38
Transcripts
NM_001127197.1 NM_001421.3 XM_005262389.3 XM_011531307.2 XM_011531308.2

SNP Detail

SNP ID Chromosome Location
rs756323428 Chr.X: 130065934-130065934 on Build GRCh38
rs781237784 Chr.X: 130065956-130065956 on Build GRCh38
rs1000433516 Chr.X: 130065623-130065623 on Build GRCh38
rs138817013 Chr.X: 130065953-130065953 on Build GRCh38
rs149306204 Chr.X: 130066025-130066025 on Build GRCh38
rs936175017 Chr.X: 130066035-130066035 on Build GRCh38
rs762594443 Chr.X: 130065620-130065620 on Build GRCh38
rs369607337 Chr.X: 130066055-130066055 on Build GRCh38
rs757160271 Chr.X: 130066012-130066012 on Build GRCh38
rs1018473653 Chr.X: 130065634-130065634 on Build GRCh38
rs1014796742 Chr.X: 130065825-130065825 on Build GRCh38
rs145905263 Chr.X: 130065952-130065952 on Build GRCh38
rs3788847 Chr.X: 130065841-130065841 on Build GRCh38
rs548892706 Chr.X: 130065919-130065919 on Build GRCh38
rs775137425 Chr.X: 130065601-130065601 on Build GRCh38
rs914526805 Chr.X: 130065625-130065625 on Build GRCh38
rs746900885 Chr.X: 130066009-130066009 on Build GRCh38
rs3788848 Chr.X: 130065837-130065837 on Build GRCh38
rs983183160 Chr.X: 130065739-130065739 on Build GRCh38
rs933339555 Chr.X: 130066056-130066056 on Build GRCh38
rs946157756 Chr.X: 130066011-130066011 on Build GRCh38
rs1056495 Chr.X: 130065853-130065853 on Build GRCh38
rs937218834 Chr.X: 130065998-130065998 on Build GRCh38
rs1056497 Chr.X: 130065635-130065635 on Build GRCh38
rs904733484 Chr.X: 130066034-130066034 on Build GRCh38
rs112963685 Chr.X: 130065734-130065734 on Build GRCh38
rs764486372 Chr.X: 130065640-130065640 on Build GRCh38
rs755081220 Chr.X: 130065954-130065954 on Build GRCh38
rs995865046 Chr.X: 130065721-130065721 on Build GRCh38
rs899296976 Chr.X: 130065869-130065869 on Build GRCh38
rs187926577 Chr.X: 130065946-130065946 on Build GRCh38
rs1010707194 Chr.X: 130065617-130065617 on Build GRCh38
rs924626825 Chr.X: 130065733-130065733 on Build GRCh38
rs1052308583 Chr.X: 130065813-130065813 on Build GRCh38
rs947301027 Chr.X: 130065626-130065626 on Build GRCh38
rs958556638 Chr.X: 130065611-130065611 on Build GRCh38
rs750783246 Chr.X: 130065741-130065741 on Build GRCh38
rs778887977 Chr.X: 130065984-130065985 on Build GRCh38
rs994158688 Chr.X: 130065879-130065879 on Build GRCh38
rs1056496 Chr.X: 130065732-130065732 on Build GRCh38
rs963203022 Chr.X: 130065906-130065906 on Build GRCh38
rs112432023 Chr.X: 130065837-130065837 on Build GRCh38
rs900043983 Chr.X: 130065659-130065659 on Build GRCh38
rs954505304 Chr.X: 130065735-130065735 on Build GRCh38
rs376721120 Chr.X: 130066039-130066039 on Build GRCh38
rs774874594 Chr.X: 130065730-130065730 on Build GRCh38
rs751775069 Chr.X: 130065929-130065929 on Build GRCh38
rs569539488 Chr.X: 130065842-130065842 on Build GRCh38
rs1041886523 Chr.X: 130065619-130065619 on Build GRCh38
rs1056493 Chr.X: 130065866-130065866 on Build GRCh38
rs977752363 Chr.X: 130066026-130066026 on Build GRCh38
rs748170470 Chr.X: 130066004-130066004 on Build GRCh38

View Full Product Details